Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28692761-28693186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMEB1
Entrez Gene ID
10691
Gene Name
glucocorticoid modulatory element binding protein 1
Gene Aliases
P96PIF, PIF96
UniGene
Hs.632373
Gene Chromosome Location
Chr.1: 28668728-28715603 on Build GRCh38
Transcripts
NM_001319674.1 NM_006582.3 NM_024482.2 XM_011540518.2 XM_011540519.2 XM_011540521.2 XM_011540522.2 XM_017000087.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5846291 c.405G>A p.W135* Chr.1: 28693010-28693010 on Build GRCh38
1492063 c.337-2A>T p.? Chr.1: 28692940-28692940 on Build GRCh38
5846290 c.435G>A p.W145* Chr.1: 28693010-28693010 on Build GRCh38
5531755 c.346C>T p.Q116* Chr.1: 28692951-28692951 on Build GRCh38
3386073 c.419G>A p.R140H Chr.1: 28693024-28693024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010388742 Chr.1: 28692795-28692795 on Build GRCh38
rs972188782 Chr.1: 28693139-28693139 on Build GRCh38
rs773763070 Chr.1: 28692912-28692912 on Build GRCh38
rs932073476 Chr.1: 28692885-28692885 on Build GRCh38
rs1041988211 Chr.1: 28693115-28693115 on Build GRCh38
rs748701572 Chr.1: 28692894-28692894 on Build GRCh38
rs903359775 Chr.1: 28693129-28693129 on Build GRCh38
rs1000283385 Chr.1: 28693135-28693135 on Build GRCh38
rs760346353 Chr.1: 28692962-28692962 on Build GRCh38
rs765156729 Chr.1: 28693055-28693055 on Build GRCh38
rs944965934 Chr.1: 28693004-28693004 on Build GRCh38
rs760798386 Chr.1: 28693053-28693053 on Build GRCh38
rs770261873 Chr.1: 28692899-28692899 on Build GRCh38
rs529116936 Chr.1: 28692840-28692840 on Build GRCh38
rs763346584 Chr.1: 28692918-28692918 on Build GRCh38
rs953373783 Chr.1: 28693105-28693105 on Build GRCh38
rs751288921 Chr.1: 28693084-28693084 on Build GRCh38
rs199537620 Chr.1: 28692970-28692970 on Build GRCh38
rs549255792 Chr.1: 28693081-28693081 on Build GRCh38
rs771261047 Chr.1: 28692934-28692934 on Build GRCh38
rs372654787 Chr.1: 28692978-28692978 on Build GRCh38
rs758318475 Chr.1: 28693073-28693073 on Build GRCh38
rs1020841819 Chr.1: 28692807-28692807 on Build GRCh38
rs909281005 Chr.1: 28693119-28693119 on Build GRCh38
rs966253936 Chr.1: 28692834-28692834 on Build GRCh38
rs768304885 Chr.1: 28692893-28692893 on Build GRCh38
rs773698776 Chr.1: 28692892-28692892 on Build GRCh38
rs774575840 Chr.1: 28692940-28692940 on Build GRCh38
rs750356585 Chr.1: 28693072-28693072 on Build GRCh38
rs371221699 Chr.1: 28692890-28692890 on Build GRCh38
rs890476774 Chr.1: 28692938-28692938 on Build GRCh38
rs752665315 Chr.1: 28692995-28692995 on Build GRCh38

View Full Product Details