Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18790739-18791249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TVP23B
Entrez Gene ID
51030
Gene Name
trans-golgi network vesicle protein 23 homolog B (S. cerevisiae)
Gene Aliases
CGI-148, FAM18B, FAM18B1, NPD008, YDR084C
UniGene
Hs.87295
Gene Chromosome Location
Chr.17: 18780995-18806713 on Build GRCh38
Transcripts
NM_001316919.1 NM_001316920.1 NM_001316921.1 NM_001316922.1 NM_001316923.1 NM_001316924.1 NM_016078.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6052375 c.194T>C p.M65T Chr.17: 18790994-18790994 on Build GRCh38
4456735 c.101C>T p.P34L Chr.17: 18790901-18790901 on Build GRCh38
704785 c.132A>T p.R44R Chr.17: 18790932-18790932 on Build GRCh38
3773779 c.164G>A p.G55E Chr.17: 18790964-18790964 on Build GRCh38
4934302 c.108A>G p.A36A Chr.17: 18790908-18790908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774156429 Chr.17: 18791195-18791197 on Build GRCh38
rs750206855 Chr.17: 18791050-18791050 on Build GRCh38
rs868338859 Chr.17: 18791203-18791203 on Build GRCh38
rs542278269 Chr.17: 18791018-18791018 on Build GRCh38
rs368902602 Chr.17: 18791213-18791214 on Build GRCh38
rs1054218934 Chr.17: 18790789-18790789 on Build GRCh38
rs372868254 Chr.17: 18790930-18790930 on Build GRCh38
rs756925552 Chr.17: 18790849-18790849 on Build GRCh38
rs867017720 Chr.17: 18790808-18790808 on Build GRCh38
rs373922863 Chr.17: 18790998-18790998 on Build GRCh38
rs762889436 Chr.17: 18791187-18791188 on Build GRCh38
rs1047175852 Chr.17: 18790766-18790766 on Build GRCh38
rs770323752 Chr.17: 18790878-18790878 on Build GRCh38
rs749256585 Chr.17: 18790950-18790951 on Build GRCh38
rs773632533 Chr.17: 18790924-18790926 on Build GRCh38
rs771602402 Chr.17: 18790867-18790867 on Build GRCh38
rs765066245 Chr.17: 18791031-18791031 on Build GRCh38
rs774820840 Chr.17: 18790974-18790974 on Build GRCh38
rs764086111 Chr.17: 18790911-18790911 on Build GRCh38
rs767742179 Chr.17: 18790903-18790903 on Build GRCh38
rs774740959 Chr.17: 18790894-18790894 on Build GRCh38
rs779763962 Chr.17: 18791063-18791063 on Build GRCh38
rs553575250 Chr.17: 18790830-18790830 on Build GRCh38
rs200487006 Chr.17: 18791030-18791030 on Build GRCh38
rs894113454 Chr.17: 18790836-18790836 on Build GRCh38
rs1005729836 Chr.17: 18790811-18790811 on Build GRCh38
rs763383929 Chr.17: 18790888-18790888 on Build GRCh38
rs1021066603 Chr.17: 18791213-18791213 on Build GRCh38
rs778630125 Chr.17: 18790926-18790926 on Build GRCh38
rs780532883 Chr.17: 18791088-18791088 on Build GRCh38
rs941554566 Chr.17: 18791100-18791100 on Build GRCh38
rs573504806 Chr.17: 18790908-18790908 on Build GRCh38
rs764704426 Chr.17: 18791002-18791002 on Build GRCh38
rs61075345 Chr.17: 18790964-18790964 on Build GRCh38
rs757056737 Chr.17: 18790924-18790924 on Build GRCh38
rs759884022 Chr.17: 18790980-18790980 on Build GRCh38
rs758025427 Chr.17: 18790942-18790942 on Build GRCh38
rs752796580 Chr.17: 18790906-18790906 on Build GRCh38
rs564515928 Chr.17: 18791202-18791202 on Build GRCh38
rs771032877 Chr.17: 18791188-18791197 on Build GRCh38
rs867227651 Chr.17: 18791214-18791214 on Build GRCh38
rs922595500 Chr.17: 18791009-18791009 on Build GRCh38
rs773519045 Chr.17: 18790882-18790882 on Build GRCh38
rs746526546 Chr.17: 18790948-18790948 on Build GRCh38
rs533886367 Chr.17: 18790793-18790793 on Build GRCh38
rs573544934 Chr.17: 18791005-18791005 on Build GRCh38
rs1038643704 Chr.17: 18790835-18790835 on Build GRCh38
rs555827257 Chr.17: 18790954-18790954 on Build GRCh38
rs542524262 Chr.17: 18790952-18790952 on Build GRCh38
rs748653469 Chr.17: 18790877-18790877 on Build GRCh38
rs971102975 Chr.17: 18790929-18790929 on Build GRCh38
rs759567622 Chr.17: 18791086-18791086 on Build GRCh38
rs998610293 Chr.17: 18790931-18790931 on Build GRCh38
rs771479580 Chr.17: 18791008-18791010 on Build GRCh38
rs544233866 Chr.17: 18791066-18791066 on Build GRCh38
rs758115521 Chr.17: 18791059-18791059 on Build GRCh38
rs745465681 Chr.17: 18790852-18790852 on Build GRCh38
rs767836819 Chr.17: 18790986-18790986 on Build GRCh38
rs770162920 Chr.17: 18791194-18791197 on Build GRCh38
rs760812549 Chr.17: 18791006-18791006 on Build GRCh38
rs779490008 Chr.17: 18790947-18790947 on Build GRCh38
rs571211309 Chr.17: 18790778-18790778 on Build GRCh38
rs1010476613 Chr.17: 18791164-18791164 on Build GRCh38
rs887294147 Chr.17: 18790779-18790779 on Build GRCh38
rs774874745 Chr.17: 18791016-18791018 on Build GRCh38
rs529458986 Chr.17: 18791130-18791131 on Build GRCh38
rs1012624949 Chr.17: 18790861-18790861 on Build GRCh38
rs894762435 Chr.17: 18791131-18791131 on Build GRCh38
rs753835939 Chr.17: 18791019-18791019 on Build GRCh38
rs774950164 Chr.17: 18790876-18790876 on Build GRCh38
rs771456997 Chr.17: 18790966-18790966 on Build GRCh38
rs754541845 Chr.17: 18791071-18791071 on Build GRCh38
rs766796686 Chr.17: 18790893-18790893 on Build GRCh38
rs775497308 Chr.17: 18790987-18790987 on Build GRCh38
rs577682107 Chr.17: 18791216-18791216 on Build GRCh38

View Full Product Details