Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19313019-19313516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPN2
Entrez Gene ID
22905
Gene Name
epsin 2
Gene Aliases
EHB21
UniGene
Hs.743983
Gene Chromosome Location
Chr.17: 19237377-19336715 on Build GRCh38
Transcripts
NM_001102664.1 NM_014964.4 NM_148921.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4459154 c.1114C>T p.P372S Chr.17: 19313246-19313246 on Build GRCh38
417154 c.1115C>A p.P372H Chr.17: 19313247-19313247 on Build GRCh38
3515024 c.995C>T p.T332I Chr.17: 19313127-19313127 on Build GRCh38
6532413 c.1086G>C p.Q362H Chr.17: 19313218-19313218 on Build GRCh38
6655368 c.1133C>A p.P378H Chr.17: 19313265-19313265 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024550109 Chr.17: 19313163-19313163 on Build GRCh38
rs184341621 Chr.17: 19313274-19313274 on Build GRCh38
rs764469123 Chr.17: 19313180-19313180 on Build GRCh38
rs774036660 Chr.17: 19313152-19313152 on Build GRCh38
rs1046342109 Chr.17: 19313300-19313300 on Build GRCh38
rs751847236 Chr.17: 19313191-19313191 on Build GRCh38
rs965872724 Chr.17: 19313206-19313206 on Build GRCh38
rs374508147 Chr.17: 19313266-19313266 on Build GRCh38
rs761659957 Chr.17: 19313054-19313054 on Build GRCh38
rs1001714698 Chr.17: 19313402-19313402 on Build GRCh38
rs746805290 Chr.17: 19313236-19313236 on Build GRCh38
rs762571448 Chr.17: 19313127-19313128 on Build GRCh38
rs891908276 Chr.17: 19313288-19313288 on Build GRCh38
rs766400271 Chr.17: 19313074-19313074 on Build GRCh38
rs760514767 Chr.17: 19313168-19313168 on Build GRCh38
rs545258288 Chr.17: 19313172-19313172 on Build GRCh38
rs781776788 Chr.17: 19313145-19313145 on Build GRCh38
rs748774660 Chr.17: 19313149-19313149 on Build GRCh38
rs369958146 Chr.17: 19313170-19313170 on Build GRCh38
rs772906576 Chr.17: 19313164-19313164 on Build GRCh38
rs373169143 Chr.17: 19313171-19313171 on Build GRCh38
rs144486716 Chr.17: 19313104-19313104 on Build GRCh38
rs769878520 Chr.17: 19313257-19313257 on Build GRCh38
rs758733429 Chr.17: 19313106-19313106 on Build GRCh38
rs1047449637 Chr.17: 19313241-19313241 on Build GRCh38
rs769594203 Chr.17: 19313330-19313330 on Build GRCh38
rs142878492 Chr.17: 19313366-19313366 on Build GRCh38
rs780997946 Chr.17: 19313308-19313308 on Build GRCh38
rs747509542 Chr.17: 19313130-19313130 on Build GRCh38
rs745755038 Chr.17: 19313324-19313324 on Build GRCh38
rs976355374 Chr.17: 19313210-19313210 on Build GRCh38
rs1024313685 Chr.17: 19313319-19313319 on Build GRCh38
rs754477652 Chr.17: 19313204-19313204 on Build GRCh38
rs867888163 Chr.17: 19313223-19313223 on Build GRCh38
rs943588644 Chr.17: 19313195-19313195 on Build GRCh38
rs772709034 Chr.17: 19313100-19313100 on Build GRCh38
rs971471875 Chr.17: 19313320-19313320 on Build GRCh38
rs777526208 Chr.17: 19313227-19313227 on Build GRCh38
rs200274309 Chr.17: 19313068-19313068 on Build GRCh38
rs764191775 Chr.17: 19313265-19313265 on Build GRCh38
rs770647847 Chr.17: 19313305-19313305 on Build GRCh38
rs760563498 Chr.17: 19313069-19313069 on Build GRCh38
rs752898872 Chr.17: 19313175-19313175 on Build GRCh38
rs780662603 Chr.17: 19313275-19313275 on Build GRCh38
rs979355920 Chr.17: 19313251-19313251 on Build GRCh38
rs763581599 Chr.17: 19313477-19313477 on Build GRCh38
rs768002641 Chr.17: 19313272-19313272 on Build GRCh38
rs759476384 Chr.17: 19313253-19313253 on Build GRCh38
rs367735561 Chr.17: 19313368-19313368 on Build GRCh38
rs977499739 Chr.17: 19313358-19313358 on Build GRCh38
rs777281233 Chr.17: 19313293-19313293 on Build GRCh38
rs770091641 Chr.17: 19313451-19313451 on Build GRCh38
rs778287434 Chr.17: 19313224-19313224 on Build GRCh38
rs976262699 Chr.17: 19313452-19313452 on Build GRCh38
rs906144818 Chr.17: 19313169-19313169 on Build GRCh38
rs757858739 Chr.17: 19313131-19313131 on Build GRCh38
rs770772265 Chr.17: 19313240-19313240 on Build GRCh38
rs115913076 Chr.17: 19313176-19313176 on Build GRCh38
rs765442156 Chr.17: 19313102-19313102 on Build GRCh38
rs764772166 Chr.17: 19313092-19313092 on Build GRCh38
rs753896813 Chr.17: 19313082-19313082 on Build GRCh38
rs55883526 Chr.17: 19313263-19313263 on Build GRCh38
rs371933381 Chr.17: 19313058-19313058 on Build GRCh38
rs146628526 Chr.17: 19313148-19313148 on Build GRCh38
rs370518249 Chr.17: 19313178-19313178 on Build GRCh38
rs925919626 Chr.17: 19313262-19313262 on Build GRCh38
rs372779952 Chr.17: 19313199-19313199 on Build GRCh38
rs778006597 Chr.17: 19313120-19313120 on Build GRCh38
rs375784287 Chr.17: 19313200-19313200 on Build GRCh38
rs192886894 Chr.17: 19313150-19313150 on Build GRCh38
rs887406391 Chr.17: 19313473-19313473 on Build GRCh38
rs188503298 Chr.17: 19313097-19313097 on Build GRCh38
rs951729570 Chr.17: 19313183-19313183 on Build GRCh38
rs1013376786 Chr.17: 19313306-19313306 on Build GRCh38
rs1027308466 Chr.17: 19313182-19313182 on Build GRCh38
rs752119640 Chr.17: 19313280-19313280 on Build GRCh38
rs959466276 Chr.17: 19313286-19313286 on Build GRCh38
rs114905963 Chr.17: 19313225-19313225 on Build GRCh38
rs771830294 Chr.17: 19313153-19313153 on Build GRCh38
rs931921087 Chr.17: 19313229-19313229 on Build GRCh38
rs772888620 Chr.17: 19313291-19313291 on Build GRCh38
rs990803167 Chr.17: 19313380-19313380 on Build GRCh38
rs763141070 Chr.17: 19313264-19313264 on Build GRCh38
rs765315785 Chr.17: 19313173-19313173 on Build GRCh38
rs910806617 Chr.17: 19313396-19313396 on Build GRCh38
rs746275910 Chr.17: 19313375-19313375 on Build GRCh38
rs971332886 Chr.17: 19313060-19313060 on Build GRCh38
rs1014843200 Chr.17: 19313112-19313112 on Build GRCh38
rs757735795 Chr.17: 19313196-19313196 on Build GRCh38
rs757998340 Chr.17: 19313282-19313282 on Build GRCh38
rs527260188 Chr.17: 19313242-19313242 on Build GRCh38
rs746678462 Chr.17: 19313304-19313304 on Build GRCh38
rs374596046 Chr.17: 19313080-19313080 on Build GRCh38
rs376635295 Chr.17: 19313065-19313065 on Build GRCh38

View Full Product Details