Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21190858-21191364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS7B
Entrez Gene ID
25979
Gene Name
dehydrogenase/reductase 7B
Gene Aliases
CGI-93, SDR32C1
UniGene
Hs.386989
Gene Chromosome Location
Chr.17: 21123369-21191972 on Build GRCh38
Transcripts
NM_015510.4 XM_005256595.4 XM_005256596.3 XM_011523786.2 XM_017024425.1 XM_017024426.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4064731 c.955G>T p.E319* Chr.17: 21191130-21191130 on Build GRCh38
1716114 c.898C>G p.L300V Chr.17: 21191073-21191073 on Build GRCh38
559899 c.773-2A>T p.? Chr.17: 21190946-21190946 on Build GRCh38
6407377 c.787A>T p.T263S Chr.17: 21190962-21190962 on Build GRCh38
3889426 c.852G>A p.K284K Chr.17: 21191027-21191027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766730129 Chr.17: 21190946-21190946 on Build GRCh38
rs777672324 Chr.17: 21191161-21191161 on Build GRCh38
rs190206909 Chr.17: 21191148-21191148 on Build GRCh38
rs776747278 Chr.17: 21191172-21191172 on Build GRCh38
rs779841254 Chr.17: 21190989-21190989 on Build GRCh38
rs756749478 Chr.17: 21190973-21190973 on Build GRCh38
rs927602133 Chr.17: 21190992-21190992 on Build GRCh38
rs766556569 Chr.17: 21191321-21191321 on Build GRCh38
rs764435512 Chr.17: 21191080-21191085 on Build GRCh38
rs1040330515 Chr.17: 21191043-21191043 on Build GRCh38
rs750012888 Chr.17: 21191131-21191131 on Build GRCh38
rs762697692 Chr.17: 21190928-21190932 on Build GRCh38
rs765711943 Chr.17: 21190951-21190951 on Build GRCh38
rs1043131745 Chr.17: 21191142-21191142 on Build GRCh38
rs554391712 Chr.17: 21191107-21191107 on Build GRCh38
rs950468338 Chr.17: 21191314-21191314 on Build GRCh38
rs757644233 Chr.17: 21191164-21191169 on Build GRCh38
rs773969632 Chr.17: 21190972-21190973 on Build GRCh38
rs780745796 Chr.17: 21190898-21190898 on Build GRCh38
rs775358808 Chr.17: 21190928-21190928 on Build GRCh38
rs755666136 Chr.17: 21191132-21191132 on Build GRCh38
rs111711075 Chr.17: 21191211-21191211 on Build GRCh38
rs149212384 Chr.17: 21191093-21191093 on Build GRCh38
rs765964693 Chr.17: 21190937-21190937 on Build GRCh38
rs1010312469 Chr.17: 21191326-21191326 on Build GRCh38
rs181113420 Chr.17: 21191208-21191208 on Build GRCh38
rs761064582 Chr.17: 21191028-21191028 on Build GRCh38
rs866825790 Chr.17: 21191150-21191150 on Build GRCh38
rs761842084 Chr.17: 21190937-21190937 on Build GRCh38
rs763214397 Chr.17: 21191201-21191201 on Build GRCh38
rs760735497 Chr.17: 21191076-21191076 on Build GRCh38
rs987203506 Chr.17: 21191320-21191320 on Build GRCh38
rs963517642 Chr.17: 21190883-21190883 on Build GRCh38
rs998829954 Chr.17: 21191215-21191215 on Build GRCh38
rs745477161 Chr.17: 21190903-21190903 on Build GRCh38
rs767597900 Chr.17: 21190941-21190941 on Build GRCh38
rs201363333 Chr.17: 21191112-21191112 on Build GRCh38
rs772060151 Chr.17: 21191158-21191158 on Build GRCh38
rs544201441 Chr.17: 21191176-21191176 on Build GRCh38
rs117992394 Chr.17: 21191290-21191290 on Build GRCh38
rs775021163 Chr.17: 21190975-21190975 on Build GRCh38
rs753215417 Chr.17: 21190972-21190972 on Build GRCh38
rs536195532 Chr.17: 21191059-21191059 on Build GRCh38
rs759857847 Chr.17: 21191102-21191102 on Build GRCh38
rs747137702 Chr.17: 21191166-21191166 on Build GRCh38
rs755354010 Chr.17: 21190950-21190950 on Build GRCh38
rs759754778 Chr.17: 21191174-21191174 on Build GRCh38
rs749090466 Chr.17: 21190931-21190931 on Build GRCh38
rs763459399 Chr.17: 21191113-21191113 on Build GRCh38
rs556531276 Chr.17: 21191025-21191025 on Build GRCh38
rs772573432 Chr.17: 21191247-21191247 on Build GRCh38
rs911539499 Chr.17: 21191018-21191018 on Build GRCh38
rs760825417 Chr.17: 21190945-21190945 on Build GRCh38
rs1003087492 Chr.17: 21191034-21191034 on Build GRCh38
rs754265684 Chr.17: 21190947-21190947 on Build GRCh38
rs774325707 Chr.17: 21190933-21190933 on Build GRCh38
rs921461017 Chr.17: 21191100-21191100 on Build GRCh38
rs748074034 Chr.17: 21191039-21191039 on Build GRCh38
rs780800134 Chr.17: 21190974-21190974 on Build GRCh38
rs372791302 Chr.17: 21191200-21191200 on Build GRCh38
rs998894043 Chr.17: 21191193-21191193 on Build GRCh38
rs370322349 Chr.17: 21191163-21191163 on Build GRCh38
rs200914428 Chr.17: 21191151-21191151 on Build GRCh38
rs773448781 Chr.17: 21190942-21190942 on Build GRCh38
rs12936412 Chr.17: 21191325-21191325 on Build GRCh38
rs1023282689 Chr.17: 21191256-21191256 on Build GRCh38
rs377059764 Chr.17: 21190983-21190983 on Build GRCh38
rs143358693 Chr.17: 21191134-21191134 on Build GRCh38
rs957838699 Chr.17: 21190914-21190914 on Build GRCh38
rs749087723 Chr.17: 21190993-21190993 on Build GRCh38
rs572706396 Chr.17: 21191128-21191128 on Build GRCh38
rs539921759 Chr.17: 21191133-21191133 on Build GRCh38
rs1026148035 Chr.17: 21191291-21191291 on Build GRCh38
rs772529738 Chr.17: 21190918-21190919 on Build GRCh38
rs895794252 Chr.17: 21191233-21191233 on Build GRCh38
rs773206705 Chr.17: 21191056-21191056 on Build GRCh38
rs770964893 Chr.17: 21191170-21191170 on Build GRCh38
rs755897701 Chr.17: 21190985-21190985 on Build GRCh38
rs1033022601 Chr.17: 21191265-21191265 on Build GRCh38
rs374530443 Chr.17: 21190932-21190932 on Build GRCh38
rs115723383 Chr.17: 21191037-21191037 on Build GRCh38
rs533062769 Chr.17: 21191285-21191285 on Build GRCh38
rs754421752 Chr.17: 21191080-21191081 on Build GRCh38
rs773519017 Chr.17: 21191259-21191259 on Build GRCh38
rs112951635 Chr.17: 21190919-21190919 on Build GRCh38
rs973196094 Chr.17: 21190888-21190888 on Build GRCh38
rs750105193 Chr.17: 21190979-21190979 on Build GRCh38

View Full Product Details