Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44798689-44799210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJC1
Entrez Gene ID
10052
Gene Name
gap junction protein gamma 1
Gene Aliases
CX45, GJA7
UniGene
Hs.712052
Gene Chromosome Location
Chr.17: 44794988-44830811 on Build GRCh38
Transcripts
NM_001080383.1 NM_005497.3 XM_005256920.1 XM_005256921.1

SNP Detail

SNP ID Chromosome Location
rs151047627 Chr.17: 44799030-44799030 on Build GRCh38
rs79102764 Chr.17: 44798803-44798803 on Build GRCh38
rs11546869 Chr.17: 44799026-44799026 on Build GRCh38
rs972515543 Chr.17: 44799173-44799173 on Build GRCh38
rs909092748 Chr.17: 44798721-44798721 on Build GRCh38
rs11315 Chr.17: 44798743-44798743 on Build GRCh38
rs948337170 Chr.17: 44798902-44798902 on Build GRCh38
rs1026565151 Chr.17: 44799138-44799138 on Build GRCh38
rs1016230134 Chr.17: 44799084-44799084 on Build GRCh38
rs567411530 Chr.17: 44798970-44798970 on Build GRCh38
rs972352639 Chr.17: 44798816-44798816 on Build GRCh38
rs959936538 Chr.17: 44799143-44799143 on Build GRCh38
rs962074633 Chr.17: 44799171-44799171 on Build GRCh38
rs564074941 Chr.17: 44798833-44798833 on Build GRCh38
rs993079873 Chr.17: 44799178-44799178 on Build GRCh38
rs1007490069 Chr.17: 44799069-44799069 on Build GRCh38
rs11546868 Chr.17: 44799110-44799110 on Build GRCh38
rs753346208 Chr.17: 44798979-44798979 on Build GRCh38
rs1056922885 Chr.17: 44798860-44798860 on Build GRCh38
rs754330318 Chr.17: 44799132-44799132 on Build GRCh38
rs911913853 Chr.17: 44798791-44798791 on Build GRCh38
rs11546866 Chr.17: 44798957-44798957 on Build GRCh38
rs1053020969 Chr.17: 44798992-44798992 on Build GRCh38
rs549305325 Chr.17: 44798877-44798877 on Build GRCh38
rs754464968 Chr.17: 44799014-44799014 on Build GRCh38
rs201061895 Chr.17: 44799159-44799160 on Build GRCh38
rs987412975 Chr.17: 44798790-44798790 on Build GRCh38
rs915556859 Chr.17: 44798890-44798890 on Build GRCh38
rs761240057 Chr.17: 44799057-44799057 on Build GRCh38
rs953490307 Chr.17: 44798782-44798782 on Build GRCh38
rs760742405 Chr.17: 44798950-44798950 on Build GRCh38
rs192652957 Chr.17: 44799050-44799050 on Build GRCh38
rs767486570 Chr.17: 44798771-44798771 on Build GRCh38
rs188130952 Chr.17: 44799017-44799017 on Build GRCh38
rs557976494 Chr.17: 44799045-44799045 on Build GRCh38
rs893747387 Chr.17: 44799092-44799092 on Build GRCh38
rs11546867 Chr.17: 44799096-44799096 on Build GRCh38
rs750592968 Chr.17: 44799003-44799004 on Build GRCh38
rs919123839 Chr.17: 44798820-44798820 on Build GRCh38
rs8082522 Chr.17: 44798972-44798972 on Build GRCh38
rs555863632 Chr.17: 44799016-44799016 on Build GRCh38
rs141220459 Chr.17: 44799124-44799124 on Build GRCh38
rs184499964 Chr.17: 44799117-44799117 on Build GRCh38
rs963250538 Chr.17: 44799105-44799105 on Build GRCh38
rs764882504 Chr.17: 44799065-44799065 on Build GRCh38
rs540087670 Chr.17: 44799052-44799052 on Build GRCh38
rs773267335 Chr.17: 44798855-44798855 on Build GRCh38
rs566672327 Chr.17: 44798764-44798764 on Build GRCh38

View Full Product Details