Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32378125-32378623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSDC1
Entrez Gene ID
55108
Gene Name
BSD domain containing 1
Gene Aliases
-
UniGene
Hs.353454
Gene Chromosome Location
Chr.1: 32364633-32394461 on Build GRCh38
Transcripts
NM_001143888.2 NM_001143889.2 NM_001143890.2 NM_001300958.1 NM_018045.7 NR_125352.1 XM_005270986.4 XM_011541677.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3488175 c.609C>T p.F203F Chr.1: 32378254-32378254 on Build GRCh38
6291966 c.580-7C>T p.? Chr.1: 32378290-32378290 on Build GRCh38
6291967 c.580-7C>T p.? Chr.1: 32378290-32378290 on Build GRCh38
7411910 c.617G>A p.R206Q Chr.1: 32378246-32378246 on Build GRCh38
3488176 c.609C>T p.F203F Chr.1: 32378254-32378254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049043157 Chr.1: 32378426-32378426 on Build GRCh38
rs766366862 Chr.1: 32378228-32378228 on Build GRCh38
rs765632276 Chr.1: 32378181-32378181 on Build GRCh38
rs912707978 Chr.1: 32378382-32378382 on Build GRCh38
rs996890086 Chr.1: 32378448-32378448 on Build GRCh38
rs1023321836 Chr.1: 32378564-32378564 on Build GRCh38
rs527539072 Chr.1: 32378300-32378300 on Build GRCh38
rs373402760 Chr.1: 32378322-32378322 on Build GRCh38
rs777571419 Chr.1: 32378395-32378395 on Build GRCh38
rs377336946 Chr.1: 32378294-32378294 on Build GRCh38
rs1005654836 Chr.1: 32378504-32378504 on Build GRCh38
rs898524380 Chr.1: 32378407-32378410 on Build GRCh38
rs373518583 Chr.1: 32378407-32378407 on Build GRCh38
rs536919746 Chr.1: 32378595-32378595 on Build GRCh38
rs560058496 Chr.1: 32378289-32378289 on Build GRCh38
rs935732795 Chr.1: 32378313-32378313 on Build GRCh38
rs867979679 Chr.1: 32378254-32378254 on Build GRCh38
rs886361711 Chr.1: 32378450-32378450 on Build GRCh38
rs764013404 Chr.1: 32378292-32378293 on Build GRCh38
rs776317589 Chr.1: 32378451-32378451 on Build GRCh38
rs757406630 Chr.1: 32378313-32378314 on Build GRCh38
rs760803134 Chr.1: 32378287-32378287 on Build GRCh38
rs749183364 Chr.1: 32378260-32378260 on Build GRCh38
rs550040427 Chr.1: 32378463-32378463 on Build GRCh38
rs754661592 Chr.1: 32378223-32378223 on Build GRCh38
rs754014924 Chr.1: 32378296-32378297 on Build GRCh38
rs765592300 Chr.1: 32378293-32378293 on Build GRCh38
rs992779914 Chr.1: 32378196-32378196 on Build GRCh38
rs180836589 Chr.1: 32378478-32378478 on Build GRCh38
rs539284480 Chr.1: 32378529-32378529 on Build GRCh38
rs548966731 Chr.1: 32378364-32378364 on Build GRCh38
rs760817406 Chr.1: 32378164-32378164 on Build GRCh38
rs935564596 Chr.1: 32378361-32378361 on Build GRCh38
rs1039799690 Chr.1: 32378401-32378401 on Build GRCh38
rs750992561 Chr.1: 32378182-32378182 on Build GRCh38
rs960539015 Chr.1: 32378533-32378533 on Build GRCh38
rs373057622 Chr.1: 32378233-32378233 on Build GRCh38
rs150963407 Chr.1: 32378227-32378227 on Build GRCh38
rs767010333 Chr.1: 32378333-32378333 on Build GRCh38
rs979728441 Chr.1: 32378237-32378237 on Build GRCh38
rs946372123 Chr.1: 32378314-32378314 on Build GRCh38
rs777450364 Chr.1: 32378247-32378247 on Build GRCh38
rs577793168 Chr.1: 32378184-32378184 on Build GRCh38
rs751121688 Chr.1: 32378220-32378220 on Build GRCh38
rs769503914 Chr.1: 32378315-32378315 on Build GRCh38
rs144659572 Chr.1: 32378280-32378280 on Build GRCh38
rs942775380 Chr.1: 32378400-32378400 on Build GRCh38
rs770858246 Chr.1: 32378267-32378267 on Build GRCh38
rs762062939 Chr.1: 32378291-32378291 on Build GRCh38
rs569264845 Chr.1: 32378312-32378312 on Build GRCh38
rs148506012 Chr.1: 32378535-32378535 on Build GRCh38
rs779673645 Chr.1: 32378185-32378185 on Build GRCh38
rs866514828 Chr.1: 32378271-32378271 on Build GRCh38
rs374773298 Chr.1: 32378177-32378177 on Build GRCh38
rs531846140 Chr.1: 32378421-32378421 on Build GRCh38
rs762396588 Chr.1: 32378373-32378373 on Build GRCh38
rs557271564 Chr.1: 32378584-32378584 on Build GRCh38
rs768863124 Chr.1: 32378288-32378288 on Build GRCh38
rs770363127 Chr.1: 32378470-32378470 on Build GRCh38
rs757708121 Chr.1: 32378179-32378179 on Build GRCh38

View Full Product Details