Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60159042-60159584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA4
Entrez Gene ID
762
Gene Name
carbonic anhydrase 4
Gene Aliases
CAIV, Car4, RP17
UniGene
Hs.89485
Gene Chromosome Location
Chr.17: 60149941-60178118 on Build GRCh38
Transcripts
XM_005257639.2 XM_011525183.2 XM_017025012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7097489 c.786G>A p.Q262Q Chr.17: 60159271-60159271 on Build GRCh38
5565315 c.812G>T p.R271M Chr.17: 60159297-60159297 on Build GRCh38
6081917 c.828_829GG>TT p.G277W Chr.17: 60159313-60159314 on Build GRCh38
6990012 c.846A>T p.I282I Chr.17: 60159331-60159331 on Build GRCh38
6871198 c.836G>T p.R279L Chr.17: 60159321-60159321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750641080 Chr.17: 60159320-60159320 on Build GRCh38
rs755477580 Chr.17: 60159322-60159322 on Build GRCh38
rs545060770 Chr.17: 60159419-60159419 on Build GRCh38
rs549334435 Chr.17: 60159225-60159225 on Build GRCh38
rs550394772 Chr.17: 60159168-60159168 on Build GRCh38
rs777166953 Chr.17: 60159273-60159273 on Build GRCh38
rs142401267 Chr.17: 60159072-60159072 on Build GRCh38
rs372070576 Chr.17: 60159355-60159355 on Build GRCh38
rs571708928 Chr.17: 60159118-60159118 on Build GRCh38
rs377145857 Chr.17: 60159324-60159324 on Build GRCh38
rs766938149 Chr.17: 60159295-60159295 on Build GRCh38
rs983535079 Chr.17: 60159495-60159495 on Build GRCh38
rs778523375 Chr.17: 60159340-60159340 on Build GRCh38
rs1021190457 Chr.17: 60159137-60159137 on Build GRCh38
rs566759180 Chr.17: 60159198-60159198 on Build GRCh38
rs373145386 Chr.17: 60159421-60159421 on Build GRCh38
rs962477037 Chr.17: 60159134-60159134 on Build GRCh38
rs962446098 Chr.17: 60159346-60159346 on Build GRCh38
rs187221698 Chr.17: 60159285-60159285 on Build GRCh38
rs755070090 Chr.17: 60159397-60159397 on Build GRCh38
rs1014109391 Chr.17: 60159302-60159302 on Build GRCh38
rs775069368 Chr.17: 60159256-60159257 on Build GRCh38
rs759093480 Chr.17: 60159216-60159216 on Build GRCh38
rs745999221 Chr.17: 60159255-60159255 on Build GRCh38
rs138395053 Chr.17: 60159289-60159289 on Build GRCh38
rs775003473 Chr.17: 60159298-60159298 on Build GRCh38
rs376414377 Chr.17: 60159407-60159407 on Build GRCh38
rs977820796 Chr.17: 60159466-60159466 on Build GRCh38
rs973565442 Chr.17: 60159162-60159162 on Build GRCh38
rs150432787 Chr.17: 60159246-60159246 on Build GRCh38
rs372423531 Chr.17: 60159502-60159502 on Build GRCh38
rs772911800 Chr.17: 60159464-60159464 on Build GRCh38
rs372344598 Chr.17: 60159321-60159321 on Build GRCh38
rs1032238872 Chr.17: 60159175-60159175 on Build GRCh38
rs757026218 Chr.17: 60159310-60159310 on Build GRCh38
rs767546771 Chr.17: 60159221-60159221 on Build GRCh38
rs764055659 Chr.17: 60159299-60159299 on Build GRCh38
rs879449973 Chr.17: 60159100-60159100 on Build GRCh38
rs780678842 Chr.17: 60159260-60159260 on Build GRCh38
rs748580097 Chr.17: 60159325-60159325 on Build GRCh38
rs922136078 Chr.17: 60159547-60159547 on Build GRCh38
rs758397326 Chr.17: 60159259-60159259 on Build GRCh38
rs1003897421 Chr.17: 60159088-60159088 on Build GRCh38
rs569363525 Chr.17: 60159233-60159233 on Build GRCh38
rs753677966 Chr.17: 60159301-60159301 on Build GRCh38
rs973506549 Chr.17: 60159545-60159545 on Build GRCh38
rs766089463 Chr.17: 60159204-60159204 on Build GRCh38
rs763106204 Chr.17: 60159379-60159379 on Build GRCh38
rs146141867 Chr.17: 60159354-60159354 on Build GRCh38
rs963099156 Chr.17: 60159530-60159530 on Build GRCh38
rs1030289775 Chr.17: 60159075-60159075 on Build GRCh38
rs748492425 Chr.17: 60159275-60159275 on Build GRCh38
rs779300156 Chr.17: 60159123-60159123 on Build GRCh38
rs529983471 Chr.17: 60159193-60159193 on Build GRCh38
rs768095636 Chr.17: 60159458-60159458 on Build GRCh38
rs146362767 Chr.17: 60159202-60159202 on Build GRCh38
rs779739325 Chr.17: 60159427-60159427 on Build GRCh38
rs771703329 Chr.17: 60159345-60159345 on Build GRCh38
rs1000661685 Chr.17: 60159409-60159409 on Build GRCh38
rs747477911 Chr.17: 60159266-60159266 on Build GRCh38
rs970447678 Chr.17: 60159490-60159490 on Build GRCh38
rs564813136 Chr.17: 60159436-60159436 on Build GRCh38
rs765023035 Chr.17: 60159372-60159372 on Build GRCh38
rs765504256 Chr.17: 60159312-60159312 on Build GRCh38
rs567212140 Chr.17: 60159174-60159174 on Build GRCh38
rs760198248 Chr.17: 60159258-60159259 on Build GRCh38
rs761755192 Chr.17: 60159442-60159442 on Build GRCh38
rs768153190 Chr.17: 60159440-60159440 on Build GRCh38
rs745887096 Chr.17: 60159145-60159145 on Build GRCh38
rs558803561 Chr.17: 60159226-60159226 on Build GRCh38
rs200329982 Chr.17: 60159337-60159337 on Build GRCh38
rs142961963 Chr.17: 60159292-60159292 on Build GRCh38
rs201580764 Chr.17: 60159408-60159408 on Build GRCh38
rs2229179 Chr.17: 60159441-60159441 on Build GRCh38
rs761665677 Chr.17: 60159369-60159369 on Build GRCh38
rs769149664 Chr.17: 60159269-60159269 on Build GRCh38
rs926018782 Chr.17: 60159291-60159291 on Build GRCh38
rs768900579 Chr.17: 60159201-60159201 on Build GRCh38
rs769821291 Chr.17: 60159338-60159338 on Build GRCh38
rs546924276 Chr.17: 60159195-60159195 on Build GRCh38
rs367980627 Chr.17: 60159352-60159352 on Build GRCh38
rs751638856 Chr.17: 60159396-60159396 on Build GRCh38
rs774043643 Chr.17: 60159205-60159205 on Build GRCh38
rs1803230 Chr.17: 60159435-60159435 on Build GRCh38
rs766591923 Chr.17: 60159388-60159388 on Build GRCh38
rs936009055 Chr.17: 60159538-60159538 on Build GRCh38
rs565056769 Chr.17: 60159112-60159112 on Build GRCh38
rs140515828 Chr.17: 60159238-60159238 on Build GRCh38
rs757505800 Chr.17: 60159243-60159243 on Build GRCh38
rs745518288 Chr.17: 60159342-60159342 on Build GRCh38
rs745912445 Chr.17: 60159184-60159184 on Build GRCh38
rs771400562 Chr.17: 60159315-60159315 on Build GRCh38
rs751214164 Chr.17: 60159489-60159489 on Build GRCh38
rs530720914 Chr.17: 60159483-60159483 on Build GRCh38
rs774889872 Chr.17: 60159350-60159350 on Build GRCh38
rs907592817 Chr.17: 60159519-60159519 on Build GRCh38
rs966599918 Chr.17: 60159167-60159167 on Build GRCh38
rs750163649 Chr.17: 60159378-60159378 on Build GRCh38
rs770755416 Chr.17: 60159278-60159278 on Build GRCh38

View Full Product Details