Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60807983-60808516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAS3
Entrez Gene ID
54828
Gene Name
BCAS3, microtubule associated cell migration factor
Gene Aliases
GAOB1, MAAB
UniGene
Hs.655028
Gene Chromosome Location
Chr.17: 60677811-61392838 on Build GRCh38
Transcripts
NM_001099432.1 NM_001320470.1 NM_017679.3 XM_005257472.1 XM_011524939.1 XM_011524940.1 XM_011524941.1 XM_011524942.2 XM_011524943.2 XM_011524944.1 XM_011524945.2 XM_011524946.2 XM_011524947.1 XM_011524948.2 XM_011524949.2 XM_011524950.1 XM_011524951.2 XM_017024783.1 XM_017024784.1 XM_017024785.1 XM_017024786.1 XM_017024787.1 XM_017024788.1 XM_017024789.1 XM_017024790.1 XM_017024791.1 XM_017024792.1 XM_017024793.1 XM_017024794.1 XM_017024795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6435099 c.458A>G p.K153R Chr.17: 60808058-60808058 on Build GRCh38
5464129 c.476+8A>C p.? Chr.17: 60808084-60808084 on Build GRCh38
5784992 c.437G>C p.R146T Chr.17: 60808037-60808037 on Build GRCh38
5784993 c.437G>C p.R146T Chr.17: 60808037-60808037 on Build GRCh38
3421747 c.437G>T p.R146I Chr.17: 60808037-60808037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746138997 Chr.17: 60808067-60808067 on Build GRCh38
rs572193796 Chr.17: 60808142-60808142 on Build GRCh38
rs757347131 Chr.17: 60808045-60808045 on Build GRCh38
rs980110806 Chr.17: 60808242-60808242 on Build GRCh38
rs929486321 Chr.17: 60808123-60808123 on Build GRCh38
rs561792991 Chr.17: 60808459-60808459 on Build GRCh38
rs756650173 Chr.17: 60808015-60808015 on Build GRCh38
rs1056414149 Chr.17: 60808366-60808366 on Build GRCh38
rs777654873 Chr.17: 60808019-60808019 on Build GRCh38
rs1035743198 Chr.17: 60808047-60808047 on Build GRCh38
rs201042830 Chr.17: 60808052-60808052 on Build GRCh38
rs773093242 Chr.17: 60808110-60808110 on Build GRCh38
rs36036625 Chr.17: 60808385-60808386 on Build GRCh38
rs901833375 Chr.17: 60808210-60808210 on Build GRCh38
rs747966407 Chr.17: 60808103-60808103 on Build GRCh38
rs891681894 Chr.17: 60808304-60808304 on Build GRCh38
rs768518772 Chr.17: 60808089-60808089 on Build GRCh38
rs776596789 Chr.17: 60808102-60808102 on Build GRCh38
rs762497525 Chr.17: 60808113-60808113 on Build GRCh38
rs868267948 Chr.17: 60808042-60808042 on Build GRCh38
rs758788416 Chr.17: 60808414-60808414 on Build GRCh38
rs914660656 Chr.17: 60808220-60808220 on Build GRCh38
rs541641894 Chr.17: 60808146-60808146 on Build GRCh38
rs959970170 Chr.17: 60808399-60808399 on Build GRCh38
rs909321622 Chr.17: 60808145-60808145 on Build GRCh38
rs749373140 Chr.17: 60808028-60808028 on Build GRCh38
rs943430466 Chr.17: 60808162-60808162 on Build GRCh38
rs534388325 Chr.17: 60808284-60808284 on Build GRCh38
rs779366709 Chr.17: 60808209-60808209 on Build GRCh38
rs147364009 Chr.17: 60808229-60808229 on Build GRCh38
rs950947408 Chr.17: 60808486-60808486 on Build GRCh38
rs756778697 Chr.17: 60808060-60808060 on Build GRCh38
rs929182088 Chr.17: 60808404-60808404 on Build GRCh38
rs1035393829 Chr.17: 60808371-60808371 on Build GRCh38
rs1011428256 Chr.17: 60808317-60808317 on Build GRCh38
rs1021444148 Chr.17: 60808332-60808332 on Build GRCh38
rs543759770 Chr.17: 60808260-60808260 on Build GRCh38
rs186171820 Chr.17: 60808198-60808198 on Build GRCh38
rs761199701 Chr.17: 60808082-60808082 on Build GRCh38
rs1032324424 Chr.17: 60808134-60808134 on Build GRCh38
rs563941625 Chr.17: 60808283-60808283 on Build GRCh38
rs966711516 Chr.17: 60808064-60808064 on Build GRCh38
rs750930580 Chr.17: 60808403-60808403 on Build GRCh38
rs139042614 Chr.17: 60808410-60808410 on Build GRCh38
rs531347801 Chr.17: 60808361-60808361 on Build GRCh38
rs769759860 Chr.17: 60808107-60808107 on Build GRCh38
rs1039063783 Chr.17: 60808191-60808193 on Build GRCh38

View Full Product Details