Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49158808-49159312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALNT2
Entrez Gene ID
124872
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 2
Gene Aliases
B4GALT, GALGT2
UniGene
Hs.374679
Gene Chromosome Location
Chr.17: 49120347-49170722 on Build GRCh38
Transcripts
NM_001159387.1 NM_001159388.1 NM_153446.2 XM_011524314.2 XM_017024173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3518881 c.721G>A p.D241N Chr.17: 49159079-49159079 on Build GRCh38
4819675 c.786G>C p.R262R Chr.17: 49159144-49159144 on Build GRCh38
1630249 c.706C>T p.L236L Chr.17: 49159064-49159064 on Build GRCh38
4791656 c.726C>T p.V242V Chr.17: 49159084-49159084 on Build GRCh38
5812934 c.743A>T p.Q248L Chr.17: 49159101-49159101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775950015 Chr.17: 49159191-49159191 on Build GRCh38
rs973604855 Chr.17: 49158984-49158984 on Build GRCh38
rs750940885 Chr.17: 49159110-49159110 on Build GRCh38
rs201083609 Chr.17: 49159016-49159016 on Build GRCh38
rs72835413 Chr.17: 49159289-49159289 on Build GRCh38
rs754898668 Chr.17: 49159163-49159163 on Build GRCh38
rs761295584 Chr.17: 49159133-49159133 on Build GRCh38
rs530802842 Chr.17: 49158943-49158943 on Build GRCh38
rs781272860 Chr.17: 49159166-49159166 on Build GRCh38
rs1056256291 Chr.17: 49158863-49158863 on Build GRCh38
rs750016761 Chr.17: 49159142-49159142 on Build GRCh38
rs758102207 Chr.17: 49158988-49158988 on Build GRCh38
rs755017029 Chr.17: 49158987-49158987 on Build GRCh38
rs373738918 Chr.17: 49159260-49159260 on Build GRCh38
rs769982513 Chr.17: 49159059-49159059 on Build GRCh38
rs749420906 Chr.17: 49159176-49159176 on Build GRCh38
rs191262044 Chr.17: 49159036-49159036 on Build GRCh38
rs1009055200 Chr.17: 49158967-49158967 on Build GRCh38
rs746900137 Chr.17: 49159007-49159007 on Build GRCh38
rs771196540 Chr.17: 49159188-49159188 on Build GRCh38
rs971495392 Chr.17: 49159173-49159173 on Build GRCh38
rs773292459 Chr.17: 49159065-49159065 on Build GRCh38
rs200502885 Chr.17: 49159171-49159171 on Build GRCh38
rs762809201 Chr.17: 49159271-49159271 on Build GRCh38
rs909819099 Chr.17: 49158844-49158844 on Build GRCh38
rs188023919 Chr.17: 49158949-49158949 on Build GRCh38
rs781144105 Chr.17: 49159029-49159038 on Build GRCh38
rs538069169 Chr.17: 49158831-49158831 on Build GRCh38
rs533914071 Chr.17: 49158924-49158924 on Build GRCh38
rs764847797 Chr.17: 49159135-49159135 on Build GRCh38
rs145087753 Chr.17: 49159182-49159182 on Build GRCh38
rs995951965 Chr.17: 49158914-49158915 on Build GRCh38
rs574326799 Chr.17: 49158830-49158830 on Build GRCh38
rs751415856 Chr.17: 49159160-49159160 on Build GRCh38
rs1043373393 Chr.17: 49158915-49158915 on Build GRCh38
rs769649482 Chr.17: 49159147-49159148 on Build GRCh38
rs900020561 Chr.17: 49158965-49158965 on Build GRCh38
rs748069796 Chr.17: 49159038-49159039 on Build GRCh38
rs781360639 Chr.17: 49159023-49159023 on Build GRCh38
rs532625676 Chr.17: 49158848-49158848 on Build GRCh38
rs902463471 Chr.17: 49158857-49158857 on Build GRCh38
rs1029713257 Chr.17: 49158976-49158976 on Build GRCh38
rs548372174 Chr.17: 49158927-49158927 on Build GRCh38
rs768035640 Chr.17: 49158856-49158856 on Build GRCh38
rs568586812 Chr.17: 49158937-49158937 on Build GRCh38
rs779668398 Chr.17: 49159143-49159143 on Build GRCh38
rs140662851 Chr.17: 49159172-49159172 on Build GRCh38
rs773251393 Chr.17: 49159151-49159161 on Build GRCh38
rs764616906 Chr.17: 49159231-49159231 on Build GRCh38
rs748155124 Chr.17: 49159170-49159170 on Build GRCh38
rs114986987 Chr.17: 49159189-49159189 on Build GRCh38
rs951441256 Chr.17: 49159032-49159032 on Build GRCh38
rs753267089 Chr.17: 49159123-49159123 on Build GRCh38
rs945043134 Chr.17: 49158901-49158901 on Build GRCh38
rs963033214 Chr.17: 49159080-49159080 on Build GRCh38
rs926518245 Chr.17: 49158860-49158860 on Build GRCh38
rs768733765 Chr.17: 49159012-49159012 on Build GRCh38
rs767062318 Chr.17: 49158972-49158972 on Build GRCh38
rs926522274 Chr.17: 49159153-49159153 on Build GRCh38
rs377719029 Chr.17: 49159006-49159006 on Build GRCh38
rs563818463 Chr.17: 49158833-49158833 on Build GRCh38
rs746224531 Chr.17: 49159190-49159190 on Build GRCh38
rs546090350 Chr.17: 49158861-49158861 on Build GRCh38
rs768014101 Chr.17: 49159092-49159092 on Build GRCh38
rs1040356577 Chr.17: 49158964-49158964 on Build GRCh38
rs890210892 Chr.17: 49158958-49158958 on Build GRCh38
rs913626370 Chr.17: 49158888-49158888 on Build GRCh38
rs915148877 Chr.17: 49159272-49159272 on Build GRCh38
rs775552144 Chr.17: 49158862-49158862 on Build GRCh38
rs373524103 Chr.17: 49158975-49158975 on Build GRCh38
rs761289698 Chr.17: 49159201-49159201 on Build GRCh38
rs559492185 Chr.17: 49158909-49158909 on Build GRCh38
rs774799306 Chr.17: 49159072-49159072 on Build GRCh38
rs762457674 Chr.17: 49159251-49159251 on Build GRCh38
rs771159365 Chr.17: 49159070-49159070 on Build GRCh38
rs201616383 Chr.17: 49159246-49159246 on Build GRCh38
rs935254503 Chr.17: 49159255-49159255 on Build GRCh38

View Full Product Details