Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58089448-58089956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNLL2
Entrez Gene ID
140735
Gene Name
dynein light chain LC8-type 2
Gene Aliases
DNCL1B, Dlc2, RSPH22
UniGene
Hs.720595
Gene Chromosome Location
Chr.17: 58083419-58090257 on Build GRCh38
Transcripts
NM_080677.2

SNP Detail

SNP ID Chromosome Location
rs1036440148 Chr.17: 58089863-58089863 on Build GRCh38
rs887876702 Chr.17: 58089655-58089655 on Build GRCh38
rs985469725 Chr.17: 58089599-58089599 on Build GRCh38
rs74470907 Chr.17: 58089690-58089690 on Build GRCh38
rs781525575 Chr.17: 58089802-58089802 on Build GRCh38
rs980747351 Chr.17: 58089838-58089838 on Build GRCh38
rs991313983 Chr.17: 58089843-58089843 on Build GRCh38
rs997435911 Chr.17: 58089713-58089713 on Build GRCh38
rs761198615 Chr.17: 58089562-58089562 on Build GRCh38
rs923478106 Chr.17: 58089805-58089805 on Build GRCh38
rs992858856 Chr.17: 58089597-58089597 on Build GRCh38
rs1043358397 Chr.17: 58089858-58089858 on Build GRCh38
rs1013413158 Chr.17: 58089909-58089909 on Build GRCh38
rs543169163 Chr.17: 58089818-58089818 on Build GRCh38
rs138574553 Chr.17: 58089648-58089648 on Build GRCh38
rs925198950 Chr.17: 58089859-58089859 on Build GRCh38
rs1030691422 Chr.17: 58089717-58089717 on Build GRCh38
rs562221344 Chr.17: 58089739-58089739 on Build GRCh38
rs190895072 Chr.17: 58089489-58089489 on Build GRCh38
rs909264149 Chr.17: 58089847-58089847 on Build GRCh38
rs573061023 Chr.17: 58089614-58089614 on Build GRCh38
rs948051742 Chr.17: 58089837-58089837 on Build GRCh38
rs968463743 Chr.17: 58089765-58089765 on Build GRCh38
rs933171326 Chr.17: 58089929-58089929 on Build GRCh38
rs755391385 Chr.17: 58089781-58089781 on Build GRCh38
rs941836303 Chr.17: 58089677-58089677 on Build GRCh38
rs1021406474 Chr.17: 58089471-58089471 on Build GRCh38
rs937975796 Chr.17: 58089862-58089862 on Build GRCh38
rs1013599694 Chr.17: 58089558-58089558 on Build GRCh38
rs1002012086 Chr.17: 58089494-58089494 on Build GRCh38
rs916581746 Chr.17: 58089848-58089848 on Build GRCh38
rs1034197980 Chr.17: 58089506-58089506 on Build GRCh38
rs950695041 Chr.17: 58089591-58089591 on Build GRCh38
rs561343921 Chr.17: 58089918-58089918 on Build GRCh38
rs921016652 Chr.17: 58089637-58089637 on Build GRCh38
rs935599133 Chr.17: 58089584-58089584 on Build GRCh38
rs536425372 Chr.17: 58089557-58089557 on Build GRCh38
rs1044045879 Chr.17: 58089691-58089691 on Build GRCh38
rs913623367 Chr.17: 58089844-58089844 on Build GRCh38
rs918183088 Chr.17: 58089825-58089825 on Build GRCh38
rs979607644 Chr.17: 58089555-58089555 on Build GRCh38
rs1026351834 Chr.17: 58089573-58089573 on Build GRCh38
rs756792245 Chr.17: 58089678-58089678 on Build GRCh38
rs903742889 Chr.17: 58089696-58089696 on Build GRCh38
rs1007248721 Chr.17: 58089750-58089750 on Build GRCh38
rs896344522 Chr.17: 58089890-58089890 on Build GRCh38
rs919085973 Chr.17: 58089910-58089910 on Build GRCh38
rs974918030 Chr.17: 58089635-58089635 on Build GRCh38
rs554502436 Chr.17: 58089609-58089609 on Build GRCh38
rs992406044 Chr.17: 58089822-58089822 on Build GRCh38
rs150717233 Chr.17: 58089772-58089772 on Build GRCh38
rs954874077 Chr.17: 58089810-58089810 on Build GRCh38
rs376105178 Chr.17: 58089643-58089643 on Build GRCh38
rs576441333 Chr.17: 58089698-58089698 on Build GRCh38
rs543849978 Chr.17: 58089730-58089730 on Build GRCh38
rs1021353970 Chr.17: 58089764-58089764 on Build GRCh38
rs1013715851 Chr.17: 58089733-58089733 on Build GRCh38
rs960741233 Chr.17: 58089551-58089552 on Build GRCh38
rs950724305 Chr.17: 58089753-58089753 on Build GRCh38
rs749994305 Chr.17: 58089613-58089613 on Build GRCh38
rs948074951 Chr.17: 58089645-58089645 on Build GRCh38

View Full Product Details