Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58194882-58195385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPX
Entrez Gene ID
8288
Gene Name
eosinophil peroxidase
Gene Aliases
EPO, EPP, EPX-PEN, EPXD
UniGene
Hs.279259
Gene Chromosome Location
Chr.17: 58192724-58205174 on Build GRCh38
Transcripts
NM_000502.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6355120 c.661G>A p.A221T Chr.17: 58195030-58195030 on Build GRCh38
6350790 c.660A>G p.R220R Chr.17: 58195029-58195029 on Build GRCh38
5636404 c.709T>G p.F237V Chr.17: 58195078-58195078 on Build GRCh38
3692003 c.717G>A p.P239P Chr.17: 58195086-58195086 on Build GRCh38
6303633 c.683G>T p.G228V Chr.17: 58195052-58195052 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775657051 Chr.17: 58195111-58195111 on Build GRCh38
rs765474483 Chr.17: 58195129-58195129 on Build GRCh38
rs747916329 Chr.17: 58194967-58194967 on Build GRCh38
rs1026877493 Chr.17: 58195338-58195338 on Build GRCh38
rs772449221 Chr.17: 58195107-58195107 on Build GRCh38
rs201161563 Chr.17: 58195163-58195163 on Build GRCh38
rs35617692 Chr.17: 58195140-58195140 on Build GRCh38
rs35620058 Chr.17: 58195204-58195204 on Build GRCh38
rs759130733 Chr.17: 58195138-58195138 on Build GRCh38
rs746503272 Chr.17: 58194949-58194949 on Build GRCh38
rs779165615 Chr.17: 58195097-58195097 on Build GRCh38
rs530146339 Chr.17: 58195022-58195022 on Build GRCh38
rs756633571 Chr.17: 58195218-58195219 on Build GRCh38
rs758012923 Chr.17: 58195011-58195011 on Build GRCh38
rs777640435 Chr.17: 58195092-58195092 on Build GRCh38
rs753110345 Chr.17: 58194933-58194933 on Build GRCh38
rs200004363 Chr.17: 58195027-58195027 on Build GRCh38
rs943260454 Chr.17: 58195136-58195136 on Build GRCh38
rs967878818 Chr.17: 58195073-58195073 on Build GRCh38
rs140411083 Chr.17: 58195078-58195078 on Build GRCh38
rs967259851 Chr.17: 58194902-58194902 on Build GRCh38
rs960199599 Chr.17: 58195252-58195252 on Build GRCh38
rs750130352 Chr.17: 58194939-58194939 on Build GRCh38
rs775174536 Chr.17: 58195217-58195217 on Build GRCh38
rs540908451 Chr.17: 58194936-58194936 on Build GRCh38
rs984495725 Chr.17: 58195333-58195333 on Build GRCh38
rs2240815 Chr.17: 58195029-58195029 on Build GRCh38
rs751065435 Chr.17: 58195014-58195014 on Build GRCh38
rs527517013 Chr.17: 58195065-58195065 on Build GRCh38
rs766438754 Chr.17: 58194922-58194922 on Build GRCh38
rs748555057 Chr.17: 58195198-58195198 on Build GRCh38
rs559293837 Chr.17: 58195288-58195288 on Build GRCh38
rs148671148 Chr.17: 58195001-58195001 on Build GRCh38
rs755681100 Chr.17: 58195141-58195141 on Build GRCh38
rs757249995 Chr.17: 58195154-58195154 on Build GRCh38
rs34599426 Chr.17: 58195171-58195171 on Build GRCh38
rs768655305 Chr.17: 58195179-58195179 on Build GRCh38
rs763183947 Chr.17: 58194991-58194991 on Build GRCh38
rs903265253 Chr.17: 58195303-58195303 on Build GRCh38
rs760148616 Chr.17: 58195218-58195218 on Build GRCh38
rs762907071 Chr.17: 58194917-58194917 on Build GRCh38
rs747092340 Chr.17: 58195112-58195112 on Build GRCh38
rs980754446 Chr.17: 58195242-58195242 on Build GRCh38
rs200969873 Chr.17: 58195054-58195054 on Build GRCh38
rs774499008 Chr.17: 58195028-58195028 on Build GRCh38
rs762138875 Chr.17: 58195068-58195068 on Build GRCh38
rs376355129 Chr.17: 58195048-58195048 on Build GRCh38
rs779637519 Chr.17: 58195013-58195013 on Build GRCh38
rs371439084 Chr.17: 58195005-58195005 on Build GRCh38
rs143977496 Chr.17: 58195021-58195021 on Build GRCh38
rs781359068 Chr.17: 58195185-58195185 on Build GRCh38
rs374074563 Chr.17: 58194950-58194950 on Build GRCh38
rs759604568 Chr.17: 58194998-58194998 on Build GRCh38
rs1002948590 Chr.17: 58195306-58195306 on Build GRCh38
rs982431499 Chr.17: 58195354-58195354 on Build GRCh38
rs758630472 Chr.17: 58195168-58195168 on Build GRCh38
rs781164005 Chr.17: 58194957-58194957 on Build GRCh38
rs202146551 Chr.17: 58195150-58195150 on Build GRCh38
rs745353396 Chr.17: 58194968-58194968 on Build GRCh38
rs755977833 Chr.17: 58195085-58195085 on Build GRCh38
rs894336766 Chr.17: 58195332-58195332 on Build GRCh38
rs144947854 Chr.17: 58195082-58195082 on Build GRCh38
rs188328305 Chr.17: 58194937-58194937 on Build GRCh38
rs779569442 Chr.17: 58194944-58194944 on Build GRCh38
rs181842900 Chr.17: 58195335-58195335 on Build GRCh38
rs1035474783 Chr.17: 58195329-58195329 on Build GRCh38
rs760028611 Chr.17: 58194929-58194929 on Build GRCh38
rs373476557 Chr.17: 58195174-58195174 on Build GRCh38
rs765494671 Chr.17: 58194903-58194903 on Build GRCh38
rs746052693 Chr.17: 58195106-58195106 on Build GRCh38
rs767429068 Chr.17: 58195315-58195315 on Build GRCh38
rs62636621 Chr.17: 58195094-58195094 on Build GRCh38
rs770346476 Chr.17: 58195201-58195201 on Build GRCh38
rs763472367 Chr.17: 58195220-58195220 on Build GRCh38
rs201189852 Chr.17: 58194986-58194986 on Build GRCh38
rs777050228 Chr.17: 58195118-58195118 on Build GRCh38
rs754178675 Chr.17: 58195009-58195009 on Build GRCh38
rs371395875 Chr.17: 58195203-58195203 on Build GRCh38
rs373468375 Chr.17: 58194932-58194932 on Build GRCh38
rs577615530 Chr.17: 58195334-58195334 on Build GRCh38
rs763478702 Chr.17: 58195247-58195247 on Build GRCh38
rs367654802 Chr.17: 58195002-58195002 on Build GRCh38
rs758018685 Chr.17: 58194940-58194940 on Build GRCh38
rs773648393 Chr.17: 58195134-58195134 on Build GRCh38
rs766964937 Chr.17: 58195079-58195079 on Build GRCh38
rs762203083 Chr.17: 58195120-58195120 on Build GRCh38
rs760730928 Chr.17: 58195245-58195245 on Build GRCh38
rs764714152 Chr.17: 58195055-58195055 on Build GRCh38
rs769612241 Chr.17: 58194973-58194973 on Build GRCh38
rs867500484 Chr.17: 58194904-58194904 on Build GRCh38
rs775743235 Chr.17: 58195051-58195051 on Build GRCh38
rs774425519 Chr.17: 58194996-58194996 on Build GRCh38
rs766029400 Chr.17: 58195074-58195074 on Build GRCh38
rs943769778 Chr.17: 58195119-58195119 on Build GRCh38
rs200843597 Chr.17: 58194992-58194992 on Build GRCh38
rs919019603 Chr.17: 58195290-58195290 on Build GRCh38
rs201328305 Chr.17: 58195018-58195018 on Build GRCh38
rs751610120 Chr.17: 58194927-58194927 on Build GRCh38
rs149721417 Chr.17: 58195149-58195149 on Build GRCh38
rs35896669 Chr.17: 58195115-58195115 on Build GRCh38
rs981258813 Chr.17: 58194918-58194918 on Build GRCh38
rs972851361 Chr.17: 58195263-58195263 on Build GRCh38
rs574158253 Chr.17: 58195350-58195350 on Build GRCh38
rs771702321 Chr.17: 58195215-58195215 on Build GRCh38
rs748033685 Chr.17: 58195020-58195020 on Build GRCh38
rs368783186 Chr.17: 58195095-58195095 on Build GRCh38
rs34444359 Chr.17: 58195017-58195017 on Build GRCh38
rs770078560 Chr.17: 58195086-58195086 on Build GRCh38
rs768128933 Chr.17: 58195000-58195000 on Build GRCh38

View Full Product Details