Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76008252-76008750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVPL
Entrez Gene ID
2125
Gene Name
envoplakin
Gene Aliases
EVPK
UniGene
Hs.500635
Gene Chromosome Location
Chr.17: 76006845-76027452 on Build GRCh38
Transcripts
NM_001320747.1 NM_001988.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
707171 c.4631A>G p.N1544S Chr.17: 76008574-76008574 on Build GRCh38
5530656 c.4534G>A p.A1512T Chr.17: 76008671-76008671 on Build GRCh38
4069758 c.4917G>A p.S1639S Chr.17: 76008288-76008288 on Build GRCh38
6523905 c.4574G>T p.R1525L Chr.17: 76008631-76008631 on Build GRCh38
5530661 c.4619G>A p.W1540* Chr.17: 76008586-76008586 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377562936 Chr.17: 76008359-76008359 on Build GRCh38
rs774663667 Chr.17: 76008355-76008355 on Build GRCh38
rs779417063 Chr.17: 76008589-76008589 on Build GRCh38
rs780444775 Chr.17: 76008323-76008323 on Build GRCh38
rs759305677 Chr.17: 76008632-76008632 on Build GRCh38
rs866995164 Chr.17: 76008652-76008652 on Build GRCh38
rs148808819 Chr.17: 76008726-76008726 on Build GRCh38
rs753898829 Chr.17: 76008509-76008509 on Build GRCh38
rs750335053 Chr.17: 76008657-76008659 on Build GRCh38
rs764339679 Chr.17: 76008479-76008479 on Build GRCh38
rs777889344 Chr.17: 76008462-76008462 on Build GRCh38
rs901488371 Chr.17: 76008441-76008441 on Build GRCh38
rs941376493 Chr.17: 76008284-76008284 on Build GRCh38
rs781674876 Chr.17: 76008701-76008701 on Build GRCh38
rs555554617 Chr.17: 76008334-76008334 on Build GRCh38
rs202156490 Chr.17: 76008278-76008278 on Build GRCh38
rs770188677 Chr.17: 76008282-76008282 on Build GRCh38
rs951858556 Chr.17: 76008274-76008274 on Build GRCh38
rs767807959 Chr.17: 76008504-76008504 on Build GRCh38
rs757077717 Chr.17: 76008315-76008315 on Build GRCh38
rs763560367 Chr.17: 76008497-76008497 on Build GRCh38
rs766131192 Chr.17: 76008547-76008547 on Build GRCh38
rs753917041 Chr.17: 76008573-76008573 on Build GRCh38
rs550288224 Chr.17: 76008425-76008425 on Build GRCh38
rs776295618 Chr.17: 76008529-76008529 on Build GRCh38
rs750859177 Chr.17: 76008479-76008479 on Build GRCh38
rs756132170 Chr.17: 76008305-76008305 on Build GRCh38
rs1026002607 Chr.17: 76008457-76008457 on Build GRCh38
rs148554122 Chr.17: 76008532-76008532 on Build GRCh38
rs760944332 Chr.17: 76008475-76008476 on Build GRCh38
rs746169602 Chr.17: 76008480-76008480 on Build GRCh38
rs199755125 Chr.17: 76008557-76008557 on Build GRCh38
rs894998721 Chr.17: 76008580-76008580 on Build GRCh38
rs1052833673 Chr.17: 76008591-76008591 on Build GRCh38
rs199627871 Chr.17: 76008404-76008404 on Build GRCh38
rs750250014 Chr.17: 76008499-76008499 on Build GRCh38
rs760035712 Chr.17: 76008376-76008376 on Build GRCh38
rs1033972008 Chr.17: 76008473-76008473 on Build GRCh38
rs147437042 Chr.17: 76008590-76008590 on Build GRCh38
rs781208062 Chr.17: 76008440-76008440 on Build GRCh38
rs753669410 Chr.17: 76008312-76008312 on Build GRCh38
rs774027925 Chr.17: 76008630-76008630 on Build GRCh38
rs752141888 Chr.17: 76008304-76008304 on Build GRCh38
rs201217983 Chr.17: 76008493-76008493 on Build GRCh38
rs776783322 Chr.17: 76008648-76008648 on Build GRCh38
rs1030266308 Chr.17: 76008503-76008503 on Build GRCh38
rs539062061 Chr.17: 76008451-76008451 on Build GRCh38
rs746261242 Chr.17: 76008517-76008517 on Build GRCh38
rs757230950 Chr.17: 76008416-76008416 on Build GRCh38
rs866439899 Chr.17: 76008275-76008275 on Build GRCh38
rs547644932 Chr.17: 76008347-76008347 on Build GRCh38
rs775982970 Chr.17: 76008448-76008457 on Build GRCh38
rs964678260 Chr.17: 76008390-76008390 on Build GRCh38
rs773642532 Chr.17: 76008286-76008286 on Build GRCh38
rs867129386 Chr.17: 76008635-76008635 on Build GRCh38
rs747340763 Chr.17: 76008598-76008598 on Build GRCh38
rs747847879 Chr.17: 76008714-76008714 on Build GRCh38
rs966584955 Chr.17: 76008609-76008609 on Build GRCh38
rs777511755 Chr.17: 76008311-76008311 on Build GRCh38
rs749831530 Chr.17: 76008468-76008468 on Build GRCh38
rs944207499 Chr.17: 76008521-76008521 on Build GRCh38
rs146092902 Chr.17: 76008318-76008318 on Build GRCh38
rs1039147899 Chr.17: 76008643-76008645 on Build GRCh38
rs756324680 Chr.17: 76008505-76008505 on Build GRCh38
rs764241294 Chr.17: 76008567-76008567 on Build GRCh38
rs563372463 Chr.17: 76008536-76008536 on Build GRCh38
rs917873460 Chr.17: 76008333-76008333 on Build GRCh38
rs747276036 Chr.17: 76008326-76008326 on Build GRCh38
rs763165225 Chr.17: 76008287-76008287 on Build GRCh38
rs748770869 Chr.17: 76008722-76008722 on Build GRCh38
rs572601816 Chr.17: 76008615-76008615 on Build GRCh38
rs759163593 Chr.17: 76008291-76008291 on Build GRCh38
rs761341904 Chr.17: 76008530-76008530 on Build GRCh38
rs766845248 Chr.17: 76008686-76008686 on Build GRCh38
rs528132734 Chr.17: 76008688-76008688 on Build GRCh38
rs142844241 Chr.17: 76008354-76008354 on Build GRCh38
rs200073444 Chr.17: 76008671-76008671 on Build GRCh38
rs780341143 Chr.17: 76008593-76008593 on Build GRCh38
rs113394630 Chr.17: 76008519-76008519 on Build GRCh38
rs369166155 Chr.17: 76008281-76008281 on Build GRCh38
rs139365903 Chr.17: 76008647-76008647 on Build GRCh38
rs764088999 Chr.17: 76008327-76008327 on Build GRCh38
rs770775128 Chr.17: 76008723-76008723 on Build GRCh38
rs948686823 Chr.17: 76008537-76008537 on Build GRCh38
rs765264329 Chr.17: 76008646-76008647 on Build GRCh38
rs757772040 Chr.17: 76008510-76008510 on Build GRCh38
rs772338750 Chr.17: 76008434-76008434 on Build GRCh38
rs759249866 Chr.17: 76008357-76008357 on Build GRCh38
rs146378151 Chr.17: 76008523-76008523 on Build GRCh38
rs761906854 Chr.17: 76008643-76008643 on Build GRCh38
rs748292820 Chr.17: 76008449-76008449 on Build GRCh38
rs755265848 Chr.17: 76008430-76008430 on Build GRCh38
rs764329412 Chr.17: 76008643-76008643 on Build GRCh38
rs1006046510 Chr.17: 76008467-76008467 on Build GRCh38
rs1052757657 Chr.17: 76008348-76008348 on Build GRCh38
rs774963318 Chr.17: 76008494-76008494 on Build GRCh38
rs997021420 Chr.17: 76008452-76008452 on Build GRCh38
rs892895479 Chr.17: 76008602-76008602 on Build GRCh38
rs776277179 Chr.17: 76008498-76008498 on Build GRCh38
rs373960086 Chr.17: 76008716-76008716 on Build GRCh38
rs775027325 Chr.17: 76008322-76008322 on Build GRCh38
rs936809047 Chr.17: 76008353-76008353 on Build GRCh38
rs755349309 Chr.17: 76008695-76008695 on Build GRCh38
rs150027671 Chr.17: 76008683-76008683 on Build GRCh38
rs751361738 Chr.17: 76008548-76008548 on Build GRCh38
rs767051147 Chr.17: 76008292-76008292 on Build GRCh38
rs867668690 Chr.17: 76008644-76008644 on Build GRCh38
rs959240068 Chr.17: 76008478-76008478 on Build GRCh38
rs769481718 Chr.17: 76008617-76008617 on Build GRCh38
rs201429207 Chr.17: 76008629-76008629 on Build GRCh38
rs760018710 Chr.17: 76008377-76008378 on Build GRCh38
rs377216983 Chr.17: 76008661-76008661 on Build GRCh38
rs144390976 Chr.17: 76008684-76008684 on Build GRCh38
rs768199812 Chr.17: 76008599-76008599 on Build GRCh38
rs140054971 Chr.17: 76008340-76008340 on Build GRCh38
rs757285861 Chr.17: 76008577-76008577 on Build GRCh38
rs564557598 Chr.17: 76008601-76008601 on Build GRCh38
rs534116655 Chr.17: 76008514-76008514 on Build GRCh38
rs751902650 Chr.17: 76008689-76008689 on Build GRCh38
rs962803888 Chr.17: 76008713-76008713 on Build GRCh38
rs897387869 Chr.17: 76008533-76008533 on Build GRCh38
rs754041039 Chr.17: 76008556-76008557 on Build GRCh38
rs144149788 Chr.17: 76008625-76008625 on Build GRCh38
rs1022516170 Chr.17: 76008360-76008360 on Build GRCh38
rs570061775 Chr.17: 76008447-76008447 on Build GRCh38
rs200253222 Chr.17: 76008508-76008508 on Build GRCh38
rs535957560 Chr.17: 76008524-76008524 on Build GRCh38
rs777479943 Chr.17: 76008720-76008720 on Build GRCh38
rs372600391 Chr.17: 76008358-76008358 on Build GRCh38
rs748121990 Chr.17: 76008339-76008339 on Build GRCh38
rs868094270 Chr.17: 76008631-76008631 on Build GRCh38
rs572362721 Chr.17: 76008501-76008501 on Build GRCh38

View Full Product Details