Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82960945-82961450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNTL1
Entrez Gene ID
146712
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase-like 1
Gene Aliases
3-Gn-T8, B3GNT8, BGnT-8, beta-1, beta3Gn-T8, beta3GnTL1
UniGene
Hs.607824
Gene Chromosome Location
Chr.17: 82942149-83051845 on Build GRCh38
Transcripts
NM_001009905.2 NM_001320742.1 NM_001320743.1 NR_135465.1 NR_135466.1 XM_006722272.3 XM_011523532.2 XM_011523535.1 XM_017024205.1 XM_017024206.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6418967 c.715T>C p.C239R Chr.17: 82961067-82961067 on Build GRCh38
1710917 c.710C>G p.T237R Chr.17: 82961072-82961072 on Build GRCh38
4984867 c.704C>T p.A235V Chr.17: 82961078-82961078 on Build GRCh38
3523924 c.667G>A p.D223N Chr.17: 82961115-82961115 on Build GRCh38
4450416 c.601-1G>A p.? Chr.17: 82961182-82961182 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753004340 Chr.17: 82961211-82961211 on Build GRCh38
rs532299736 Chr.17: 82961355-82961355 on Build GRCh38
rs776022363 Chr.17: 82961106-82961106 on Build GRCh38
rs935937495 Chr.17: 82961396-82961396 on Build GRCh38
rs755576903 Chr.17: 82961227-82961227 on Build GRCh38
rs202145788 Chr.17: 82961098-82961098 on Build GRCh38
rs553723708 Chr.17: 82960992-82960992 on Build GRCh38
rs185300831 Chr.17: 82960997-82960997 on Build GRCh38
rs539854147 Chr.17: 82961310-82961310 on Build GRCh38
rs888127642 Chr.17: 82960999-82960999 on Build GRCh38
rs780761733 Chr.17: 82961093-82961093 on Build GRCh38
rs145422923 Chr.17: 82961072-82961072 on Build GRCh38
rs372601883 Chr.17: 82961037-82961037 on Build GRCh38
rs775355207 Chr.17: 82961173-82961173 on Build GRCh38
rs564841477 Chr.17: 82961086-82961086 on Build GRCh38
rs906567402 Chr.17: 82961332-82961332 on Build GRCh38
rs866299734 Chr.17: 82961333-82961333 on Build GRCh38
rs922268258 Chr.17: 82961364-82961364 on Build GRCh38
rs1012136711 Chr.17: 82961416-82961416 on Build GRCh38
rs1049673788 Chr.17: 82961265-82961265 on Build GRCh38
rs201021215 Chr.17: 82961078-82961078 on Build GRCh38
rs201686706 Chr.17: 82961130-82961130 on Build GRCh38
rs866413289 Chr.17: 82960985-82960985 on Build GRCh38
rs139832462 Chr.17: 82961266-82961266 on Build GRCh38
rs985332552 Chr.17: 82961382-82961382 on Build GRCh38
rs200253539 Chr.17: 82961096-82961096 on Build GRCh38
rs1033963481 Chr.17: 82961339-82961339 on Build GRCh38
rs914072840 Chr.17: 82960979-82960979 on Build GRCh38
rs1017520493 Chr.17: 82961345-82961345 on Build GRCh38
rs988446859 Chr.17: 82960971-82960971 on Build GRCh38
rs369421536 Chr.17: 82961034-82961034 on Build GRCh38
rs371962087 Chr.17: 82961030-82961030 on Build GRCh38
rs915888613 Chr.17: 82961281-82961281 on Build GRCh38
rs374979627 Chr.17: 82961009-82961009 on Build GRCh38
rs765834239 Chr.17: 82961207-82961207 on Build GRCh38
rs375357344 Chr.17: 82961176-82961176 on Build GRCh38
rs777594830 Chr.17: 82961002-82961002 on Build GRCh38
rs959484370 Chr.17: 82961412-82961418 on Build GRCh38
rs769260899 Chr.17: 82961257-82961257 on Build GRCh38
rs759108497 Chr.17: 82961115-82961115 on Build GRCh38
rs202197391 Chr.17: 82961044-82961044 on Build GRCh38
rs983925891 Chr.17: 82961089-82961089 on Build GRCh38
rs543985120 Chr.17: 82961365-82961365 on Build GRCh38
rs756702102 Chr.17: 82961092-82961092 on Build GRCh38
rs376226349 Chr.17: 82961226-82961226 on Build GRCh38
rs747556194 Chr.17: 82961239-82961239 on Build GRCh38
rs982253391 Chr.17: 82961069-82961069 on Build GRCh38
rs1048495387 Chr.17: 82960998-82960998 on Build GRCh38
rs148004417 Chr.17: 82961178-82961178 on Build GRCh38
rs939929830 Chr.17: 82961001-82961001 on Build GRCh38
rs770370413 Chr.17: 82961059-82961059 on Build GRCh38
rs200668312 Chr.17: 82961203-82961203 on Build GRCh38
rs141479851 Chr.17: 82961146-82961146 on Build GRCh38
rs780891865 Chr.17: 82961157-82961157 on Build GRCh38
rs371588569 Chr.17: 82961221-82961221 on Build GRCh38
rs776592296 Chr.17: 82961064-82961064 on Build GRCh38
rs572705594 Chr.17: 82961294-82961294 on Build GRCh38
rs915906851 Chr.17: 82961154-82961156 on Build GRCh38
rs759478290 Chr.17: 82961065-82961065 on Build GRCh38
rs564831800 Chr.17: 82961340-82961340 on Build GRCh38
rs746380517 Chr.17: 82961039-82961039 on Build GRCh38
rs757318927 Chr.17: 82961036-82961036 on Build GRCh38
rs377431775 Chr.17: 82960986-82960986 on Build GRCh38
rs1032624196 Chr.17: 82961260-82961260 on Build GRCh38
rs1024518561 Chr.17: 82960970-82960970 on Build GRCh38
rs753770494 Chr.17: 82961011-82961011 on Build GRCh38
rs997944678 Chr.17: 82961028-82961028 on Build GRCh38
rs375983406 Chr.17: 82961134-82961134 on Build GRCh38
rs138531570 Chr.17: 82961121-82961121 on Build GRCh38
rs540644774 Chr.17: 82961228-82961228 on Build GRCh38
rs962282103 Chr.17: 82960978-82960978 on Build GRCh38
rs145393038 Chr.17: 82961151-82961151 on Build GRCh38
rs746618486 Chr.17: 82961155-82961155 on Build GRCh38
rs367926270 Chr.17: 82961029-82961029 on Build GRCh38
rs773606774 Chr.17: 82961190-82961190 on Build GRCh38
rs745354692 Chr.17: 82961162-82961162 on Build GRCh38
rs896438804 Chr.17: 82961403-82961403 on Build GRCh38
rs541399980 Chr.17: 82961382-82961382 on Build GRCh38
rs545627717 Chr.17: 82961007-82961007 on Build GRCh38
rs997123462 Chr.17: 82961366-82961366 on Build GRCh38
rs192413231 Chr.17: 82961094-82961094 on Build GRCh38
rs762711617 Chr.17: 82961120-82961120 on Build GRCh38
rs376882588 Chr.17: 82961033-82961033 on Build GRCh38
rs760789715 Chr.17: 82961191-82961191 on Build GRCh38
rs770409462 Chr.17: 82961100-82961100 on Build GRCh38
rs751775590 Chr.17: 82961076-82961076 on Build GRCh38
rs767067875 Chr.17: 82961006-82961006 on Build GRCh38
rs370798688 Chr.17: 82961164-82961164 on Build GRCh38
rs1005694740 Chr.17: 82961342-82961342 on Build GRCh38
rs562508596 Chr.17: 82961381-82961381 on Build GRCh38
rs541423458 Chr.17: 82961108-82961109 on Build GRCh38
rs756850040 Chr.17: 82961156-82961156 on Build GRCh38
rs754330521 Chr.17: 82961198-82961198 on Build GRCh38
rs754719769 Chr.17: 82961020-82961020 on Build GRCh38
rs757309602 Chr.17: 82961077-82961077 on Build GRCh38
rs950044363 Chr.17: 82961290-82961290 on Build GRCh38
rs773513113 Chr.17: 82961311-82961311 on Build GRCh38
rs150669869 Chr.17: 82961040-82961040 on Build GRCh38
rs893868530 Chr.17: 82961299-82961299 on Build GRCh38
rs754420546 Chr.17: 82961149-82961149 on Build GRCh38
rs767626973 Chr.17: 82961179-82961179 on Build GRCh38
rs375629511 Chr.17: 82961081-82961081 on Build GRCh38
rs769339281 Chr.17: 82961166-82961166 on Build GRCh38
rs542325165 Chr.17: 82961238-82961238 on Build GRCh38
rs970930848 Chr.17: 82961165-82961165 on Build GRCh38
rs751762558 Chr.17: 82961127-82961127 on Build GRCh38
rs145143559 Chr.17: 82961119-82961119 on Build GRCh38
rs933899336 Chr.17: 82961258-82961258 on Build GRCh38
rs563915065 Chr.17: 82961409-82961409 on Build GRCh38
rs765047984 Chr.17: 82961022-82961022 on Build GRCh38
rs369103228 Chr.17: 82961370-82961370 on Build GRCh38
rs149224289 Chr.17: 82961118-82961118 on Build GRCh38
rs529920238 Chr.17: 82961393-82961393 on Build GRCh38
rs764342701 Chr.17: 82961074-82961074 on Build GRCh38
rs777108247 Chr.17: 82961280-82961280 on Build GRCh38
rs1020721822 Chr.17: 82961125-82961125 on Build GRCh38
rs567523605 Chr.17: 82961131-82961131 on Build GRCh38
rs758839747 Chr.17: 82961213-82961213 on Build GRCh38
rs761944601 Chr.17: 82961128-82961128 on Build GRCh38
rs553662197 Chr.17: 82961152-82961152 on Build GRCh38
rs370187811 Chr.17: 82961043-82961043 on Build GRCh38
rs760158723 Chr.17: 82961008-82961008 on Build GRCh38
rs1032138419 Chr.17: 82961409-82961409 on Build GRCh38
rs374980165 Chr.17: 82961071-82961071 on Build GRCh38
rs972488528 Chr.17: 82961356-82961356 on Build GRCh38
rs572010424 Chr.17: 82961174-82961174 on Build GRCh38
rs1049514844 Chr.17: 82961129-82961129 on Build GRCh38
rs550306136 Chr.17: 82961058-82961058 on Build GRCh38
rs756328004 Chr.17: 82961133-82961133 on Build GRCh38
rs749631176 Chr.17: 82961336-82961336 on Build GRCh38

View Full Product Details