Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31622539-31623054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT6
Entrez Gene ID
9331
Gene Name
beta-1,4-galactosyltransferase 6
Gene Aliases
B4Gal-T6, beta4Gal-T6
UniGene
Hs.591063
Gene Chromosome Location
Chr.18: 31622246-31724670 on Build GRCh38
Transcripts
NM_004775.3 XM_005258387.4 XM_006722578.3 XM_006722579.3 XM_017026088.1 XM_017026089.1 XM_017026090.1 XM_017026091.1 XM_017026092.1

SNP Detail

SNP ID Chromosome Location
rs536068413 Chr.18: 31622653-31622653 on Build GRCh38
rs951928098 Chr.18: 31622611-31622611 on Build GRCh38
rs201948853 Chr.18: 31622656-31622656 on Build GRCh38
rs1049861687 Chr.18: 31622907-31622907 on Build GRCh38
rs137938811 Chr.18: 31622867-31622867 on Build GRCh38
rs187794863 Chr.18: 31622793-31622793 on Build GRCh38
rs925149076 Chr.18: 31622742-31622743 on Build GRCh38
rs986281277 Chr.18: 31622965-31622965 on Build GRCh38
rs549070624 Chr.18: 31622964-31622964 on Build GRCh38
rs940323809 Chr.18: 31622811-31622811 on Build GRCh38
rs990599918 Chr.18: 31622753-31622753 on Build GRCh38
rs569628554 Chr.18: 31622952-31622952 on Build GRCh38
rs576069391 Chr.18: 31622789-31622789 on Build GRCh38
rs932150851 Chr.18: 31622675-31622675 on Build GRCh38
rs551738913 Chr.18: 31622926-31622926 on Build GRCh38
rs761549628 Chr.18: 31622659-31622659 on Build GRCh38
rs193170324 Chr.18: 31622887-31622887 on Build GRCh38
rs938647432 Chr.18: 31623024-31623024 on Build GRCh38
rs923522289 Chr.18: 31622927-31622927 on Build GRCh38
rs546207991 Chr.18: 31622699-31622699 on Build GRCh38
rs547190829 Chr.18: 31623000-31623000 on Build GRCh38
rs951797393 Chr.18: 31622879-31622879 on Build GRCh38
rs143811512 Chr.18: 31622809-31622809 on Build GRCh38
rs529053166 Chr.18: 31622819-31622819 on Build GRCh38
rs949075844 Chr.18: 31622775-31622775 on Build GRCh38
rs1012081172 Chr.18: 31622848-31622848 on Build GRCh38
rs1014521497 Chr.18: 31622901-31622901 on Build GRCh38
rs984950228 Chr.18: 31622891-31622891 on Build GRCh38
rs115186394 Chr.18: 31622962-31622962 on Build GRCh38
rs772818932 Chr.18: 31623016-31623016 on Build GRCh38
rs955429447 Chr.18: 31622672-31622672 on Build GRCh38
rs568622263 Chr.18: 31622632-31622632 on Build GRCh38
rs903294657 Chr.18: 31623011-31623011 on Build GRCh38
rs975749453 Chr.18: 31622914-31622914 on Build GRCh38
rs17803165 Chr.18: 31622988-31622988 on Build GRCh38
rs568732092 Chr.18: 31622715-31622718 on Build GRCh38
rs183304335 Chr.18: 31622572-31622572 on Build GRCh38
rs898695366 Chr.18: 31622838-31622838 on Build GRCh38
rs140359087 Chr.18: 31623008-31623008 on Build GRCh38
rs116203117 Chr.18: 31622655-31622655 on Build GRCh38
rs961701014 Chr.18: 31622912-31622912 on Build GRCh38
rs893211535 Chr.18: 31622690-31622690 on Build GRCh38
rs917689672 Chr.18: 31622769-31622769 on Build GRCh38
rs937810198 Chr.18: 31622745-31622745 on Build GRCh38
rs557728644 Chr.18: 31622784-31622784 on Build GRCh38
rs916315600 Chr.18: 31622589-31622589 on Build GRCh38
rs1008694387 Chr.18: 31622959-31622959 on Build GRCh38
rs572684121 Chr.18: 31622678-31622678 on Build GRCh38
rs984467749 Chr.18: 31622613-31622613 on Build GRCh38
rs903201606 Chr.18: 31622719-31622719 on Build GRCh38
rs1017135789 Chr.18: 31622592-31622592 on Build GRCh38
rs576000830 Chr.18: 31622728-31622733 on Build GRCh38
rs555510363 Chr.18: 31622597-31622597 on Build GRCh38
rs143416107 Chr.18: 31622911-31622911 on Build GRCh38
rs1041699300 Chr.18: 31622701-31622701 on Build GRCh38
rs898665778 Chr.18: 31622600-31622600 on Build GRCh38
rs147177441 Chr.18: 31622995-31622995 on Build GRCh38

View Full Product Details