Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2476391-2476910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GADD45B
Entrez Gene ID
4616
Gene Name
growth arrest and DNA damage inducible beta
Gene Aliases
GADD45BETA, MYD118
UniGene
Hs.110571
Gene Chromosome Location
Chr.19: 2476125-2478259 on Build GRCh38
Transcripts
NM_015675.3 XM_017026822.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3357107 c.55G>C p.V19L Chr.19: 2476539-2476539 on Build GRCh38
6198682 c.91C>A p.Q31K Chr.19: 2476575-2476575 on Build GRCh38
1190930 c.42_44+1delGAAG p.? Chr.19: 2476400-2476403 on Build GRCh38
6988925 c.106C>T p.L36F Chr.19: 2476590-2476590 on Build GRCh38
5418635 c.122A>T p.Y41F Chr.19: 2476606-2476606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759168082 Chr.19: 2476846-2476846 on Build GRCh38
rs775132438 Chr.19: 2476483-2476483 on Build GRCh38
rs3783497 Chr.19: 2476787-2476787 on Build GRCh38
rs748068108 Chr.19: 2476671-2476671 on Build GRCh38
rs920866062 Chr.19: 2476824-2476824 on Build GRCh38
rs748563947 Chr.19: 2476417-2476417 on Build GRCh38
rs1045355581 Chr.19: 2476450-2476450 on Build GRCh38
rs771815739 Chr.19: 2476526-2476526 on Build GRCh38
rs187488334 Chr.19: 2476860-2476860 on Build GRCh38
rs377490192 Chr.19: 2476593-2476593 on Build GRCh38
rs767719964 Chr.19: 2476597-2476597 on Build GRCh38
rs548744711 Chr.19: 2476795-2476795 on Build GRCh38
rs751761346 Chr.19: 2476584-2476584 on Build GRCh38
rs759274501 Chr.19: 2476425-2476425 on Build GRCh38
rs941033641 Chr.19: 2476806-2476806 on Build GRCh38
rs746660483 Chr.19: 2476653-2476653 on Build GRCh38
rs3783498 Chr.19: 2476792-2476792 on Build GRCh38
rs958529587 Chr.19: 2476482-2476482 on Build GRCh38
rs753186019 Chr.19: 2476598-2476598 on Build GRCh38
rs3783496 Chr.19: 2476718-2476718 on Build GRCh38
rs938208423 Chr.19: 2476715-2476715 on Build GRCh38
rs770224212 Chr.19: 2476518-2476518 on Build GRCh38
rs576751807 Chr.19: 2476505-2476505 on Build GRCh38
rs145778154 Chr.19: 2476437-2476438 on Build GRCh38
rs776500657 Chr.19: 2476559-2476559 on Build GRCh38
rs755302110 Chr.19: 2476638-2476639 on Build GRCh38
rs753595292 Chr.19: 2476488-2476488 on Build GRCh38
rs932130696 Chr.19: 2476825-2476825 on Build GRCh38
rs544782085 Chr.19: 2476484-2476484 on Build GRCh38
rs776192378 Chr.19: 2476667-2476667 on Build GRCh38
rs533308934 Chr.19: 2476757-2476757 on Build GRCh38
rs766638467 Chr.19: 2476452-2476453 on Build GRCh38
rs11541535 Chr.19: 2476583-2476583 on Build GRCh38
rs768079196 Chr.19: 2476545-2476545 on Build GRCh38
rs773051830 Chr.19: 2476572-2476572 on Build GRCh38
rs940992930 Chr.19: 2476543-2476543 on Build GRCh38
rs567426077 Chr.19: 2476799-2476799 on Build GRCh38
rs1024048728 Chr.19: 2476785-2476785 on Build GRCh38
rs530650211 Chr.19: 2476793-2476793 on Build GRCh38
rs986821140 Chr.19: 2476678-2476678 on Build GRCh38
rs567433567 Chr.19: 2476415-2476415 on Build GRCh38
rs939642617 Chr.19: 2476764-2476764 on Build GRCh38
rs1020846303 Chr.19: 2476859-2476859 on Build GRCh38
rs3783495 Chr.19: 2476447-2476448 on Build GRCh38
rs908113275 Chr.19: 2476517-2476517 on Build GRCh38
rs1046756200 Chr.19: 2476512-2476512 on Build GRCh38
rs375019788 Chr.19: 2476499-2476499 on Build GRCh38
rs541678798 Chr.19: 2476733-2476733 on Build GRCh38
rs112291544 Chr.19: 2476632-2476632 on Build GRCh38
rs886888778 Chr.19: 2476475-2476475 on Build GRCh38
rs925814760 Chr.19: 2476468-2476468 on Build GRCh38
rs763067262 Chr.19: 2476574-2476574 on Build GRCh38
rs202192151 Chr.19: 2476489-2476489 on Build GRCh38
rs191275929 Chr.19: 2476742-2476742 on Build GRCh38
rs773547471 Chr.19: 2476429-2476431 on Build GRCh38
rs373430316 Chr.19: 2476568-2476568 on Build GRCh38
rs771801287 Chr.19: 2476887-2476887 on Build GRCh38
rs757952919 Chr.19: 2476644-2476644 on Build GRCh38
rs1020952778 Chr.19: 2476689-2476689 on Build GRCh38
rs753361521 Chr.19: 2476493-2476493 on Build GRCh38
rs747471704 Chr.19: 2476412-2476412 on Build GRCh38
rs756751009 Chr.19: 2476601-2476601 on Build GRCh38
rs745511007 Chr.19: 2476522-2476522 on Build GRCh38
rs1050449706 Chr.19: 2476661-2476661 on Build GRCh38
rs746565575 Chr.19: 2476536-2476536 on Build GRCh38
rs1045407768 Chr.19: 2476734-2476734 on Build GRCh38
rs749711441 Chr.19: 2476643-2476643 on Build GRCh38
rs1037922107 Chr.19: 2476549-2476549 on Build GRCh38
rs750580180 Chr.19: 2476496-2476496 on Build GRCh38
rs769267957 Chr.19: 2476413-2476413 on Build GRCh38
rs370255741 Chr.19: 2476506-2476506 on Build GRCh38
rs758717181 Chr.19: 2476504-2476504 on Build GRCh38
rs764136373 Chr.19: 2476486-2476486 on Build GRCh38
rs766663792 Chr.19: 2476578-2476578 on Build GRCh38
rs562399667 Chr.19: 2476662-2476662 on Build GRCh38
rs899307907 Chr.19: 2476580-2476580 on Build GRCh38
rs200157147 Chr.19: 2476443-2476443 on Build GRCh38
rs1008139855 Chr.19: 2476828-2476828 on Build GRCh38
rs770460270 Chr.19: 2476494-2476494 on Build GRCh38
rs375169076 Chr.19: 2476419-2476419 on Build GRCh38
rs778785769 Chr.19: 2476707-2476707 on Build GRCh38
rs544016078 Chr.19: 2476604-2476604 on Build GRCh38
rs1020600024 Chr.19: 2476439-2476439 on Build GRCh38
rs979544590 Chr.19: 2476700-2476700 on Build GRCh38
rs538630599 Chr.19: 2476876-2476876 on Build GRCh38
rs571203018 Chr.19: 2476864-2476864 on Build GRCh38
rs765015136 Chr.19: 2476570-2476570 on Build GRCh38
rs779713116 Chr.19: 2476648-2476648 on Build GRCh38
rs754172340 Chr.19: 2476510-2476535 on Build GRCh38
rs1050581121 Chr.19: 2476827-2476827 on Build GRCh38
rs781696614 Chr.19: 2476511-2476511 on Build GRCh38
rs774836462 Chr.19: 2476531-2476531 on Build GRCh38
rs967347954 Chr.19: 2476463-2476463 on Build GRCh38
rs778268064 Chr.19: 2476641-2476641 on Build GRCh38
rs967229327 Chr.19: 2476691-2476691 on Build GRCh38
rs1057003664 Chr.19: 2476818-2476818 on Build GRCh38
rs897053540 Chr.19: 2476823-2476823 on Build GRCh38
rs978993095 Chr.19: 2476464-2476464 on Build GRCh38
rs552714984 Chr.19: 2476422-2476422 on Build GRCh38
rs769776936 Chr.19: 2476673-2476673 on Build GRCh38
rs755410215 Chr.19: 2476510-2476510 on Build GRCh38
rs183137231 Chr.19: 2476820-2476820 on Build GRCh38
rs898209221 Chr.19: 2476770-2476770 on Build GRCh38
rs1038130160 Chr.19: 2476769-2476769 on Build GRCh38
rs979814864 Chr.19: 2476886-2476886 on Build GRCh38
rs778286178 Chr.19: 2476521-2476521 on Build GRCh38
rs918193045 Chr.19: 2476722-2476722 on Build GRCh38
rs543408242 Chr.19: 2476836-2476836 on Build GRCh38

View Full Product Details