Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3136310-3136846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNA15
Entrez Gene ID
2769
Gene Name
G protein subunit alpha 15
Gene Aliases
GNA16
UniGene
Hs.73797
Gene Chromosome Location
Chr.19: 3136032-3163769 on Build GRCh38
Transcripts
NM_002068.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1208392 c.108G>T p.K36N Chr.19: 3136558-3136558 on Build GRCh38
4884085 c.4G>T p.A2S Chr.19: 3136454-3136454 on Build GRCh38
1711603 c.144G>C p.L48F Chr.19: 3136594-3136594 on Build GRCh38
3668236 c.63C>A p.A21A Chr.19: 3136513-3136513 on Build GRCh38
5718779 c.61G>A p.A21T Chr.19: 3136511-3136511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372463760 Chr.19: 3136616-3136616 on Build GRCh38
rs968683586 Chr.19: 3136680-3136680 on Build GRCh38
rs766182128 Chr.19: 3136571-3136571 on Build GRCh38
rs763604036 Chr.19: 3136426-3136426 on Build GRCh38
rs577523583 Chr.19: 3136411-3136411 on Build GRCh38
rs900777389 Chr.19: 3136622-3136622 on Build GRCh38
rs921752713 Chr.19: 3136355-3136355 on Build GRCh38
rs567578119 Chr.19: 3136357-3136357 on Build GRCh38
rs776216111 Chr.19: 3136433-3136433 on Build GRCh38
rs979316332 Chr.19: 3136501-3136501 on Build GRCh38
rs768354703 Chr.19: 3136551-3136551 on Build GRCh38
rs940701272 Chr.19: 3136387-3136387 on Build GRCh38
rs749833575 Chr.19: 3136517-3136517 on Build GRCh38
rs567762404 Chr.19: 3136808-3136808 on Build GRCh38
rs115417983 Chr.19: 3136391-3136391 on Build GRCh38
rs761540533 Chr.19: 3136568-3136568 on Build GRCh38
rs556784328 Chr.19: 3136474-3136474 on Build GRCh38
rs766240650 Chr.19: 3136495-3136495 on Build GRCh38
rs752976720 Chr.19: 3136617-3136617 on Build GRCh38
rs371766518 Chr.19: 3136513-3136513 on Build GRCh38
rs751274515 Chr.19: 3136498-3136498 on Build GRCh38
rs941710074 Chr.19: 3136613-3136613 on Build GRCh38
rs972520780 Chr.19: 3136809-3136809 on Build GRCh38
rs763019719 Chr.19: 3136570-3136570 on Build GRCh38
rs781728223 Chr.19: 3136649-3136649 on Build GRCh38
rs997460342 Chr.19: 3136778-3136782 on Build GRCh38
rs75723333 Chr.19: 3136458-3136458 on Build GRCh38
rs374281001 Chr.19: 3136510-3136510 on Build GRCh38
rs411210 Chr.19: 3136694-3136694 on Build GRCh38
rs411474 Chr.19: 3136800-3136800 on Build GRCh38
rs866766675 Chr.19: 3136712-3136712 on Build GRCh38
rs755254026 Chr.19: 3136500-3136500 on Build GRCh38
rs867190636 Chr.19: 3136446-3136446 on Build GRCh38
rs1055275564 Chr.19: 3136382-3136382 on Build GRCh38
rs937884858 Chr.19: 3136381-3136381 on Build GRCh38
rs964868296 Chr.19: 3136427-3136427 on Build GRCh38
rs771387678 Chr.19: 3136518-3136518 on Build GRCh38
rs1045703679 Chr.19: 3136616-3136617 on Build GRCh38
rs953163545 Chr.19: 3136462-3136462 on Build GRCh38
rs774548264 Chr.19: 3136410-3136410 on Build GRCh38
rs879906024 Chr.19: 3136642-3136643 on Build GRCh38
rs892924949 Chr.19: 3136674-3136674 on Build GRCh38
rs1036346735 Chr.19: 3136389-3136389 on Build GRCh38
rs549281796 Chr.19: 3136803-3136803 on Build GRCh38
rs376509314 Chr.19: 3136472-3136472 on Build GRCh38
rs768015037 Chr.19: 3136374-3136374 on Build GRCh38
rs996390197 Chr.19: 3136428-3136428 on Build GRCh38
rs1637665 Chr.19: 3136643-3136643 on Build GRCh38
rs781127081 Chr.19: 3136802-3136802 on Build GRCh38
rs376134188 Chr.19: 3136789-3136789 on Build GRCh38
rs112493638 Chr.19: 3136351-3136351 on Build GRCh38
rs1013651705 Chr.19: 3136398-3136398 on Build GRCh38
rs867164143 Chr.19: 3136356-3136356 on Build GRCh38
rs1009932719 Chr.19: 3136689-3136689 on Build GRCh38
rs758392333 Chr.19: 3136494-3136494 on Build GRCh38
rs905321287 Chr.19: 3136405-3136405 on Build GRCh38
rs909320865 Chr.19: 3136482-3136482 on Build GRCh38
rs200281819 Chr.19: 3136531-3136531 on Build GRCh38
rs370588174 Chr.19: 3136375-3136375 on Build GRCh38
rs411453 Chr.19: 3136771-3136771 on Build GRCh38
rs368219827 Chr.19: 3136473-3136473 on Build GRCh38
rs772534730 Chr.19: 3136730-3136730 on Build GRCh38
rs564829431 Chr.19: 3136648-3136648 on Build GRCh38
rs754663163 Chr.19: 3136609-3136609 on Build GRCh38
rs752212338 Chr.19: 3136623-3136624 on Build GRCh38
rs386805964 Chr.19: 3136800-3136802 on Build GRCh38
rs866418527 Chr.19: 3136709-3136709 on Build GRCh38
rs925118575 Chr.19: 3136378-3136378 on Build GRCh38
rs751384202 Chr.19: 3136587-3136587 on Build GRCh38
rs923416738 Chr.19: 3136348-3136348 on Build GRCh38
rs371765154 Chr.19: 3136516-3136516 on Build GRCh38
rs751170883 Chr.19: 3136661-3136661 on Build GRCh38
rs777860419 Chr.19: 3136638-3136638 on Build GRCh38
rs756296808 Chr.19: 3136620-3136620 on Build GRCh38
rs1049135309 Chr.19: 3136372-3136372 on Build GRCh38
rs1047404912 Chr.19: 3136404-3136404 on Build GRCh38
rs764892056 Chr.19: 3136569-3136569 on Build GRCh38
rs746158062 Chr.19: 3136550-3136550 on Build GRCh38
rs866327080 Chr.19: 3136528-3136528 on Build GRCh38
rs959336680 Chr.19: 3136733-3136733 on Build GRCh38
rs369043343 Chr.19: 3136563-3136563 on Build GRCh38
rs1032203022 Chr.19: 3136724-3136724 on Build GRCh38
rs374928184 Chr.19: 3136514-3136514 on Build GRCh38
rs1029354453 Chr.19: 3136786-3136786 on Build GRCh38
rs370713281 Chr.19: 3136665-3136665 on Build GRCh38
rs537198789 Chr.19: 3136349-3136349 on Build GRCh38
rs1053862184 Chr.19: 3136672-3136672 on Build GRCh38

View Full Product Details