Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12891406-12891902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCDH
Entrez Gene ID
2639
Gene Name
glutaryl-CoA dehydrogenase
Gene Aliases
ACAD5, GCD
UniGene
Hs.532699
Gene Chromosome Location
Chr.19: 12891129-12915345 on Build GRCh38
Transcripts
NM_000159.3 NM_013976.3 NR_102316.1 NR_102317.1 XM_006722721.2 XM_011527899.2 XM_011527900.2 XM_017026580.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3822041 c.155A>G p.D52G Chr.19: 12891858-12891858 on Build GRCh38
4686601 c.125A>G p.K42R Chr.19: 12891520-12891520 on Build GRCh38
991526 c.93G>T p.E31D Chr.19: 12891488-12891488 on Build GRCh38
6669919 c.101G>A p.G34E Chr.19: 12891496-12891496 on Build GRCh38
6388925 c.134G>A p.R45H Chr.19: 12891837-12891837 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754542775 Chr.19: 12891761-12891761 on Build GRCh38
rs564016367 Chr.19: 12891441-12891441 on Build GRCh38
rs1057516412 Chr.19: 12891875-12891875 on Build GRCh38
rs933702964 Chr.19: 12891711-12891711 on Build GRCh38
rs749490439 Chr.19: 12891578-12891578 on Build GRCh38
rs760596722 Chr.19: 12891592-12891592 on Build GRCh38
rs760536298 Chr.19: 12891528-12891528 on Build GRCh38
rs764813144 Chr.19: 12891567-12891570 on Build GRCh38
rs776355205 Chr.19: 12891699-12891699 on Build GRCh38
rs775583509 Chr.19: 12891591-12891591 on Build GRCh38
rs1040493033 Chr.19: 12891560-12891560 on Build GRCh38
rs761411060 Chr.19: 12891615-12891615 on Build GRCh38
rs572680595 Chr.19: 12891574-12891574 on Build GRCh38
rs755546201 Chr.19: 12891546-12891546 on Build GRCh38
rs749637416 Chr.19: 12891665-12891665 on Build GRCh38
rs773055912 Chr.19: 12891634-12891634 on Build GRCh38
rs764954552 Chr.19: 12891784-12891784 on Build GRCh38
rs772284492 Chr.19: 12891676-12891676 on Build GRCh38
rs758646992 Chr.19: 12891851-12891851 on Build GRCh38
rs753621195 Chr.19: 12891595-12891595 on Build GRCh38
rs775673559 Chr.19: 12891678-12891678 on Build GRCh38
rs376124947 Chr.19: 12891469-12891469 on Build GRCh38
rs763827330 Chr.19: 12891593-12891593 on Build GRCh38
rs759599442 Chr.19: 12891505-12891505 on Build GRCh38
rs980499037 Chr.19: 12891698-12891698 on Build GRCh38
rs751044366 Chr.19: 12891790-12891790 on Build GRCh38
rs888068604 Chr.19: 12891603-12891603 on Build GRCh38
rs578242746 Chr.19: 12891538-12891538 on Build GRCh38
rs748595546 Chr.19: 12891565-12891565 on Build GRCh38
rs752609314 Chr.19: 12891512-12891512 on Build GRCh38
rs746944518 Chr.19: 12891685-12891685 on Build GRCh38
rs532241386 Chr.19: 12891757-12891757 on Build GRCh38
rs767535651 Chr.19: 12891509-12891509 on Build GRCh38
rs200145853 Chr.19: 12891809-12891809 on Build GRCh38
rs772854475 Chr.19: 12891786-12891786 on Build GRCh38
rs778420613 Chr.19: 12891443-12891443 on Build GRCh38
rs1037898844 Chr.19: 12891736-12891736 on Build GRCh38
rs776643576 Chr.19: 12891872-12891872 on Build GRCh38
rs1031082837 Chr.19: 12891856-12891856 on Build GRCh38
rs758133485 Chr.19: 12891698-12891698 on Build GRCh38
rs781654177 Chr.19: 12891555-12891555 on Build GRCh38
rs765892776 Chr.19: 12891789-12891789 on Build GRCh38
rs1012645652 Chr.19: 12891455-12891455 on Build GRCh38
rs185661323 Chr.19: 12891473-12891473 on Build GRCh38
rs538220975 Chr.19: 12891480-12891480 on Build GRCh38
rs922305439 Chr.19: 12891646-12891646 on Build GRCh38
rs749991534 Chr.19: 12891616-12891616 on Build GRCh38
rs761852050 Chr.19: 12891581-12891581 on Build GRCh38
rs539354912 Chr.19: 12891543-12891543 on Build GRCh38
rs76650855 Chr.19: 12891752-12891752 on Build GRCh38
rs373149331 Chr.19: 12891438-12891438 on Build GRCh38
rs371026901 Chr.19: 12891539-12891539 on Build GRCh38
rs142568445 Chr.19: 12891531-12891531 on Build GRCh38
rs1024035620 Chr.19: 12891755-12891755 on Build GRCh38
rs201970551 Chr.19: 12891834-12891834 on Build GRCh38
rs1006150317 Chr.19: 12891875-12891875 on Build GRCh38
rs769128625 Chr.19: 12891465-12891465 on Build GRCh38
rs765806472 Chr.19: 12891620-12891620 on Build GRCh38
rs762549022 Chr.19: 12891788-12891788 on Build GRCh38
rs771159682 Chr.19: 12891580-12891580 on Build GRCh38
rs373749152 Chr.19: 12891499-12891499 on Build GRCh38
rs761653702 Chr.19: 12891701-12891701 on Build GRCh38
rs886893711 Chr.19: 12891728-12891728 on Build GRCh38
rs377485394 Chr.19: 12891520-12891520 on Build GRCh38
rs779267319 Chr.19: 12891669-12891669 on Build GRCh38
rs759689270 Chr.19: 12891584-12891584 on Build GRCh38
rs763622379 Chr.19: 12891704-12891705 on Build GRCh38
rs770111154 Chr.19: 12891472-12891472 on Build GRCh38
rs761099433 Chr.19: 12891556-12891556 on Build GRCh38
rs532647383 Chr.19: 12891697-12891697 on Build GRCh38
rs533991873 Chr.19: 12891439-12891439 on Build GRCh38
rs2242517 Chr.19: 12891749-12891749 on Build GRCh38
rs754165281 Chr.19: 12891579-12891579 on Build GRCh38
rs757492521 Chr.19: 12891668-12891668 on Build GRCh38
rs561029001 Chr.19: 12891670-12891670 on Build GRCh38
rs752521726 Chr.19: 12891426-12891426 on Build GRCh38
rs781576170 Chr.19: 12891468-12891468 on Build GRCh38
rs1006415164 Chr.19: 12891627-12891627 on Build GRCh38
rs144678094 Chr.19: 12891832-12891832 on Build GRCh38
rs369789314 Chr.19: 12891429-12891429 on Build GRCh38
rs770942722 Chr.19: 12891497-12891497 on Build GRCh38
rs780332187 Chr.19: 12891858-12891858 on Build GRCh38
rs752133601 Chr.19: 12891633-12891634 on Build GRCh38
rs943857937 Chr.19: 12891750-12891750 on Build GRCh38
rs747546938 Chr.19: 12891458-12891458 on Build GRCh38
rs778044164 Chr.19: 12891653-12891653 on Build GRCh38
rs757915352 Chr.19: 12891619-12891619 on Build GRCh38
rs760467479 Chr.19: 12891731-12891731 on Build GRCh38
rs112314659 Chr.19: 12891749-12891749 on Build GRCh38
rs3745647 Chr.19: 12891570-12891570 on Build GRCh38
rs766105340 Chr.19: 12891734-12891734 on Build GRCh38
rs924929359 Chr.19: 12891702-12891702 on Build GRCh38
rs768617997 Chr.19: 12891692-12891692 on Build GRCh38
rs764661339 Chr.19: 12891800-12891800 on Build GRCh38
rs371007485 Chr.19: 12891530-12891530 on Build GRCh38
rs1799918 Chr.19: 12891586-12891586 on Build GRCh38
rs113028448 Chr.19: 12891601-12891601 on Build GRCh38
rs547301120 Chr.19: 12891766-12891766 on Build GRCh38
rs936598138 Chr.19: 12891723-12891723 on Build GRCh38
rs768532620 Chr.19: 12891860-12891860 on Build GRCh38

View Full Product Details