Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43936577-43937100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARTN
Entrez Gene ID
9048
Gene Name
artemin
Gene Aliases
ENOVIN, EVN, NBN
UniGene
Hs.632404
Gene Chromosome Location
Chr.1: 43933320-43937240 on Build GRCh38
Transcripts
NM_001136215.1 NM_057090.2 NM_057091.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7106126 c.590G>A p.R197Q Chr.1: 43936668-43936668 on Build GRCh38
1191757 c.652C>T p.R218C Chr.1: 43936730-43936730 on Build GRCh38
1245521 c.556C>A p.P186T Chr.1: 43936634-43936634 on Build GRCh38
1342918 c.592C>T p.P198S Chr.1: 43936670-43936670 on Build GRCh38
4933440 c.671G>T p.C224F Chr.1: 43936749-43936749 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766463007 Chr.1: 43936685-43936685 on Build GRCh38
rs1000683913 Chr.1: 43936922-43936922 on Build GRCh38
rs776841585 Chr.1: 43937003-43937003 on Build GRCh38
rs192619513 Chr.1: 43937038-43937038 on Build GRCh38
rs952859631 Chr.1: 43937011-43937011 on Build GRCh38
rs1025490202 Chr.1: 43936766-43936766 on Build GRCh38
rs998260853 Chr.1: 43936967-43936967 on Build GRCh38
rs941282030 Chr.1: 43936953-43936953 on Build GRCh38
rs752643971 Chr.1: 43936719-43936719 on Build GRCh38
rs969362504 Chr.1: 43936972-43936972 on Build GRCh38
rs1050758051 Chr.1: 43936977-43936977 on Build GRCh38
rs771462996 Chr.1: 43936963-43936963 on Build GRCh38
rs370236511 Chr.1: 43936736-43936736 on Build GRCh38
rs981433986 Chr.1: 43936864-43936864 on Build GRCh38
rs576387859 Chr.1: 43936817-43936817 on Build GRCh38
rs940357012 Chr.1: 43936835-43936835 on Build GRCh38
rs752447756 Chr.1: 43936631-43936631 on Build GRCh38
rs1040072041 Chr.1: 43936851-43936851 on Build GRCh38
rs781736783 Chr.1: 43936804-43936804 on Build GRCh38
rs777421734 Chr.1: 43936633-43936633 on Build GRCh38
rs751487648 Chr.1: 43936625-43936625 on Build GRCh38
rs745323674 Chr.1: 43936771-43936771 on Build GRCh38
rs748894349 Chr.1: 43936639-43936639 on Build GRCh38
rs778342419 Chr.1: 43936658-43936658 on Build GRCh38
rs769248831 Chr.1: 43936663-43936663 on Build GRCh38
rs748673937 Chr.1: 43936810-43936810 on Build GRCh38
rs766437568 Chr.1: 43936622-43936622 on Build GRCh38
rs148086985 Chr.1: 43936919-43936919 on Build GRCh38
rs552771679 Chr.1: 43936850-43936850 on Build GRCh38
rs767392376 Chr.1: 43936707-43936707 on Build GRCh38
rs778741890 Chr.1: 43936767-43936767 on Build GRCh38
rs376893754 Chr.1: 43936667-43936667 on Build GRCh38
rs927373784 Chr.1: 43936875-43936875 on Build GRCh38
rs373484831 Chr.1: 43936662-43936662 on Build GRCh38
rs762214862 Chr.1: 43936672-43936672 on Build GRCh38
rs755866875 Chr.1: 43936723-43936723 on Build GRCh38
rs1027045343 Chr.1: 43936999-43936999 on Build GRCh38
rs892350624 Chr.1: 43936956-43936956 on Build GRCh38
rs1013914015 Chr.1: 43936959-43936959 on Build GRCh38
rs997980023 Chr.1: 43936648-43936648 on Build GRCh38
rs12127737 Chr.1: 43936893-43936893 on Build GRCh38
rs1036942600 Chr.1: 43936955-43936955 on Build GRCh38
rs548593022 Chr.1: 43937033-43937033 on Build GRCh38
rs374554315 Chr.1: 43936798-43936798 on Build GRCh38
rs763023264 Chr.1: 43936615-43936615 on Build GRCh38
rs780932734 Chr.1: 43936629-43936629 on Build GRCh38
rs770059461 Chr.1: 43936674-43936674 on Build GRCh38
rs756907005 Chr.1: 43936761-43936761 on Build GRCh38
rs765454863 Chr.1: 43936609-43936609 on Build GRCh38
rs750628728 Chr.1: 43936613-43936613 on Build GRCh38
rs375263972 Chr.1: 43937059-43937059 on Build GRCh38
rs754955889 Chr.1: 43936627-43936627 on Build GRCh38
rs1054979502 Chr.1: 43936909-43936909 on Build GRCh38
rs770466998 Chr.1: 43936646-43936646 on Build GRCh38
rs751676511 Chr.1: 43936699-43936699 on Build GRCh38
rs1052786911 Chr.1: 43936932-43936932 on Build GRCh38
rs763144695 Chr.1: 43936682-43936682 on Build GRCh38
rs957644671 Chr.1: 43936768-43936768 on Build GRCh38
rs558178801 Chr.1: 43937000-43937000 on Build GRCh38
rs760976812 Chr.1: 43936702-43936702 on Build GRCh38
rs982936673 Chr.1: 43937046-43937046 on Build GRCh38
rs1033109701 Chr.1: 43936724-43936724 on Build GRCh38
rs763858286 Chr.1: 43936730-43936730 on Build GRCh38
rs759642007 Chr.1: 43936703-43936703 on Build GRCh38
rs773480609 Chr.1: 43936675-43936675 on Build GRCh38
rs889545858 Chr.1: 43936979-43936979 on Build GRCh38
rs1033161886 Chr.1: 43937054-43937054 on Build GRCh38

View Full Product Details