Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14120208-14120716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASF1B
Entrez Gene ID
55723
Gene Name
anti-silencing function 1B histone chaperone
Gene Aliases
CIA-II
UniGene
Hs.26516
Gene Chromosome Location
Chr.19: 14119509-14136628 on Build GRCh38
Transcripts
NM_018154.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4454627 c.541A>T p.I181F Chr.19: 14120527-14120527 on Build GRCh38
6588312 c.506C>T p.S169F Chr.19: 14120562-14120562 on Build GRCh38
6564911 c.490G>C p.E164Q Chr.19: 14120578-14120578 on Build GRCh38
266049 c.476G>T p.R159M Chr.19: 14120592-14120592 on Build GRCh38
390131 c.417C>T p.I139I Chr.19: 14120651-14120651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965726314 Chr.19: 14120362-14120362 on Build GRCh38
rs745344857 Chr.19: 14120556-14120556 on Build GRCh38
rs749689410 Chr.19: 14120631-14120632 on Build GRCh38
rs775946328 Chr.19: 14120242-14120242 on Build GRCh38
rs779333083 Chr.19: 14120548-14120548 on Build GRCh38
rs151302500 Chr.19: 14120619-14120619 on Build GRCh38
rs905405216 Chr.19: 14120472-14120472 on Build GRCh38
rs755215195 Chr.19: 14120635-14120635 on Build GRCh38
rs769406622 Chr.19: 14120509-14120509 on Build GRCh38
rs139612745 Chr.19: 14120464-14120464 on Build GRCh38
rs754251890 Chr.19: 14120538-14120538 on Build GRCh38
rs551860719 Chr.19: 14120446-14120446 on Build GRCh38
rs758936633 Chr.19: 14120643-14120643 on Build GRCh38
rs3745463 Chr.19: 14120444-14120444 on Build GRCh38
rs374362551 Chr.19: 14120426-14120426 on Build GRCh38
rs141700566 Chr.19: 14120630-14120630 on Build GRCh38
rs769237306 Chr.19: 14120462-14120462 on Build GRCh38
rs926994495 Chr.19: 14120277-14120277 on Build GRCh38
rs542174932 Chr.19: 14120429-14120429 on Build GRCh38
rs775070439 Chr.19: 14120567-14120567 on Build GRCh38
rs918037927 Chr.19: 14120695-14120695 on Build GRCh38
rs774052434 Chr.19: 14120684-14120684 on Build GRCh38
rs781566713 Chr.19: 14120432-14120432 on Build GRCh38
rs1014711925 Chr.19: 14120597-14120597 on Build GRCh38
rs936068483 Chr.19: 14120595-14120595 on Build GRCh38
rs988175320 Chr.19: 14120694-14120694 on Build GRCh38
rs577237560 Chr.19: 14120351-14120351 on Build GRCh38
rs142438175 Chr.19: 14120551-14120551 on Build GRCh38
rs375618333 Chr.19: 14120443-14120443 on Build GRCh38
rs756444397 Chr.19: 14120531-14120531 on Build GRCh38
rs768399721 Chr.19: 14120594-14120594 on Build GRCh38
rs889092150 Chr.19: 14120663-14120663 on Build GRCh38
rs768327995 Chr.19: 14120678-14120678 on Build GRCh38
rs778206991 Chr.19: 14120657-14120657 on Build GRCh38
rs779614573 Chr.19: 14120669-14120669 on Build GRCh38
rs541229380 Chr.19: 14120373-14120373 on Build GRCh38
rs769115670 Chr.19: 14120317-14120317 on Build GRCh38
rs866884143 Chr.19: 14120403-14120403 on Build GRCh38
rs367584606 Chr.19: 14120546-14120546 on Build GRCh38
rs750730597 Chr.19: 14120481-14120482 on Build GRCh38
rs762814793 Chr.19: 14120568-14120568 on Build GRCh38
rs201366561 Chr.19: 14120685-14120685 on Build GRCh38
rs775299757 Chr.19: 14120511-14120511 on Build GRCh38
rs759365999 Chr.19: 14120626-14120626 on Build GRCh38
rs749631039 Chr.19: 14120254-14120254 on Build GRCh38
rs917055237 Chr.19: 14120328-14120328 on Build GRCh38
rs764994662 Chr.19: 14120633-14120633 on Build GRCh38
rs756369453 Chr.19: 14120485-14120487 on Build GRCh38
rs760840829 Chr.19: 14120693-14120693 on Build GRCh38
rs1057000732 Chr.19: 14120325-14120325 on Build GRCh38
rs750560881 Chr.19: 14120624-14120624 on Build GRCh38
rs780383299 Chr.19: 14120532-14120532 on Build GRCh38
rs200801036 Chr.19: 14120634-14120634 on Build GRCh38
rs565977296 Chr.19: 14120566-14120566 on Build GRCh38
rs745397948 Chr.19: 14120658-14120658 on Build GRCh38
rs371339968 Chr.19: 14120671-14120671 on Build GRCh38
rs149484550 Chr.19: 14120259-14120259 on Build GRCh38
rs750505525 Chr.19: 14120530-14120530 on Build GRCh38
rs776107476 Chr.19: 14120491-14120491 on Build GRCh38
rs559024702 Chr.19: 14120302-14120302 on Build GRCh38
rs780722232 Chr.19: 14120552-14120552 on Build GRCh38
rs760785660 Chr.19: 14120625-14120625 on Build GRCh38
rs548895792 Chr.19: 14120689-14120689 on Build GRCh38
rs774178777 Chr.19: 14120602-14120602 on Build GRCh38
rs759236968 Chr.19: 14120499-14120499 on Build GRCh38
rs748947729 Chr.19: 14120672-14120672 on Build GRCh38
rs761802938 Chr.19: 14120520-14120520 on Build GRCh38
rs761703228 Chr.19: 14120603-14120603 on Build GRCh38
rs549601434 Chr.19: 14120642-14120642 on Build GRCh38
rs144221648 Chr.19: 14120486-14120486 on Build GRCh38
rs780071194 Chr.19: 14120549-14120550 on Build GRCh38
rs3745464 Chr.19: 14120431-14120431 on Build GRCh38
rs945881274 Chr.19: 14120346-14120346 on Build GRCh38
rs969950256 Chr.19: 14120252-14120252 on Build GRCh38
rs907515083 Chr.19: 14120353-14120353 on Build GRCh38
rs746157443 Chr.19: 14120441-14120441 on Build GRCh38
rs992673073 Chr.19: 14120542-14120542 on Build GRCh38
rs764108702 Chr.19: 14120517-14120517 on Build GRCh38
rs370433719 Chr.19: 14120483-14120483 on Build GRCh38
rs756816563 Chr.19: 14120368-14120368 on Build GRCh38
rs755558637 Chr.19: 14120659-14120659 on Build GRCh38
rs757718620 Chr.19: 14120457-14120457 on Build GRCh38
rs767760898 Chr.19: 14120527-14120527 on Build GRCh38
rs1050406028 Chr.19: 14120392-14120392 on Build GRCh38
rs752467925 Chr.19: 14120427-14120427 on Build GRCh38
rs8102267 Chr.19: 14120513-14120513 on Build GRCh38
rs199525441 Chr.19: 14120428-14120428 on Build GRCh38
rs8102698 Chr.19: 14120518-14120518 on Build GRCh38
rs145956683 Chr.19: 14120391-14120391 on Build GRCh38
rs748661613 Chr.19: 14120549-14120549 on Build GRCh38

View Full Product Details