Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15227009-15227518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHX3
Entrez Gene ID
79852
Gene Name
epoxide hydrolase 3
Gene Aliases
ABHD9, EH3
UniGene
Hs.156457
Gene Chromosome Location
Chr.19: 15226919-15233047 on Build GRCh38
Transcripts
NM_001142886.1 NM_024794.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7161461 c.1062C>T p.F354F Chr.19: 15227458-15227458 on Build GRCh38
249063 c.1062C>G p.F354L Chr.19: 15227458-15227458 on Build GRCh38
6655314 c.1050C>A p.Y350* Chr.19: 15227470-15227470 on Build GRCh38
1391067 c.1033delC p.Q345fs*>16 Chr.19: 15227487-15227487 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370419225 Chr.19: 15227351-15227351 on Build GRCh38
rs771566576 Chr.19: 15227395-15227395 on Build GRCh38
rs751003614 Chr.19: 15227490-15227490 on Build GRCh38
rs530065809 Chr.19: 15227408-15227408 on Build GRCh38
rs764166880 Chr.19: 15227464-15227464 on Build GRCh38
rs561320444 Chr.19: 15227384-15227384 on Build GRCh38
rs774304199 Chr.19: 15227466-15227466 on Build GRCh38
rs1054491350 Chr.19: 15227316-15227316 on Build GRCh38
rs199698308 Chr.19: 15227440-15227440 on Build GRCh38
rs770539760 Chr.19: 15227487-15227487 on Build GRCh38
rs980925431 Chr.19: 15227339-15227339 on Build GRCh38
rs971814182 Chr.19: 15227257-15227257 on Build GRCh38
rs907346071 Chr.19: 15227077-15227077 on Build GRCh38
rs962616489 Chr.19: 15227312-15227312 on Build GRCh38
rs760634252 Chr.19: 15227456-15227456 on Build GRCh38
rs992600546 Chr.19: 15227355-15227355 on Build GRCh38
rs745659145 Chr.19: 15227415-15227415 on Build GRCh38
rs1056452664 Chr.19: 15227104-15227104 on Build GRCh38
rs747806786 Chr.19: 15227389-15227389 on Build GRCh38
rs773269339 Chr.19: 15227323-15227323 on Build GRCh38
rs952165050 Chr.19: 15227338-15227338 on Build GRCh38
rs926626209 Chr.19: 15227096-15227096 on Build GRCh38
rs1050106190 Chr.19: 15227261-15227261 on Build GRCh38
rs776609731 Chr.19: 15227411-15227411 on Build GRCh38
rs913867962 Chr.19: 15227412-15227412 on Build GRCh38
rs547415258 Chr.19: 15227350-15227350 on Build GRCh38
rs888851519 Chr.19: 15227280-15227280 on Build GRCh38
rs1050198027 Chr.19: 15227489-15227489 on Build GRCh38
rs144741388 Chr.19: 15227218-15227218 on Build GRCh38
rs143196077 Chr.19: 15227470-15227470 on Build GRCh38
rs1009002022 Chr.19: 15227287-15227287 on Build GRCh38
rs767003446 Chr.19: 15227249-15227249 on Build GRCh38
rs775729973 Chr.19: 15227442-15227442 on Build GRCh38
rs544496378 Chr.19: 15227341-15227341 on Build GRCh38
rs191607342 Chr.19: 15227322-15227322 on Build GRCh38
rs945954828 Chr.19: 15227418-15227418 on Build GRCh38
rs144386323 Chr.19: 15227462-15227462 on Build GRCh38
rs533509644 Chr.19: 15227045-15227045 on Build GRCh38
rs766951093 Chr.19: 15227041-15227041 on Build GRCh38
rs1015869626 Chr.19: 15227227-15227227 on Build GRCh38
rs562600102 Chr.19: 15227215-15227215 on Build GRCh38
rs751990351 Chr.19: 15227114-15227114 on Build GRCh38
rs746898567 Chr.19: 15227407-15227407 on Build GRCh38
rs936566911 Chr.19: 15227314-15227314 on Build GRCh38
rs761359276 Chr.19: 15227102-15227102 on Build GRCh38
rs149370041 Chr.19: 15227160-15227160 on Build GRCh38
rs573598061 Chr.19: 15227161-15227161 on Build GRCh38
rs986046855 Chr.19: 15227308-15227308 on Build GRCh38
rs777182998 Chr.19: 15227403-15227403 on Build GRCh38
rs773928122 Chr.19: 15227046-15227046 on Build GRCh38
rs545241041 Chr.19: 15227286-15227286 on Build GRCh38
rs945975867 Chr.19: 15227115-15227115 on Build GRCh38
rs1041947546 Chr.19: 15227154-15227154 on Build GRCh38
rs999920377 Chr.19: 15227171-15227171 on Build GRCh38
rs866461767 Chr.19: 15227101-15227101 on Build GRCh38
rs917194811 Chr.19: 15227107-15227107 on Build GRCh38
rs756528252 Chr.19: 15227495-15227495 on Build GRCh38
rs923107889 Chr.19: 15227270-15227270 on Build GRCh38
rs768131182 Chr.19: 15227476-15227476 on Build GRCh38
rs980829045 Chr.19: 15227065-15227065 on Build GRCh38
rs542695119 Chr.19: 15227178-15227178 on Build GRCh38
rs952111342 Chr.19: 15227049-15227049 on Build GRCh38
rs565128667 Chr.19: 15227300-15227300 on Build GRCh38

View Full Product Details