Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18924141-18924643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX49
Entrez Gene ID
54555
Gene Name
DEAD-box helicase 49
Gene Aliases
R27090_2
UniGene
Hs.143187
Gene Chromosome Location
Chr.19: 18919675-18928633 on Build GRCh38
Transcripts
NM_019070.4 NR_033677.1 XM_011528083.1 XM_011528084.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3100634 c.798G>A p.M266I Chr.19: 18924254-18924254 on Build GRCh38
4653084 c.538C>T p.R180C Chr.19: 18924629-18924629 on Build GRCh38
4653082 c.475A>G p.M159V Chr.19: 18924252-18924252 on Build GRCh38
4653083 c.859C>T p.R287C Chr.19: 18924629-18924629 on Build GRCh38
1686786 c.853-1G>A p.? Chr.19: 18924622-18924622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368523415 Chr.19: 18924281-18924281 on Build GRCh38
rs747700700 Chr.19: 18924280-18924280 on Build GRCh38
rs115276776 Chr.19: 18924522-18924522 on Build GRCh38
rs537184854 Chr.19: 18924593-18924593 on Build GRCh38
rs188008835 Chr.19: 18924278-18924278 on Build GRCh38
rs367687762 Chr.19: 18924244-18924244 on Build GRCh38
rs760907892 Chr.19: 18924218-18924218 on Build GRCh38
rs952378098 Chr.19: 18924550-18924550 on Build GRCh38
rs780497270 Chr.19: 18924325-18924326 on Build GRCh38
rs760505884 Chr.19: 18924603-18924603 on Build GRCh38
rs750238904 Chr.19: 18924190-18924190 on Build GRCh38
rs574523833 Chr.19: 18924163-18924163 on Build GRCh38
rs751625714 Chr.19: 18924282-18924282 on Build GRCh38
rs372898180 Chr.19: 18924266-18924266 on Build GRCh38
rs755567801 Chr.19: 18924349-18924349 on Build GRCh38
rs369900868 Chr.19: 18924355-18924355 on Build GRCh38
rs1021226494 Chr.19: 18924179-18924179 on Build GRCh38
rs898652291 Chr.19: 18924471-18924471 on Build GRCh38
rs757255123 Chr.19: 18924185-18924185 on Build GRCh38
rs199563493 Chr.19: 18924357-18924357 on Build GRCh38
rs773680274 Chr.19: 18924211-18924211 on Build GRCh38
rs1028553408 Chr.19: 18924463-18924463 on Build GRCh38
rs761720601 Chr.19: 18924250-18924250 on Build GRCh38
rs926416301 Chr.19: 18924344-18924344 on Build GRCh38
rs767475173 Chr.19: 18924450-18924450 on Build GRCh38
rs386807509 Chr.19: 18924545-18924546 on Build GRCh38
rs147481044 Chr.19: 18924307-18924307 on Build GRCh38
rs772754998 Chr.19: 18924209-18924209 on Build GRCh38
rs985006155 Chr.19: 18924558-18924558 on Build GRCh38
rs140226594 Chr.19: 18924351-18924351 on Build GRCh38
rs143159618 Chr.19: 18924271-18924271 on Build GRCh38
rs903759587 Chr.19: 18924436-18924436 on Build GRCh38
rs192511312 Chr.19: 18924477-18924477 on Build GRCh38
rs371802790 Chr.19: 18924600-18924600 on Build GRCh38
rs772952517 Chr.19: 18924333-18924333 on Build GRCh38
rs2301660 Chr.19: 18924546-18924546 on Build GRCh38
rs111977431 Chr.19: 18924329-18924329 on Build GRCh38
rs556766043 Chr.19: 18924325-18924325 on Build GRCh38
rs954735698 Chr.19: 18924348-18924348 on Build GRCh38
rs748651643 Chr.19: 18924208-18924208 on Build GRCh38
rs753714388 Chr.19: 18924341-18924341 on Build GRCh38
rs776463656 Chr.19: 18924616-18924616 on Build GRCh38
rs775520659 Chr.19: 18924328-18924328 on Build GRCh38
rs894242914 Chr.19: 18924414-18924414 on Build GRCh38
rs200641433 Chr.19: 18924356-18924356 on Build GRCh38
rs573005981 Chr.19: 18924394-18924394 on Build GRCh38
rs953468638 Chr.19: 18924472-18924472 on Build GRCh38
rs752835831 Chr.19: 18924230-18924230 on Build GRCh38
rs5827425 Chr.19: 18924351-18924351 on Build GRCh38
rs775628811 Chr.19: 18924585-18924585 on Build GRCh38
rs536731978 Chr.19: 18924279-18924279 on Build GRCh38
rs750392042 Chr.19: 18924260-18924260 on Build GRCh38
rs150711492 Chr.19: 18924348-18924348 on Build GRCh38
rs752331613 Chr.19: 18924620-18924620 on Build GRCh38
rs545694455 Chr.19: 18924543-18924543 on Build GRCh38
rs902839988 Chr.19: 18924162-18924162 on Build GRCh38
rs138743473 Chr.19: 18924492-18924492 on Build GRCh38
rs986776632 Chr.19: 18924527-18924527 on Build GRCh38
rs781392464 Chr.19: 18924358-18924358 on Build GRCh38
rs760728909 Chr.19: 18924338-18924338 on Build GRCh38
rs879591068 Chr.19: 18924545-18924546 on Build GRCh38
rs1000694983 Chr.19: 18924204-18924204 on Build GRCh38
rs764225625 Chr.19: 18924165-18924165 on Build GRCh38
rs1476505 Chr.19: 18924561-18924561 on Build GRCh38
rs755765241 Chr.19: 18924201-18924201 on Build GRCh38
rs780955116 Chr.19: 18924186-18924186 on Build GRCh38
rs767486527 Chr.19: 18924251-18924251 on Build GRCh38
rs771135039 Chr.19: 18924295-18924295 on Build GRCh38
rs999453 Chr.19: 18924608-18924608 on Build GRCh38
rs771350028 Chr.19: 18924221-18924221 on Build GRCh38
rs758193551 Chr.19: 18924622-18924622 on Build GRCh38
rs768610675 Chr.19: 18924590-18924590 on Build GRCh38
rs61750860 Chr.19: 18924619-18924619 on Build GRCh38
rs2301659 Chr.19: 18924545-18924545 on Build GRCh38
rs759811880 Chr.19: 18924227-18924227 on Build GRCh38
rs777122683 Chr.19: 18924300-18924300 on Build GRCh38
rs770179786 Chr.19: 18924359-18924359 on Build GRCh38
rs764943039 Chr.19: 18924349-18924349 on Build GRCh38
rs1042275201 Chr.19: 18924432-18924432 on Build GRCh38
rs768212802 Chr.19: 18924206-18924206 on Build GRCh38
rs747565108 Chr.19: 18924285-18924285 on Build GRCh38
rs201684040 Chr.19: 18924587-18924587 on Build GRCh38
rs200277294 Chr.19: 18924544-18924545 on Build GRCh38
rs754645395 Chr.19: 18924346-18924346 on Build GRCh38
rs778631607 Chr.19: 18924207-18924207 on Build GRCh38
rs776661274 Chr.19: 18924225-18924225 on Build GRCh38
rs968580789 Chr.19: 18924557-18924557 on Build GRCh38
rs748586735 Chr.19: 18924353-18924353 on Build GRCh38
rs1000744008 Chr.19: 18924452-18924452 on Build GRCh38
rs183729370 Chr.19: 18924262-18924262 on Build GRCh38
rs375027443 Chr.19: 18924202-18924202 on Build GRCh38
rs112705706 Chr.19: 18924232-18924232 on Build GRCh38
rs201654770 Chr.19: 18924546-18924547 on Build GRCh38
rs1012645240 Chr.19: 18924421-18924423 on Build GRCh38
rs200072144 Chr.19: 18924261-18924261 on Build GRCh38
rs759666151 Chr.19: 18924317-18924317 on Build GRCh38
rs141969072 Chr.19: 18924391-18924391 on Build GRCh38
rs374715590 Chr.19: 18924205-18924205 on Build GRCh38
rs938333030 Chr.19: 18924390-18924390 on Build GRCh38
rs765299223 Chr.19: 18924228-18924228 on Build GRCh38
rs986512998 Chr.19: 18924352-18924352 on Build GRCh38

View Full Product Details