Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 32453059-32453552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L3
Entrez Gene ID
147991
Gene Name
dpy-19 like 3 (C. elegans)
Gene Aliases
-
UniGene
Hs.194392
Gene Chromosome Location
Chr.19: 32405749-32485893 on Build GRCh38
Transcripts
NM_001172774.1 NM_207325.2 XM_006723045.1 XM_011526526.2 XM_017026366.1 XM_017026367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3422649 c.857C>T p.A286V Chr.19: 32453146-32453146 on Build GRCh38
5056505 c.865C>A p.L289M Chr.19: 32453154-32453154 on Build GRCh38
5013415 c.962C>A p.S321* Chr.19: 32453251-32453251 on Build GRCh38
1250675 c.935G>C p.G312A Chr.19: 32453224-32453224 on Build GRCh38
3959896 c.924C>T p.S308S Chr.19: 32453213-32453213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043054946 Chr.19: 32453300-32453300 on Build GRCh38
rs527368198 Chr.19: 32453235-32453235 on Build GRCh38
rs746655150 Chr.19: 32453116-32453117 on Build GRCh38
rs777765410 Chr.19: 32453091-32453091 on Build GRCh38
rs149282626 Chr.19: 32453146-32453146 on Build GRCh38
rs929190046 Chr.19: 32453372-32453372 on Build GRCh38
rs756532547 Chr.19: 32453125-32453125 on Build GRCh38
rs984865532 Chr.19: 32453123-32453123 on Build GRCh38
rs776558527 Chr.19: 32453189-32453189 on Build GRCh38
rs973090290 Chr.19: 32453094-32453094 on Build GRCh38
rs757914938 Chr.19: 32453291-32453291 on Build GRCh38
rs746602544 Chr.19: 32453298-32453298 on Build GRCh38
rs1025458627 Chr.19: 32453102-32453102 on Build GRCh38
rs769797953 Chr.19: 32453284-32453284 on Build GRCh38
rs780771732 Chr.19: 32453135-32453135 on Build GRCh38
rs994078063 Chr.19: 32453481-32453481 on Build GRCh38
rs1024637327 Chr.19: 32453086-32453086 on Build GRCh38
rs988431953 Chr.19: 32453183-32453183 on Build GRCh38
rs893999952 Chr.19: 32453482-32453482 on Build GRCh38
rs755395895 Chr.19: 32453274-32453274 on Build GRCh38
rs777710442 Chr.19: 32453087-32453087 on Build GRCh38
rs780750558 Chr.19: 32453220-32453221 on Build GRCh38
rs754418389 Chr.19: 32453259-32453259 on Build GRCh38
rs771660180 Chr.19: 32453168-32453168 on Build GRCh38
rs936847473 Chr.19: 32453331-32453331 on Build GRCh38
rs777472089 Chr.19: 32453295-32453295 on Build GRCh38
rs906232030 Chr.19: 32453448-32453448 on Build GRCh38
rs1021243080 Chr.19: 32453526-32453526 on Build GRCh38
rs541427809 Chr.19: 32453088-32453088 on Build GRCh38
rs1044164771 Chr.19: 32453415-32453415 on Build GRCh38
rs759337098 Chr.19: 32453196-32453196 on Build GRCh38
rs767902213 Chr.19: 32453232-32453232 on Build GRCh38
rs781644799 Chr.19: 32453107-32453107 on Build GRCh38
rs555694876 Chr.19: 32453285-32453285 on Build GRCh38
rs1046034159 Chr.19: 32453434-32453434 on Build GRCh38
rs367971073 Chr.19: 32453282-32453282 on Build GRCh38
rs776782588 Chr.19: 32453202-32453202 on Build GRCh38
rs1047663202 Chr.19: 32453386-32453386 on Build GRCh38
rs1025997538 Chr.19: 32453522-32453522 on Build GRCh38
rs775589839 Chr.19: 32453296-32453297 on Build GRCh38
rs761923638 Chr.19: 32453212-32453212 on Build GRCh38
rs779442466 Chr.19: 32453280-32453280 on Build GRCh38
rs999952150 Chr.19: 32453450-32453450 on Build GRCh38
rs374185599 Chr.19: 32453182-32453182 on Build GRCh38
rs564666545 Chr.19: 32453197-32453197 on Build GRCh38
rs757372282 Chr.19: 32453106-32453106 on Build GRCh38
rs964666635 Chr.19: 32453187-32453187 on Build GRCh38
rs536858783 Chr.19: 32453312-32453312 on Build GRCh38
rs79614333 Chr.19: 32453292-32453292 on Build GRCh38
rs368428740 Chr.19: 32453326-32453326 on Build GRCh38
rs188020683 Chr.19: 32453447-32453447 on Build GRCh38
rs34259672 Chr.19: 32453228-32453228 on Build GRCh38
rs745375585 Chr.19: 32453283-32453284 on Build GRCh38
rs774255214 Chr.19: 32453209-32453209 on Build GRCh38
rs75752714 Chr.19: 32453293-32453293 on Build GRCh38
rs746868275 Chr.19: 32453174-32453174 on Build GRCh38
rs894896287 Chr.19: 32453349-32453349 on Build GRCh38
rs749232322 Chr.19: 32453399-32453399 on Build GRCh38
rs1011874734 Chr.19: 32453084-32453084 on Build GRCh38
rs913875734 Chr.19: 32453192-32453192 on Build GRCh38
rs541650176 Chr.19: 32453151-32453151 on Build GRCh38
rs371136278 Chr.19: 32453147-32453147 on Build GRCh38
rs1011986025 Chr.19: 32453400-32453400 on Build GRCh38
rs905570099 Chr.19: 32453426-32453426 on Build GRCh38
rs772825844 Chr.19: 32453169-32453169 on Build GRCh38
rs756919363 Chr.19: 32453207-32453208 on Build GRCh38
rs750859418 Chr.19: 32453110-32453110 on Build GRCh38
rs1053433275 Chr.19: 32453335-32453335 on Build GRCh38
rs923298047 Chr.19: 32453193-32453193 on Build GRCh38
rs770711359 Chr.19: 32453490-32453490 on Build GRCh38
rs766792616 Chr.19: 32453238-32453238 on Build GRCh38
rs1013962423 Chr.19: 32453403-32453403 on Build GRCh38
rs747657838 Chr.19: 32453281-32453281 on Build GRCh38
rs749745501 Chr.19: 32453141-32453141 on Build GRCh38
rs76155137 Chr.19: 32453229-32453229 on Build GRCh38
rs770729672 Chr.19: 32453302-32453302 on Build GRCh38
rs764017568 Chr.19: 32453098-32453098 on Build GRCh38
rs78879458 Chr.19: 32453319-32453319 on Build GRCh38

View Full Product Details