Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 32900797-32901330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP89
Entrez Gene ID
84902
Gene Name
centrosomal protein 89
Gene Aliases
CCDC123, CEP123
UniGene
Hs.599703
Gene Chromosome Location
Chr.19: 32875925-32972029 on Build GRCh38
Transcripts
NM_032816.4 XM_005259344.3 XM_011527425.2 XM_017027398.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3970899 c.1685A>G p.Q562R Chr.19: 32901293-32901293 on Build GRCh38
1235980 c.1661T>C p.L554S Chr.19: 32901317-32901317 on Build GRCh38
994626 c.1733+2T>C p.? Chr.19: 32901243-32901243 on Build GRCh38
2751189 c.1705G>A p.E569K Chr.19: 32901273-32901273 on Build GRCh38
1199809 c.1733+2T>G p.? Chr.19: 32901243-32901243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs8111033 Chr.19: 32900832-32900832 on Build GRCh38
rs899780698 Chr.19: 32901075-32901075 on Build GRCh38
rs998224725 Chr.19: 32901076-32901076 on Build GRCh38
rs187153462 Chr.19: 32901239-32901239 on Build GRCh38
rs570043573 Chr.19: 32901217-32901217 on Build GRCh38
rs7253415 Chr.19: 32901181-32901181 on Build GRCh38
rs963440645 Chr.19: 32900860-32900860 on Build GRCh38
rs779353102 Chr.19: 32901202-32901202 on Build GRCh38
rs775079961 Chr.19: 32901273-32901273 on Build GRCh38
rs7252134 Chr.19: 32900947-32900947 on Build GRCh38
rs571238520 Chr.19: 32901179-32901179 on Build GRCh38
rs755679621 Chr.19: 32901032-32901032 on Build GRCh38
rs750473202 Chr.19: 32901206-32901206 on Build GRCh38
rs902907561 Chr.19: 32901105-32901105 on Build GRCh38
rs1032791218 Chr.19: 32901160-32901160 on Build GRCh38
rs779345339 Chr.19: 32901289-32901289 on Build GRCh38
rs189911102 Chr.19: 32901043-32901043 on Build GRCh38
rs989717533 Chr.19: 32901017-32901017 on Build GRCh38
rs150896465 Chr.19: 32901276-32901276 on Build GRCh38
rs8107020 Chr.19: 32900954-32900954 on Build GRCh38
rs541718983 Chr.19: 32900963-32900963 on Build GRCh38
rs960395630 Chr.19: 32901185-32901185 on Build GRCh38
rs377584191 Chr.19: 32901278-32901278 on Build GRCh38
rs772397469 Chr.19: 32901240-32901240 on Build GRCh38
rs922416719 Chr.19: 32901304-32901304 on Build GRCh38
rs958175240 Chr.19: 32901166-32901166 on Build GRCh38
rs551244250 Chr.19: 32901129-32901129 on Build GRCh38
rs925788806 Chr.19: 32901270-32901270 on Build GRCh38
rs960041079 Chr.19: 32900955-32900955 on Build GRCh38
rs888327004 Chr.19: 32900869-32900869 on Build GRCh38
rs1007723648 Chr.19: 32900846-32900846 on Build GRCh38
rs747706449 Chr.19: 32901019-32901019 on Build GRCh38
rs368697866 Chr.19: 32900888-32900888 on Build GRCh38
rs182195430 Chr.19: 32901197-32901197 on Build GRCh38
rs770439772 Chr.19: 32901286-32901286 on Build GRCh38
rs34188586 Chr.19: 32900897-32900898 on Build GRCh38
rs377466027 Chr.19: 32900886-32900886 on Build GRCh38
rs899476057 Chr.19: 32900949-32900949 on Build GRCh38
rs756464830 Chr.19: 32901211-32901211 on Build GRCh38
rs528283729 Chr.19: 32901128-32901128 on Build GRCh38
rs757470989 Chr.19: 32900886-32900887 on Build GRCh38
rs967159981 Chr.19: 32900971-32900971 on Build GRCh38
rs763925684 Chr.19: 32901275-32901275 on Build GRCh38
rs748558193 Chr.19: 32901207-32901208 on Build GRCh38
rs765367427 Chr.19: 32901294-32901294 on Build GRCh38
rs563386367 Chr.19: 32901056-32901056 on Build GRCh38
rs751414230 Chr.19: 32900847-32900847 on Build GRCh38
rs759139017 Chr.19: 32901263-32901263 on Build GRCh38
rs771763599 Chr.19: 32901005-32901005 on Build GRCh38
rs777078940 Chr.19: 32901033-32901033 on Build GRCh38
rs765110361 Chr.19: 32901264-32901264 on Build GRCh38
rs56712030 Chr.19: 32900903-32900903 on Build GRCh38
rs776241494 Chr.19: 32901261-32901261 on Build GRCh38
rs778219369 Chr.19: 32901306-32901306 on Build GRCh38
rs528939882 Chr.19: 32901061-32901061 on Build GRCh38
rs1046890000 Chr.19: 32900855-32900855 on Build GRCh38
rs969676098 Chr.19: 32901186-32901186 on Build GRCh38
rs746514877 Chr.19: 32901248-32901248 on Build GRCh38
rs35171769 Chr.19: 32901050-32901050 on Build GRCh38
rs867971683 Chr.19: 32900904-32900904 on Build GRCh38
rs7253310 Chr.19: 32901104-32901104 on Build GRCh38
rs541410529 Chr.19: 32900889-32900889 on Build GRCh38
rs770608768 Chr.19: 32901258-32901258 on Build GRCh38
rs374230640 Chr.19: 32901238-32901238 on Build GRCh38
rs369591029 Chr.19: 32901204-32901204 on Build GRCh38
rs749712696 Chr.19: 32901222-32901222 on Build GRCh38
rs781336634 Chr.19: 32901205-32901205 on Build GRCh38
rs748444728 Chr.19: 32901293-32901293 on Build GRCh38
rs759008543 Chr.19: 32901305-32901305 on Build GRCh38
rs772833328 Chr.19: 32901245-32901245 on Build GRCh38
rs532918018 Chr.19: 32901044-32901044 on Build GRCh38
rs889401714 Chr.19: 32901124-32901124 on Build GRCh38
rs1005446599 Chr.19: 32900883-32900883 on Build GRCh38
rs548669220 Chr.19: 32901074-32901074 on Build GRCh38
rs779102717 Chr.19: 32901229-32901229 on Build GRCh38
rs953613751 Chr.19: 32900839-32900839 on Build GRCh38
rs747155129 Chr.19: 32900888-32900889 on Build GRCh38
rs761903654 Chr.19: 32901274-32901274 on Build GRCh38
rs534441287 Chr.19: 32901067-32901067 on Build GRCh38
rs755119951 Chr.19: 32901280-32901280 on Build GRCh38
rs899316304 Chr.19: 32901174-32901174 on Build GRCh38
rs756233920 Chr.19: 32901277-32901277 on Build GRCh38
rs969792924 Chr.19: 32901029-32901029 on Build GRCh38
rs542402572 Chr.19: 32900865-32900865 on Build GRCh38
rs958246911 Chr.19: 32901011-32901011 on Build GRCh38
rs761525792 Chr.19: 32900889-32900890 on Build GRCh38
rs780871281 Chr.19: 32900981-32900981 on Build GRCh38
rs1031192188 Chr.19: 32901010-32901010 on Build GRCh38

View Full Product Details