Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44512635-44513180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM20
Entrez Gene ID
125931
Gene Name
carcinoembryonic antigen related cell adhesion molecule 20
Gene Aliases
UNQ9366
UniGene
Hs.689632
Gene Chromosome Location
Chr.19: 44503838-44530100 on Build GRCh38
Transcripts
NM_001102597.2 NM_001102598.2 NM_001102599.2 NM_001102600.2 XM_011526429.2 XM_011526430.2 XM_011526431.2 XM_011526432.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6525091 c.1510G>T p.E504* Chr.19: 44512868-44512868 on Build GRCh38
3404362 c.1482G>A p.K494K Chr.19: 44512896-44512896 on Build GRCh38
4167864 c.1426C>T p.R476W Chr.19: 44513173-44513173 on Build GRCh38
5856093 c.1482G>T p.K494N Chr.19: 44512896-44512896 on Build GRCh38
6151525 c.1488G>A p.E496E Chr.19: 44512890-44512890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766055132 Chr.19: 44512998-44512998 on Build GRCh38
rs373001010 Chr.19: 44512775-44512775 on Build GRCh38
rs556065734 Chr.19: 44512805-44512805 on Build GRCh38
rs376778236 Chr.19: 44513003-44513003 on Build GRCh38
rs776278748 Chr.19: 44512861-44512861 on Build GRCh38
rs375276498 Chr.19: 44512908-44512908 on Build GRCh38
rs751847021 Chr.19: 44512856-44512857 on Build GRCh38
rs756445358 Chr.19: 44513130-44513130 on Build GRCh38
rs762918024 Chr.19: 44512968-44512968 on Build GRCh38
rs914805774 Chr.19: 44513121-44513121 on Build GRCh38
rs990700671 Chr.19: 44513114-44513114 on Build GRCh38
rs73938680 Chr.19: 44512914-44512914 on Build GRCh38
rs184877244 Chr.19: 44512869-44512869 on Build GRCh38
rs199955865 Chr.19: 44512868-44512868 on Build GRCh38
rs73938679 Chr.19: 44512855-44512855 on Build GRCh38
rs973911127 Chr.19: 44513105-44513105 on Build GRCh38
rs779016364 Chr.19: 44513151-44513151 on Build GRCh38
rs967237188 Chr.19: 44512748-44512748 on Build GRCh38
rs368515417 Chr.19: 44512890-44512890 on Build GRCh38
rs1027950150 Chr.19: 44513004-44513004 on Build GRCh38
rs544772689 Chr.19: 44512824-44512824 on Build GRCh38
rs754675884 Chr.19: 44512891-44512891 on Build GRCh38
rs779508477 Chr.19: 44513130-44513131 on Build GRCh38
rs772262656 Chr.19: 44512950-44512950 on Build GRCh38
rs768214430 Chr.19: 44513135-44513136 on Build GRCh38
rs760438014 Chr.19: 44513092-44513092 on Build GRCh38
rs1015337788 Chr.19: 44513054-44513054 on Build GRCh38
rs747930065 Chr.19: 44512961-44512961 on Build GRCh38
rs900722426 Chr.19: 44512981-44512981 on Build GRCh38
rs35216557 Chr.19: 44512936-44512936 on Build GRCh38
rs759684861 Chr.19: 44512844-44512844 on Build GRCh38
rs372107713 Chr.19: 44513145-44513145 on Build GRCh38
rs563648499 Chr.19: 44513100-44513101 on Build GRCh38
rs777029307 Chr.19: 44512737-44512737 on Build GRCh38
rs778391010 Chr.19: 44513145-44513146 on Build GRCh38
rs965362385 Chr.19: 44512664-44512664 on Build GRCh38
rs961442757 Chr.19: 44513099-44513099 on Build GRCh38
rs955071640 Chr.19: 44512688-44512688 on Build GRCh38
rs949382516 Chr.19: 44513137-44513137 on Build GRCh38
rs762574240 Chr.19: 44512877-44512877 on Build GRCh38
rs370490069 Chr.19: 44513139-44513139 on Build GRCh38
rs777948369 Chr.19: 44512833-44512833 on Build GRCh38
rs908179812 Chr.19: 44513093-44513093 on Build GRCh38
rs773038104 Chr.19: 44512967-44512967 on Build GRCh38
rs986548446 Chr.19: 44512752-44512752 on Build GRCh38
rs959285694 Chr.19: 44513109-44513109 on Build GRCh38
rs754406725 Chr.19: 44513141-44513141 on Build GRCh38
rs189634421 Chr.19: 44513106-44513106 on Build GRCh38
rs1011685224 Chr.19: 44512665-44512665 on Build GRCh38
rs17800819 Chr.19: 44513005-44513005 on Build GRCh38
rs991784184 Chr.19: 44512829-44512829 on Build GRCh38
rs780166258 Chr.19: 44513156-44513156 on Build GRCh38
rs767105799 Chr.19: 44513124-44513124 on Build GRCh38
rs982854182 Chr.19: 44513090-44513090 on Build GRCh38
rs1369101 Chr.19: 44512780-44512780 on Build GRCh38
rs749357672 Chr.19: 44512836-44512836 on Build GRCh38
rs996500093 Chr.19: 44512672-44512672 on Build GRCh38
rs765931173 Chr.19: 44512887-44512887 on Build GRCh38
rs771722563 Chr.19: 44512996-44512996 on Build GRCh38
rs752345012 Chr.19: 44513125-44513125 on Build GRCh38
rs768504526 Chr.19: 44512854-44512854 on Build GRCh38
rs752971964 Chr.19: 44512899-44512899 on Build GRCh38
rs952448442 Chr.19: 44513044-44513044 on Build GRCh38
rs553353755 Chr.19: 44513101-44513101 on Build GRCh38
rs770944375 Chr.19: 44512837-44512837 on Build GRCh38
rs748812159 Chr.19: 44513131-44513132 on Build GRCh38
rs373968773 Chr.19: 44512820-44512820 on Build GRCh38
rs375433735 Chr.19: 44513153-44513153 on Build GRCh38
rs769582901 Chr.19: 44512964-44512964 on Build GRCh38
rs774565722 Chr.19: 44512842-44512842 on Build GRCh38
rs778957179 Chr.19: 44512917-44512917 on Build GRCh38
rs750338712 Chr.19: 44512873-44512873 on Build GRCh38
rs398034748 Chr.19: 44512933-44512933 on Build GRCh38
rs1028003881 Chr.19: 44512687-44512687 on Build GRCh38
rs1007930785 Chr.19: 44513015-44513015 on Build GRCh38
rs1050457318 Chr.19: 44512663-44512663 on Build GRCh38
rs917875219 Chr.19: 44513117-44513117 on Build GRCh38
rs923460282 Chr.19: 44513110-44513110 on Build GRCh38
rs371778076 Chr.19: 44512960-44512960 on Build GRCh38
rs533013677 Chr.19: 44513149-44513149 on Build GRCh38
rs923407959 Chr.19: 44512898-44512898 on Build GRCh38
rs770177998 Chr.19: 44512830-44512830 on Build GRCh38
rs73560086 Chr.19: 44512892-44512892 on Build GRCh38
rs192193273 Chr.19: 44512806-44512806 on Build GRCh38
rs908330751 Chr.19: 44512764-44512764 on Build GRCh38
rs773974429 Chr.19: 44512999-44512999 on Build GRCh38
rs778160697 Chr.19: 44512900-44512900 on Build GRCh38

View Full Product Details