Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45407202-45407716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD3EAP
Entrez Gene ID
10849
Gene Name
CD3e molecule associated protein
Gene Aliases
ASE-1, ASE1, CAST, PAF49
UniGene
Hs.710495
Gene Chromosome Location
Chr.19: 45406129-45410766 on Build GRCh38
Transcripts
NM_001297590.1 NM_012099.1 XM_017026180.1 XM_017026181.1 NM_001166049.1 NM_001983.3 XM_017026460.1 XM_017026461.1 XM_017026462.1 XM_017026463.1 XM_017026464.1 XM_017026465.1 XM_017026466.1

Gene Symbol
ERCC1
Entrez Gene ID
2067
Gene Name
ERCC excision repair 1, endonuclease non-catalytic subunit
Gene Aliases
COFS4, RAD10, UV20
UniGene
Hs.435981
Gene Chromosome Location
Chr.19: 45407333-45478866 on Build GRCh38
Transcripts
NM_001297590.1 NM_012099.1 XM_017026180.1 XM_017026181.1 NM_001166049.1 NM_001983.3 XM_017026460.1 XM_017026461.1 XM_017026462.1 XM_017026463.1 XM_017026464.1 XM_017026465.1 XM_017026466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5550045 c.146C>T p.A49V Chr.19: 45407217-45407217 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779704319 Chr.19: 45407258-45407258 on Build GRCh38
rs924484490 Chr.19: 45407677-45407677 on Build GRCh38
rs188831361 Chr.19: 45407533-45407533 on Build GRCh38
rs56839686 Chr.19: 45407300-45407301 on Build GRCh38
rs138557840 Chr.19: 45407594-45407594 on Build GRCh38
rs985826900 Chr.19: 45407628-45407628 on Build GRCh38
rs1054921615 Chr.19: 45407394-45407394 on Build GRCh38
rs1049295952 Chr.19: 45407490-45407490 on Build GRCh38
rs552921074 Chr.19: 45407277-45407277 on Build GRCh38
rs920297224 Chr.19: 45407619-45407619 on Build GRCh38
rs910508460 Chr.19: 45407329-45407329 on Build GRCh38
rs761507705 Chr.19: 45407340-45407340 on Build GRCh38
rs1022920429 Chr.19: 45407460-45407460 on Build GRCh38
rs891808539 Chr.19: 45407608-45407608 on Build GRCh38
rs910205288 Chr.19: 45407635-45407635 on Build GRCh38
rs1031751155 Chr.19: 45407328-45407328 on Build GRCh38
rs761629114 Chr.19: 45407232-45407232 on Build GRCh38
rs533637524 Chr.19: 45407496-45407496 on Build GRCh38
rs572885712 Chr.19: 45407295-45407295 on Build GRCh38
rs1046282 Chr.19: 45407414-45407414 on Build GRCh38
rs758040811 Chr.19: 45407254-45407254 on Build GRCh38
rs376694674 Chr.19: 45407437-45407446 on Build GRCh38
rs544748650 Chr.19: 45407439-45407439 on Build GRCh38
rs1050334988 Chr.19: 45407319-45407319 on Build GRCh38
rs990235116 Chr.19: 45407356-45407356 on Build GRCh38
rs919005100 Chr.19: 45407448-45407448 on Build GRCh38
rs771863554 Chr.19: 45407377-45407377 on Build GRCh38
rs1002901802 Chr.19: 45407309-45407309 on Build GRCh38
rs951770832 Chr.19: 45407626-45407626 on Build GRCh38
rs746791432 Chr.19: 45407289-45407289 on Build GRCh38
rs748674099 Chr.19: 45407287-45407287 on Build GRCh38
rs929019825 Chr.19: 45407480-45407480 on Build GRCh38
rs1040234266 Chr.19: 45407367-45407367 on Build GRCh38
rs910483488 Chr.19: 45407399-45407399 on Build GRCh38
rs149272244 Chr.19: 45407639-45407639 on Build GRCh38
rs781751749 Chr.19: 45407286-45407286 on Build GRCh38
rs930261391 Chr.19: 45407288-45407288 on Build GRCh38
rs117128015 Chr.19: 45407645-45407645 on Build GRCh38
rs753794315 Chr.19: 45407228-45407228 on Build GRCh38
rs1031636605 Chr.19: 45407601-45407601 on Build GRCh38
rs750030260 Chr.19: 45407253-45407253 on Build GRCh38
rs548752904 Chr.19: 45407543-45407543 on Build GRCh38
rs971656717 Chr.19: 45407505-45407505 on Build GRCh38
rs915044390 Chr.19: 45407371-45407371 on Build GRCh38
rs760296724 Chr.19: 45407459-45407459 on Build GRCh38
rs568889783 Chr.19: 45407592-45407592 on Build GRCh38
rs111448570 Chr.19: 45407534-45407534 on Build GRCh38
rs73559395 Chr.19: 45407351-45407351 on Build GRCh38
rs887924730 Chr.19: 45407495-45407495 on Build GRCh38
rs764945531 Chr.19: 45407251-45407251 on Build GRCh38
rs564538579 Chr.19: 45407457-45407457 on Build GRCh38
rs920286500 Chr.19: 45407271-45407271 on Build GRCh38
rs751152098 Chr.19: 45407269-45407269 on Build GRCh38
rs183371964 Chr.19: 45407373-45407373 on Build GRCh38
rs540904237 Chr.19: 45407498-45407498 on Build GRCh38
rs533771641 Chr.19: 45407678-45407678 on Build GRCh38

View Full Product Details