Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47349202-47349698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX34
Entrez Gene ID
9704
Gene Name
DEAH-box helicase 34
Gene Aliases
DDX34, HRH1
UniGene
Hs.151706
Gene Chromosome Location
Chr.19: 47349281-47382704 on Build GRCh38
Transcripts
NM_014681.5 XM_005259443.3

SNP Detail

SNP ID Chromosome Location
rs2859069 Chr.19: 47349457-47349457 on Build GRCh38
rs553933968 Chr.19: 47349283-47349283 on Build GRCh38
rs1002084634 Chr.19: 47349339-47349339 on Build GRCh38
rs545700105 Chr.19: 47349295-47349295 on Build GRCh38
rs913937133 Chr.19: 47349571-47349571 on Build GRCh38
rs922497099 Chr.19: 47349235-47349235 on Build GRCh38
rs960782192 Chr.19: 47349471-47349471 on Build GRCh38
rs181267350 Chr.19: 47349361-47349361 on Build GRCh38
rs962876530 Chr.19: 47349380-47349380 on Build GRCh38
rs942500002 Chr.19: 47349228-47349228 on Build GRCh38
rs565412759 Chr.19: 47349314-47349314 on Build GRCh38
rs562748380 Chr.19: 47349241-47349241 on Build GRCh38
rs781525789 Chr.19: 47349261-47349261 on Build GRCh38
rs984236786 Chr.19: 47349399-47349399 on Build GRCh38
rs1052343839 Chr.19: 47349414-47349414 on Build GRCh38
rs866355004 Chr.19: 47349538-47349538 on Build GRCh38
rs976569932 Chr.19: 47349453-47349453 on Build GRCh38
rs1043655946 Chr.19: 47349231-47349231 on Build GRCh38
rs1004504899 Chr.19: 47349362-47349362 on Build GRCh38
rs748420970 Chr.19: 47349521-47349521 on Build GRCh38
rs561256756 Chr.19: 47349347-47349347 on Build GRCh38
rs903438021 Chr.19: 47349276-47349276 on Build GRCh38
rs976716397 Chr.19: 47349233-47349233 on Build GRCh38
rs952788145 Chr.19: 47349371-47349371 on Build GRCh38
rs890656793 Chr.19: 47349425-47349425 on Build GRCh38
rs999156359 Chr.19: 47349409-47349409 on Build GRCh38
rs922549557 Chr.19: 47349550-47349550 on Build GRCh38
rs1017129985 Chr.19: 47349376-47349376 on Build GRCh38
rs980874544 Chr.19: 47349384-47349384 on Build GRCh38
rs377506442 Chr.19: 47349526-47349526 on Build GRCh38
rs761552171 Chr.19: 47349353-47349354 on Build GRCh38
rs148911212 Chr.19: 47349671-47349671 on Build GRCh38
rs186966545 Chr.19: 47349636-47349636 on Build GRCh38
rs903716375 Chr.19: 47349244-47349244 on Build GRCh38
rs1025941401 Chr.19: 47349357-47349357 on Build GRCh38
rs1045177326 Chr.19: 47349290-47349290 on Build GRCh38
rs2859068 Chr.19: 47349455-47349455 on Build GRCh38
rs547509398 Chr.19: 47349344-47349344 on Build GRCh38
rs988015886 Chr.19: 47349656-47349656 on Build GRCh38
rs926715889 Chr.19: 47349395-47349395 on Build GRCh38
rs527963747 Chr.19: 47349315-47349315 on Build GRCh38
rs1018753124 Chr.19: 47349402-47349402 on Build GRCh38
rs550250784 Chr.19: 47349363-47349363 on Build GRCh38
rs535409555 Chr.19: 47349259-47349259 on Build GRCh38
rs963937276 Chr.19: 47349432-47349432 on Build GRCh38
rs945355759 Chr.19: 47349610-47349610 on Build GRCh38
rs145547388 Chr.19: 47349279-47349279 on Build GRCh38
rs901565580 Chr.19: 47349282-47349282 on Build GRCh38
rs756524423 Chr.19: 47349664-47349664 on Build GRCh38

View Full Product Details