Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49799537-49800040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP2A1
Entrez Gene ID
160
Gene Name
adaptor related protein complex 2 alpha 1 subunit
Gene Aliases
ADTAA, AP2-ALPHA, CLAPA1
UniGene
Hs.467125
Gene Chromosome Location
Chr.19: 49766923-49807114 on Build GRCh38
Transcripts
NM_014203.2 NM_130787.2 XM_011526556.2 XM_011526557.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1304924 c.1196G>A p.R399Q Chr.19: 49799690-49799690 on Build GRCh38
2753316 c.1272C>T p.I424I Chr.19: 49799766-49799766 on Build GRCh38
5584468 c.1261C>T p.R421C Chr.19: 49799755-49799755 on Build GRCh38
999431 c.1155G>A p.V385V Chr.19: 49799649-49799649 on Build GRCh38
6601588 c.1244C>T p.T415M Chr.19: 49799738-49799738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745506973 Chr.19: 49799962-49799962 on Build GRCh38
rs543455936 Chr.19: 49799739-49799739 on Build GRCh38
rs918956811 Chr.19: 49799577-49799577 on Build GRCh38
rs1017229429 Chr.19: 49799858-49799858 on Build GRCh38
rs770288996 Chr.19: 49799982-49799982 on Build GRCh38
rs763288834 Chr.19: 49799796-49799797 on Build GRCh38
rs757374551 Chr.19: 49799620-49799620 on Build GRCh38
rs781014128 Chr.19: 49799943-49799943 on Build GRCh38
rs368684290 Chr.19: 49799688-49799688 on Build GRCh38
rs115948131 Chr.19: 49799579-49799579 on Build GRCh38
rs73590490 Chr.19: 49799941-49799941 on Build GRCh38
rs769699463 Chr.19: 49799754-49799755 on Build GRCh38
rs765347166 Chr.19: 49799717-49799717 on Build GRCh38
rs768733259 Chr.19: 49799942-49799943 on Build GRCh38
rs374039488 Chr.19: 49799748-49799748 on Build GRCh38
rs896697817 Chr.19: 49799957-49799957 on Build GRCh38
rs770655556 Chr.19: 49799791-49799791 on Build GRCh38
rs201906388 Chr.19: 49800000-49800000 on Build GRCh38
rs1026068157 Chr.19: 49799776-49799776 on Build GRCh38
rs377523946 Chr.19: 49799715-49799715 on Build GRCh38
rs185296139 Chr.19: 49799782-49799782 on Build GRCh38
rs1053985478 Chr.19: 49799575-49799575 on Build GRCh38
rs774463341 Chr.19: 49800004-49800004 on Build GRCh38
rs755250608 Chr.19: 49799942-49799942 on Build GRCh38
rs150789308 Chr.19: 49799745-49799745 on Build GRCh38
rs146010013 Chr.19: 49799991-49799991 on Build GRCh38
rs754378520 Chr.19: 49799714-49799714 on Build GRCh38
rs189554440 Chr.19: 49799923-49799923 on Build GRCh38
rs200754162 Chr.19: 49799964-49799964 on Build GRCh38
rs781118779 Chr.19: 49799651-49799651 on Build GRCh38
rs965003478 Chr.19: 49799899-49799899 on Build GRCh38
rs775137106 Chr.19: 49799758-49799760 on Build GRCh38
rs915533421 Chr.19: 49799826-49799826 on Build GRCh38
rs961724333 Chr.19: 49799670-49799670 on Build GRCh38
rs574649207 Chr.19: 49799578-49799578 on Build GRCh38
rs756655264 Chr.19: 49799595-49799595 on Build GRCh38
rs533348822 Chr.19: 49799811-49799811 on Build GRCh38
rs141155718 Chr.19: 49799742-49799742 on Build GRCh38
rs761892112 Chr.19: 49799956-49799956 on Build GRCh38
rs1054184861 Chr.19: 49799822-49799822 on Build GRCh38
rs376939716 Chr.19: 49799952-49799952 on Build GRCh38
rs770393212 Chr.19: 49799785-49799785 on Build GRCh38
rs770745252 Chr.19: 49799689-49799689 on Build GRCh38
rs759719552 Chr.19: 49800012-49800012 on Build GRCh38
rs761145930 Chr.19: 49799708-49799708 on Build GRCh38
rs764726365 Chr.19: 49799603-49799603 on Build GRCh38
rs777302536 Chr.19: 49799784-49799784 on Build GRCh38
rs764640528 Chr.19: 49799710-49799710 on Build GRCh38
rs1052458308 Chr.19: 49799641-49799641 on Build GRCh38
rs775953622 Chr.19: 49799690-49799690 on Build GRCh38
rs572245833 Chr.19: 49799630-49799630 on Build GRCh38
rs753923015 Chr.19: 49799617-49799617 on Build GRCh38
rs1010858993 Chr.19: 49799650-49799650 on Build GRCh38
rs565962932 Chr.19: 49799766-49799766 on Build GRCh38
rs778636010 Chr.19: 49799960-49799960 on Build GRCh38
rs768708447 Chr.19: 49799569-49799569 on Build GRCh38
rs149504055 Chr.19: 49799959-49799959 on Build GRCh38
rs746102918 Chr.19: 49799624-49799624 on Build GRCh38
rs766432944 Chr.19: 49800001-49800001 on Build GRCh38
rs370959134 Chr.19: 49799646-49799646 on Build GRCh38
rs570465699 Chr.19: 49799910-49799910 on Build GRCh38
rs774504131 Chr.19: 49799942-49799945 on Build GRCh38
rs774649528 Chr.19: 49799949-49799949 on Build GRCh38
rs750478513 Chr.19: 49799928-49799928 on Build GRCh38
rs551896115 Chr.19: 49799895-49799895 on Build GRCh38
rs750518472 Chr.19: 49799718-49799718 on Build GRCh38
rs531790097 Chr.19: 49799703-49799703 on Build GRCh38
rs138602676 Chr.19: 49799576-49799576 on Build GRCh38
rs1003423 Chr.19: 49799640-49799640 on Build GRCh38
rs747545364 Chr.19: 49799657-49799657 on Build GRCh38
rs373788390 Chr.19: 49799631-49799631 on Build GRCh38
rs199728232 Chr.19: 49799947-49799947 on Build GRCh38
rs982165838 Chr.19: 49799757-49799757 on Build GRCh38
rs758617042 Chr.19: 49799725-49799725 on Build GRCh38
rs748102575 Chr.19: 49799749-49799749 on Build GRCh38
rs748493956 Chr.19: 49799972-49799972 on Build GRCh38
rs377250932 Chr.19: 49799796-49799796 on Build GRCh38
rs751774802 Chr.19: 49799936-49799936 on Build GRCh38
rs374256135 Chr.19: 49799676-49799676 on Build GRCh38
rs374167558 Chr.19: 49799924-49799924 on Build GRCh38
rs372135964 Chr.19: 49799621-49799621 on Build GRCh38
rs1027601299 Chr.19: 49799681-49799681 on Build GRCh38
rs763032812 Chr.19: 49799933-49799933 on Build GRCh38
rs769341765 Chr.19: 49799661-49799661 on Build GRCh38
rs181564717 Chr.19: 49799954-49799954 on Build GRCh38
rs200552286 Chr.19: 49799935-49799935 on Build GRCh38
rs745735901 Chr.19: 49799649-49799658 on Build GRCh38

View Full Product Details