Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49811280-49811733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUZ
Entrez Gene ID
80199
Gene Name
fuzzy planar cell polarity protein
Gene Aliases
FY, NTD
UniGene
Hs.288800
Gene Chromosome Location
Chr.19: 49806866-49817376 on Build GRCh38
Transcripts
NM_001171937.1 NM_025129.4 NR_033269.1 XM_006723399.3 XM_011527339.1 XM_011527340.1 XM_011527341.2 XM_011527342.1 XM_011527343.1 XM_011527345.1 XM_011527346.1 XM_011527347.1 XM_017027319.1 XM_017027320.1 XM_017027321.1 XM_017027322.1 XM_017027323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6222073 c.463G>A p.V155M Chr.19: 49811392-49811392 on Build GRCh38
2753430 c.361G>A p.V121M Chr.19: 49811657-49811657 on Build GRCh38
4607188 c.425C>T p.S142L Chr.19: 49811430-49811430 on Build GRCh38
6326936 c.379G>A p.D127N Chr.19: 49811639-49811639 on Build GRCh38
5365026 c.426G>A p.S142S Chr.19: 49811429-49811429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764796000 Chr.19: 49811682-49811682 on Build GRCh38
rs773515920 Chr.19: 49811420-49811420 on Build GRCh38
rs1048505512 Chr.19: 49811652-49811652 on Build GRCh38
rs139116714 Chr.19: 49811419-49811419 on Build GRCh38
rs778023454 Chr.19: 49811713-49811713 on Build GRCh38
rs764082165 Chr.19: 49811525-49811525 on Build GRCh38
rs751785693 Chr.19: 49811429-49811429 on Build GRCh38
rs146191083 Chr.19: 49811399-49811399 on Build GRCh38
rs1036215896 Chr.19: 49811554-49811554 on Build GRCh38
rs35994116 Chr.19: 49811604-49811605 on Build GRCh38
rs755601216 Chr.19: 49811382-49811382 on Build GRCh38
rs780706540 Chr.19: 49811713-49811717 on Build GRCh38
rs778338671 Chr.19: 49811589-49811589 on Build GRCh38
rs781092276 Chr.19: 49811369-49811369 on Build GRCh38
rs769710496 Chr.19: 49811416-49811416 on Build GRCh38
rs1009581560 Chr.19: 49811473-49811473 on Build GRCh38
rs753599532 Chr.19: 49811359-49811359 on Build GRCh38
rs12973635 Chr.19: 49811481-49811481 on Build GRCh38
rs749797231 Chr.19: 49811592-49811592 on Build GRCh38
rs191877536 Chr.19: 49811602-49811602 on Build GRCh38
rs369633124 Chr.19: 49811374-49811374 on Build GRCh38
rs558749502 Chr.19: 49811338-49811339 on Build GRCh38
rs745980135 Chr.19: 49811642-49811643 on Build GRCh38
rs1003815357 Chr.19: 49811705-49811705 on Build GRCh38
rs757468560 Chr.19: 49811335-49811335 on Build GRCh38
rs756077603 Chr.19: 49811434-49811434 on Build GRCh38
rs760517460 Chr.19: 49811662-49811662 on Build GRCh38
rs748184407 Chr.19: 49811612-49811612 on Build GRCh38
rs199608357 Chr.19: 49811708-49811708 on Build GRCh38
rs781705103 Chr.19: 49811467-49811467 on Build GRCh38
rs35832552 Chr.19: 49811658-49811658 on Build GRCh38
rs761977723 Chr.19: 49811335-49811335 on Build GRCh38
rs777244657 Chr.19: 49811405-49811405 on Build GRCh38
rs752582696 Chr.19: 49811432-49811432 on Build GRCh38
rs753405783 Chr.19: 49811440-49811440 on Build GRCh38
rs756492726 Chr.19: 49811588-49811588 on Build GRCh38
rs539964458 Chr.19: 49811546-49811546 on Build GRCh38
rs775269115 Chr.19: 49811512-49811512 on Build GRCh38
rs781631990 Chr.19: 49811537-49811537 on Build GRCh38
rs748853432 Chr.19: 49811390-49811390 on Build GRCh38
rs766049090 Chr.19: 49811351-49811351 on Build GRCh38
rs756778438 Chr.19: 49811442-49811442 on Build GRCh38
rs778626921 Chr.19: 49811449-49811449 on Build GRCh38
rs373225684 Chr.19: 49811403-49811403 on Build GRCh38
rs774447548 Chr.19: 49811634-49811634 on Build GRCh38
rs904125079 Chr.19: 49811488-49811488 on Build GRCh38
rs774536898 Chr.19: 49811508-49811508 on Build GRCh38
rs764700213 Chr.19: 49811531-49811531 on Build GRCh38
rs748537209 Chr.19: 49811584-49811584 on Build GRCh38
rs765618793 Chr.19: 49811400-49811400 on Build GRCh38
rs751373878 Chr.19: 49811360-49811360 on Build GRCh38
rs767074844 Chr.19: 49811536-49811536 on Build GRCh38
rs763445960 Chr.19: 49811436-49811436 on Build GRCh38
rs373836926 Chr.19: 49811511-49811511 on Build GRCh38
rs780228231 Chr.19: 49811393-49811393 on Build GRCh38
rs376627265 Chr.19: 49811421-49811421 on Build GRCh38
rs753787518 Chr.19: 49811530-49811530 on Build GRCh38
rs763837948 Chr.19: 49811664-49811664 on Build GRCh38
rs35499921 Chr.19: 49811450-49811450 on Build GRCh38
rs759778616 Chr.19: 49811509-49811509 on Build GRCh38
rs930461774 Chr.19: 49811610-49811610 on Build GRCh38
rs574296569 Chr.19: 49811712-49811712 on Build GRCh38
rs571143206 Chr.19: 49811544-49811544 on Build GRCh38
rs769951235 Chr.19: 49811676-49811678 on Build GRCh38
rs767351327 Chr.19: 49811431-49811431 on Build GRCh38
rs766116099 Chr.19: 49811700-49811700 on Build GRCh38
rs750044002 Chr.19: 49811533-49811533 on Build GRCh38
rs759319192 Chr.19: 49811430-49811430 on Build GRCh38
rs750585789 Chr.19: 49811339-49811339 on Build GRCh38
rs891241885 Chr.19: 49811702-49811702 on Build GRCh38
rs1001121610 Chr.19: 49811495-49811495 on Build GRCh38
rs770626233 Chr.19: 49811392-49811392 on Build GRCh38
rs762455950 Chr.19: 49811409-49811409 on Build GRCh38
rs201963135 Chr.19: 49811672-49811672 on Build GRCh38
rs141513214 Chr.19: 49811364-49811364 on Build GRCh38
rs749373287 Chr.19: 49811482-49811482 on Build GRCh38
rs370940328 Chr.19: 49811657-49811657 on Build GRCh38
rs748621332 Chr.19: 49811470-49811470 on Build GRCh38
rs749953771 Chr.19: 49811686-49811686 on Build GRCh38
rs763254372 Chr.19: 49811341-49811341 on Build GRCh38
rs761676609 Chr.19: 49811681-49811681 on Build GRCh38
rs770338238 Chr.19: 49811471-49811471 on Build GRCh38
rs760551733 Chr.19: 49811523-49811523 on Build GRCh38
rs777365619 Chr.19: 49811388-49811388 on Build GRCh38
rs557828772 Chr.19: 49811550-49811550 on Build GRCh38
rs755369802 Chr.19: 49811455-49811455 on Build GRCh38
rs762240513 Chr.19: 49811308-49811308 on Build GRCh38
rs570362032 Chr.19: 49811703-49811703 on Build GRCh38
rs770981493 Chr.19: 49811487-49811487 on Build GRCh38
rs888484946 Chr.19: 49811397-49811397 on Build GRCh38

View Full Product Details