Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 56182657-56183199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALP
Entrez Gene ID
85569
Gene Name
galanin like peptide
Gene Aliases
-
UniGene
Hs.283915
Gene Chromosome Location
Chr.19: 56176020-56185775 on Build GRCh38
Transcripts
NM_001145546.1 NM_033106.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5568903 c.250T>C p.S84P Chr.19: 56183167-56183167 on Build GRCh38
714459 c.255G>T p.K85N Chr.19: 56183172-56183172 on Build GRCh38
4535632 c.220G>A p.G74R Chr.19: 56183137-56183137 on Build GRCh38
5427207 c.219C>T p.D73D Chr.19: 56183136-56183136 on Build GRCh38
6997121 c.246G>A p.Q82Q Chr.19: 56183163-56183163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs796080360 Chr.19: 56182708-56182709 on Build GRCh38
rs77780586 Chr.19: 56182845-56182845 on Build GRCh38
rs962741802 Chr.19: 56182898-56182898 on Build GRCh38
rs115827741 Chr.19: 56182680-56182680 on Build GRCh38
rs575756 Chr.19: 56183136-56183136 on Build GRCh38
rs367729331 Chr.19: 56183122-56183122 on Build GRCh38
rs767906480 Chr.19: 56183087-56183087 on Build GRCh38
rs746181030 Chr.19: 56183131-56183131 on Build GRCh38
rs58614188 Chr.19: 56183082-56183082 on Build GRCh38
rs761996012 Chr.19: 56183142-56183142 on Build GRCh38
rs974585138 Chr.19: 56182718-56182718 on Build GRCh38
rs545425086 Chr.19: 56182928-56182928 on Build GRCh38
rs758410051 Chr.19: 56183110-56183110 on Build GRCh38
rs1045459249 Chr.19: 56182914-56182914 on Build GRCh38
rs953175570 Chr.19: 56183036-56183036 on Build GRCh38
rs951224871 Chr.19: 56182966-56182966 on Build GRCh38
rs144859428 Chr.19: 56183052-56183052 on Build GRCh38
rs186406876 Chr.19: 56182994-56182994 on Build GRCh38
rs780838797 Chr.19: 56183127-56183127 on Build GRCh38
rs1035186942 Chr.19: 56182943-56182943 on Build GRCh38
rs756276029 Chr.19: 56182916-56182916 on Build GRCh38
rs936342870 Chr.19: 56183051-56183051 on Build GRCh38
rs564347741 Chr.19: 56182969-56182969 on Build GRCh38
rs867582053 Chr.19: 56182835-56182835 on Build GRCh38
rs367913346 Chr.19: 56182953-56182953 on Build GRCh38
rs3760852 Chr.19: 56183013-56183013 on Build GRCh38
rs988351283 Chr.19: 56182808-56182808 on Build GRCh38
rs368445767 Chr.19: 56183118-56183118 on Build GRCh38
rs918422559 Chr.19: 56182748-56182748 on Build GRCh38
rs141705754 Chr.19: 56183140-56183140 on Build GRCh38
rs274175 Chr.19: 56182758-56182758 on Build GRCh38
rs565438466 Chr.19: 56183088-56183088 on Build GRCh38
rs151245481 Chr.19: 56182733-56182733 on Build GRCh38
rs777826788 Chr.19: 56182889-56182889 on Build GRCh38
rs758943176 Chr.19: 56182917-56182918 on Build GRCh38
rs553470331 Chr.19: 56182868-56182868 on Build GRCh38
rs1036204490 Chr.19: 56182890-56182890 on Build GRCh38
rs1052649411 Chr.19: 56182827-56182827 on Build GRCh38
rs138776948 Chr.19: 56183103-56183103 on Build GRCh38
rs763149312 Chr.19: 56182757-56182757 on Build GRCh38
rs537618223 Chr.19: 56182839-56182839 on Build GRCh38
rs899074169 Chr.19: 56182895-56182895 on Build GRCh38
rs573234901 Chr.19: 56182925-56182925 on Build GRCh38
rs924941852 Chr.19: 56183043-56183043 on Build GRCh38
rs1035155648 Chr.19: 56182763-56182763 on Build GRCh38
rs375520472 Chr.19: 56182841-56182841 on Build GRCh38
rs567747236 Chr.19: 56183172-56183172 on Build GRCh38
rs180740441 Chr.19: 56182702-56182702 on Build GRCh38
rs367903825 Chr.19: 56182987-56182987 on Build GRCh38
rs570945213 Chr.19: 56183104-56183104 on Build GRCh38
rs754728042 Chr.19: 56182829-56182829 on Build GRCh38
rs975342144 Chr.19: 56183024-56183024 on Build GRCh38
rs936422792 Chr.19: 56182825-56182825 on Build GRCh38
rs780943015 Chr.19: 56182836-56182836 on Build GRCh38
rs921728247 Chr.19: 56182838-56182838 on Build GRCh38
rs74182590 Chr.19: 56182851-56182851 on Build GRCh38
rs750462010 Chr.19: 56182717-56182717 on Build GRCh38
rs770483064 Chr.19: 56182963-56182963 on Build GRCh38
rs765882204 Chr.19: 56182705-56182705 on Build GRCh38
rs112817488 Chr.19: 56183073-56183073 on Build GRCh38
rs942343427 Chr.19: 56182803-56182803 on Build GRCh38
rs112965468 Chr.19: 56183008-56183008 on Build GRCh38
rs746590539 Chr.19: 56183120-56183120 on Build GRCh38
rs1001813764 Chr.19: 56182885-56182885 on Build GRCh38
rs551915 Chr.19: 56182844-56182844 on Build GRCh38
rs765181454 Chr.19: 56183100-56183100 on Build GRCh38
rs781021584 Chr.19: 56183104-56183105 on Build GRCh38
rs1009729506 Chr.19: 56182980-56182980 on Build GRCh38
rs557982643 Chr.19: 56182805-56182805 on Build GRCh38
rs188267352 Chr.19: 56182683-56182683 on Build GRCh38
rs187988 Chr.19: 56182807-56182807 on Build GRCh38
rs745542001 Chr.19: 56183076-56183076 on Build GRCh38
rs995593953 Chr.19: 56182924-56182924 on Build GRCh38
rs748691962 Chr.19: 56182909-56182909 on Build GRCh38
rs1034331451 Chr.19: 56182888-56182888 on Build GRCh38
rs940565669 Chr.19: 56183077-56183077 on Build GRCh38
rs987143190 Chr.19: 56183063-56183063 on Build GRCh38
rs956823898 Chr.19: 56182954-56182954 on Build GRCh38
rs912499256 Chr.19: 56182771-56182771 on Build GRCh38
rs151198103 Chr.19: 56183137-56183137 on Build GRCh38
rs771896109 Chr.19: 56183099-56183099 on Build GRCh38

View Full Product Details