Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 3643932-3644424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLEC11
Entrez Gene ID
78989
Gene Name
collectin subfamily member 11
Gene Aliases
3MC2, CL-K1-I, CL-K1-II, CL-K1-IIa, CL-K1-IIb, CLK1
UniGene
Hs.32603
Gene Chromosome Location
Chr.2: 3594832-3644644 on Build GRCh38
Transcripts
NM_001255982.1 NM_001255983.1 NM_001255984.1 NM_001255985.1 NM_001255986.1 NM_001255987.1 NM_001255988.1 NM_001255989.1 NM_024027.4 NM_199235.2 NR_045659.1 XM_005263853.4 XM_006711897.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6633892 c.849G>T p.E283D Chr.2: 3644109-3644109 on Build GRCh38
7260850 c.626G>A p.G209D Chr.2: 3643937-3643937 on Build GRCh38
4094033 c.627C>T p.G209G Chr.2: 3643938-3643938 on Build GRCh38
4984912 c.698A>G p.H233R Chr.2: 3643958-3643958 on Build GRCh38
6633891 c.798G>T p.E266D Chr.2: 3644109-3644109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568274049 Chr.2: 3644362-3644362 on Build GRCh38
rs756469756 Chr.2: 3644049-3644049 on Build GRCh38
rs777781960 Chr.2: 3644224-3644224 on Build GRCh38
rs959650840 Chr.2: 3644272-3644272 on Build GRCh38
rs748080980 Chr.2: 3644119-3644119 on Build GRCh38
rs978970203 Chr.2: 3644287-3644287 on Build GRCh38
rs7567833 Chr.2: 3643958-3643958 on Build GRCh38
rs1047585093 Chr.2: 3644042-3644042 on Build GRCh38
rs767176074 Chr.2: 3644141-3644141 on Build GRCh38
rs777205136 Chr.2: 3644155-3644156 on Build GRCh38
rs200835001 Chr.2: 3644028-3644028 on Build GRCh38
rs199962584 Chr.2: 3643965-3643965 on Build GRCh38
rs781189973 Chr.2: 3644223-3644223 on Build GRCh38
rs750077755 Chr.2: 3643961-3643961 on Build GRCh38
rs774378733 Chr.2: 3644364-3644364 on Build GRCh38
rs947325126 Chr.2: 3644277-3644277 on Build GRCh38
rs764669422 Chr.2: 3644153-3644153 on Build GRCh38
rs750680074 Chr.2: 3644195-3644195 on Build GRCh38
rs565076370 Chr.2: 3644016-3644016 on Build GRCh38
rs778015880 Chr.2: 3643990-3643990 on Build GRCh38
rs770882319 Chr.2: 3644231-3644231 on Build GRCh38
rs760243603 Chr.2: 3644250-3644250 on Build GRCh38
rs751043966 Chr.2: 3644038-3644038 on Build GRCh38
rs761153465 Chr.2: 3644143-3644143 on Build GRCh38
rs1026486248 Chr.2: 3644139-3644139 on Build GRCh38
rs762808688 Chr.2: 3644136-3644136 on Build GRCh38
rs983557430 Chr.2: 3644346-3644346 on Build GRCh38
rs753234170 Chr.2: 3644142-3644142 on Build GRCh38
rs137922152 Chr.2: 3644059-3644059 on Build GRCh38
rs372572907 Chr.2: 3644050-3644050 on Build GRCh38
rs548461104 Chr.2: 3644311-3644311 on Build GRCh38
rs772174556 Chr.2: 3644107-3644107 on Build GRCh38
rs756455956 Chr.2: 3643988-3643988 on Build GRCh38
rs772944173 Chr.2: 3644134-3644134 on Build GRCh38
rs567826922 Chr.2: 3644238-3644238 on Build GRCh38
rs112999974 Chr.2: 3644176-3644176 on Build GRCh38
rs755606600 Chr.2: 3643967-3643967 on Build GRCh38
rs149123195 Chr.2: 3643992-3643992 on Build GRCh38
rs754095135 Chr.2: 3644165-3644165 on Build GRCh38
rs950845895 Chr.2: 3644330-3644330 on Build GRCh38
rs768525658 Chr.2: 3644007-3644007 on Build GRCh38
rs1001008242 Chr.2: 3644235-3644235 on Build GRCh38
rs758863458 Chr.2: 3644013-3644013 on Build GRCh38
rs780169464 Chr.2: 3644209-3644209 on Build GRCh38
rs573309933 Chr.2: 3644394-3644394 on Build GRCh38
rs941815801 Chr.2: 3644360-3644360 on Build GRCh38
rs368184468 Chr.2: 3643957-3643957 on Build GRCh38
rs768518451 Chr.2: 3644355-3644355 on Build GRCh38
rs1039665047 Chr.2: 3643950-3643950 on Build GRCh38
rs370041024 Chr.2: 3644254-3644254 on Build GRCh38
rs114885075 Chr.2: 3644140-3644140 on Build GRCh38
rs965069748 Chr.2: 3644191-3644191 on Build GRCh38
rs1021066752 Chr.2: 3644205-3644205 on Build GRCh38
rs975087111 Chr.2: 3644192-3644192 on Build GRCh38
rs774482090 Chr.2: 3644004-3644004 on Build GRCh38
rs762567908 Chr.2: 3644027-3644027 on Build GRCh38
rs114716171 Chr.2: 3644079-3644079 on Build GRCh38
rs770647475 Chr.2: 3644137-3644137 on Build GRCh38
rs553471012 Chr.2: 3644275-3644275 on Build GRCh38
rs781765198 Chr.2: 3643971-3643971 on Build GRCh38
rs753041393 Chr.2: 3643960-3643960 on Build GRCh38
rs115403931 Chr.2: 3644376-3644376 on Build GRCh38
rs895809395 Chr.2: 3644391-3644391 on Build GRCh38
rs982568460 Chr.2: 3644151-3644151 on Build GRCh38
rs201597655 Chr.2: 3643993-3643993 on Build GRCh38
rs934745308 Chr.2: 3644339-3644339 on Build GRCh38
rs748116580 Chr.2: 3644214-3644214 on Build GRCh38
rs745954101 Chr.2: 3644005-3644005 on Build GRCh38
rs1023668823 Chr.2: 3644073-3644073 on Build GRCh38
rs774795506 Chr.2: 3644246-3644246 on Build GRCh38
rs781080059 Chr.2: 3644114-3644114 on Build GRCh38
rs772894762 Chr.2: 3644017-3644017 on Build GRCh38
rs781393704 Chr.2: 3644385-3644385 on Build GRCh38
rs143185171 Chr.2: 3644019-3644019 on Build GRCh38
rs779069817 Chr.2: 3644174-3644174 on Build GRCh38
rs765634984 Chr.2: 3644358-3644358 on Build GRCh38
rs1033857901 Chr.2: 3644255-3644255 on Build GRCh38
rs759311267 Chr.2: 3644237-3644237 on Build GRCh38
rs140226372 Chr.2: 3644045-3644045 on Build GRCh38
rs749229386 Chr.2: 3644307-3644307 on Build GRCh38
rs1042911508 Chr.2: 3643969-3643969 on Build GRCh38
rs758446960 Chr.2: 3644207-3644207 on Build GRCh38
rs748988767 Chr.2: 3644227-3644227 on Build GRCh38
rs909312746 Chr.2: 3644353-3644353 on Build GRCh38
rs369567362 Chr.2: 3644046-3644046 on Build GRCh38
rs761633168 Chr.2: 3644010-3644010 on Build GRCh38
rs549923166 Chr.2: 3644367-3644367 on Build GRCh38
rs968212094 Chr.2: 3644211-3644211 on Build GRCh38
rs747243750 Chr.2: 3644130-3644130 on Build GRCh38
rs748943052 Chr.2: 3644301-3644301 on Build GRCh38
rs755642169 Chr.2: 3644153-3644154 on Build GRCh38
rs547553238 Chr.2: 3644058-3644058 on Build GRCh38
rs900431102 Chr.2: 3644126-3644126 on Build GRCh38
rs891539374 Chr.2: 3644159-3644159 on Build GRCh38
rs757665386 Chr.2: 3644171-3644171 on Build GRCh38
rs748463932 Chr.2: 3643987-3643987 on Build GRCh38
rs995391363 Chr.2: 3644105-3644107 on Build GRCh38
rs779122949 Chr.2: 3644061-3644061 on Build GRCh38

View Full Product Details