Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24038768-24039271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP1B
Entrez Gene ID
2281
Gene Name
FK506 binding protein 1B
Gene Aliases
FKBP12.6, FKBP1L, OTK4, PKBP1L, PPIase
UniGene
Hs.709461
Gene Chromosome Location
Chr.2: 24033205-24067743 on Build GRCh38
Transcripts
XM_017003594.1 NM_001142319.1 NM_025203.2 XM_017005029.1

Gene Symbol
WDCP
Entrez Gene ID
80304
Gene Name
WD repeat and coiled coil containing
Gene Aliases
C2orf44, PP384
UniGene
Hs.24624
Gene Chromosome Location
Chr.2: 24029336-24047426 on Build GRCh38
Transcripts
XM_017003594.1 NM_001142319.1 NM_025203.2 XM_017005029.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7080631 c.294C>T p.S98S Chr.2: 24039201-24039201 on Build GRCh38
7095454 c.574C>T p.H192Y Chr.2: 24038921-24038921 on Build GRCh38
7215631 c.305C>T p.T102M Chr.2: 24039190-24039190 on Build GRCh38
4751313 c.466C>T p.Q156* Chr.2: 24039029-24039029 on Build GRCh38
3426317 c.681G>T p.K227N Chr.2: 24038814-24038814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867072431 Chr.2: 24039071-24039071 on Build GRCh38
rs773991300 Chr.2: 24039196-24039196 on Build GRCh38
rs1037806808 Chr.2: 24038963-24038963 on Build GRCh38
rs749495440 Chr.2: 24038806-24038806 on Build GRCh38
rs906285733 Chr.2: 24038923-24038924 on Build GRCh38
rs151169389 Chr.2: 24038937-24038937 on Build GRCh38
rs750555223 Chr.2: 24039095-24039095 on Build GRCh38
rs747329318 Chr.2: 24038976-24038976 on Build GRCh38
rs776485759 Chr.2: 24038874-24038874 on Build GRCh38
rs747591625 Chr.2: 24039147-24039147 on Build GRCh38
rs747856335 Chr.2: 24039005-24039005 on Build GRCh38
rs756273255 Chr.2: 24038918-24038918 on Build GRCh38
rs895061534 Chr.2: 24038866-24038866 on Build GRCh38
rs769365186 Chr.2: 24039174-24039174 on Build GRCh38
rs766373388 Chr.2: 24038913-24038913 on Build GRCh38
rs374485799 Chr.2: 24039228-24039228 on Build GRCh38
rs767995016 Chr.2: 24038851-24038851 on Build GRCh38
rs1030922560 Chr.2: 24039146-24039146 on Build GRCh38
rs751392809 Chr.2: 24038916-24038916 on Build GRCh38
rs990161262 Chr.2: 24039175-24039175 on Build GRCh38
rs374740188 Chr.2: 24038825-24038825 on Build GRCh38
rs147098169 Chr.2: 24039045-24039045 on Build GRCh38
rs535818244 Chr.2: 24039148-24039148 on Build GRCh38
rs753935659 Chr.2: 24039050-24039050 on Build GRCh38
rs772768767 Chr.2: 24039217-24039217 on Build GRCh38
rs768280739 Chr.2: 24038964-24038964 on Build GRCh38
rs766071766 Chr.2: 24039023-24039023 on Build GRCh38
rs778066997 Chr.2: 24038803-24038803 on Build GRCh38
rs368152129 Chr.2: 24038880-24038880 on Build GRCh38
rs754095890 Chr.2: 24038936-24038936 on Build GRCh38
rs756078824 Chr.2: 24038798-24038798 on Build GRCh38
rs755422514 Chr.2: 24039158-24039158 on Build GRCh38
rs758856224 Chr.2: 24039123-24039123 on Build GRCh38
rs1022074851 Chr.2: 24039184-24039184 on Build GRCh38
rs781181716 Chr.2: 24038795-24038795 on Build GRCh38
rs1024794855 Chr.2: 24038904-24038904 on Build GRCh38
rs747459667 Chr.2: 24038961-24038961 on Build GRCh38
rs77397158 Chr.2: 24038822-24038822 on Build GRCh38
rs758621577 Chr.2: 24038949-24038949 on Build GRCh38
rs780625827 Chr.2: 24038960-24038960 on Build GRCh38
rs1047981710 Chr.2: 24038883-24038883 on Build GRCh38
rs765288990 Chr.2: 24039235-24039235 on Build GRCh38
rs534675761 Chr.2: 24038879-24038879 on Build GRCh38
rs769377257 Chr.2: 24038916-24038916 on Build GRCh38
rs372053685 Chr.2: 24038940-24038940 on Build GRCh38
rs780277835 Chr.2: 24039135-24039135 on Build GRCh38
rs112608137 Chr.2: 24039201-24039201 on Build GRCh38
rs900236268 Chr.2: 24038971-24038971 on Build GRCh38
rs748886296 Chr.2: 24039189-24039189 on Build GRCh38
rs776250480 Chr.2: 24038989-24038989 on Build GRCh38
rs764072892 Chr.2: 24038934-24038934 on Build GRCh38
rs3731620 Chr.2: 24039190-24039190 on Build GRCh38
rs149037014 Chr.2: 24038843-24038843 on Build GRCh38
rs752847613 Chr.2: 24038796-24038796 on Build GRCh38
rs762610244 Chr.2: 24038907-24038907 on Build GRCh38
rs765448263 Chr.2: 24039083-24039083 on Build GRCh38
rs775336979 Chr.2: 24039225-24039225 on Build GRCh38
rs764206842 Chr.2: 24039048-24039048 on Build GRCh38
rs886065651 Chr.2: 24038919-24038919 on Build GRCh38
rs774409475 Chr.2: 24039026-24039026 on Build GRCh38
rs760239130 Chr.2: 24038832-24038832 on Build GRCh38
rs999410002 Chr.2: 24039113-24039113 on Build GRCh38
rs375594658 Chr.2: 24039022-24039022 on Build GRCh38
rs143964812 Chr.2: 24039072-24039072 on Build GRCh38
rs367731925 Chr.2: 24039162-24039162 on Build GRCh38
rs199508606 Chr.2: 24038814-24038814 on Build GRCh38
rs371452340 Chr.2: 24038820-24038820 on Build GRCh38
rs772027729 Chr.2: 24039203-24039203 on Build GRCh38
rs747694862 Chr.2: 24039168-24039168 on Build GRCh38
rs552984011 Chr.2: 24039019-24039019 on Build GRCh38
rs751209925 Chr.2: 24038850-24038850 on Build GRCh38
rs773054259 Chr.2: 24039181-24039181 on Build GRCh38

View Full Product Details