Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24971736-24972239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC27
Entrez Gene ID
51277
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Gene Aliases
RBJ, RabJS
UniGene
Hs.434993
Gene Chromosome Location
Chr.2: 24943636-24972094 on Build GRCh38
Transcripts
NM_001198559.1 NM_016544.2 XM_011532902.1 NR_034113.1

Gene Symbol
DNAJC27-AS1
Entrez Gene ID
729723
Gene Name
DNAJC27 antisense RNA 1
Gene Aliases
-
UniGene
Hs.436366
Gene Chromosome Location
Chr.2: 24972112-25039694 on Build GRCh38
Transcripts
NM_001198559.1 NM_016544.2 XM_011532902.1 NR_034113.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6582058 c.43C>G p.L15V Chr.2: 24971862-24971862 on Build GRCh38
4583404 c.34G>C p.G12R Chr.2: 24971871-24971871 on Build GRCh38
4133746 c.33C>T p.P11P Chr.2: 24971872-24971872 on Build GRCh38
107340 c.27G>A p.K9K Chr.2: 24971878-24971878 on Build GRCh38
4093249 c.7G>A p.A3T Chr.2: 24971898-24971898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771267911 Chr.2: 24971940-24971940 on Build GRCh38
rs553932691 Chr.2: 24972110-24972110 on Build GRCh38
rs145542720 Chr.2: 24971865-24971865 on Build GRCh38
rs949784430 Chr.2: 24972022-24972022 on Build GRCh38
rs539639738 Chr.2: 24972184-24972184 on Build GRCh38
rs763840059 Chr.2: 24971903-24971903 on Build GRCh38
rs899858347 Chr.2: 24972125-24972125 on Build GRCh38
rs376869349 Chr.2: 24971852-24971852 on Build GRCh38
rs898345302 Chr.2: 24972035-24972035 on Build GRCh38
rs755961670 Chr.2: 24971769-24971769 on Build GRCh38
rs772968062 Chr.2: 24971809-24971809 on Build GRCh38
rs769208639 Chr.2: 24971796-24971796 on Build GRCh38
rs374439417 Chr.2: 24971791-24971791 on Build GRCh38
rs758489050 Chr.2: 24971907-24971907 on Build GRCh38
rs770620180 Chr.2: 24971871-24971871 on Build GRCh38
rs1001322879 Chr.2: 24971785-24971785 on Build GRCh38
rs182287849 Chr.2: 24972116-24972116 on Build GRCh38
rs903277217 Chr.2: 24972179-24972179 on Build GRCh38
rs760653495 Chr.2: 24971851-24971852 on Build GRCh38
rs777075339 Chr.2: 24971802-24971802 on Build GRCh38
rs768673712 Chr.2: 24971864-24971864 on Build GRCh38
rs187423055 Chr.2: 24972200-24972200 on Build GRCh38
rs755073750 Chr.2: 24971921-24971921 on Build GRCh38
rs1033299736 Chr.2: 24972185-24972185 on Build GRCh38
rs752698800 Chr.2: 24971838-24971838 on Build GRCh38
rs553041269 Chr.2: 24971922-24971922 on Build GRCh38
rs767055009 Chr.2: 24971877-24971877 on Build GRCh38
rs991836619 Chr.2: 24972190-24972190 on Build GRCh38
rs143830482 Chr.2: 24971878-24971878 on Build GRCh38
rs998204829 Chr.2: 24972134-24972134 on Build GRCh38
rs777596983 Chr.2: 24971841-24971841 on Build GRCh38
rs546931828 Chr.2: 24972081-24972081 on Build GRCh38
rs199963470 Chr.2: 24971925-24971925 on Build GRCh38
rs767431146 Chr.2: 24971829-24971829 on Build GRCh38
rs991170493 Chr.2: 24972017-24972017 on Build GRCh38
rs1032979713 Chr.2: 24972072-24972072 on Build GRCh38
rs747437213 Chr.2: 24971793-24971793 on Build GRCh38
rs779934261 Chr.2: 24971859-24971859 on Build GRCh38
rs975299240 Chr.2: 24972080-24972080 on Build GRCh38
rs573113068 Chr.2: 24972207-24972207 on Build GRCh38
rs773733477 Chr.2: 24971870-24971870 on Build GRCh38
rs755873615 Chr.2: 24971840-24971840 on Build GRCh38
rs190216543 Chr.2: 24971902-24971902 on Build GRCh38
rs774618261 Chr.2: 24972002-24972002 on Build GRCh38
rs912231969 Chr.2: 24972120-24972120 on Build GRCh38
rs770453121 Chr.2: 24971924-24971924 on Build GRCh38
rs758680294 Chr.2: 24971856-24971856 on Build GRCh38
rs931184641 Chr.2: 24972160-24972160 on Build GRCh38
rs575335585 Chr.2: 24972128-24972128 on Build GRCh38
rs1029723463 Chr.2: 24972151-24972151 on Build GRCh38
rs201971228 Chr.2: 24971861-24971861 on Build GRCh38
rs779104270 Chr.2: 24971775-24971775 on Build GRCh38
rs921000240 Chr.2: 24971919-24971919 on Build GRCh38
rs763985395 Chr.2: 24971910-24971910 on Build GRCh38
rs966946419 Chr.2: 24971851-24971851 on Build GRCh38
rs879722219 Chr.2: 24971895-24971895 on Build GRCh38
rs780306898 Chr.2: 24971914-24971914 on Build GRCh38
rs535460137 Chr.2: 24972114-24972114 on Build GRCh38
rs377674615 Chr.2: 24971799-24971799 on Build GRCh38
rs1055878382 Chr.2: 24971836-24971836 on Build GRCh38
rs1022853428 Chr.2: 24972192-24972192 on Build GRCh38
rs762251464 Chr.2: 24971804-24971804 on Build GRCh38
rs1050715990 Chr.2: 24971951-24971951 on Build GRCh38
rs1013629143 Chr.2: 24972055-24972055 on Build GRCh38
rs981131394 Chr.2: 24972033-24972033 on Build GRCh38
rs1032897280 Chr.2: 24972003-24972003 on Build GRCh38
rs986479047 Chr.2: 24972099-24972099 on Build GRCh38
rs561708577 Chr.2: 24971807-24971807 on Build GRCh38
rs751110283 Chr.2: 24971814-24971814 on Build GRCh38
rs752612919 Chr.2: 24971881-24971881 on Build GRCh38
rs978718182 Chr.2: 24971860-24971860 on Build GRCh38
rs966663137 Chr.2: 24972061-24972061 on Build GRCh38
rs977389495 Chr.2: 24971808-24971808 on Build GRCh38
rs368139378 Chr.2: 24971905-24971905 on Build GRCh38
rs918967405 Chr.2: 24972211-24972211 on Build GRCh38
rs957304122 Chr.2: 24972186-24972186 on Build GRCh38
rs7593864 Chr.2: 24971872-24971872 on Build GRCh38
rs749478598 Chr.2: 24971854-24971854 on Build GRCh38
rs751791811 Chr.2: 24971917-24971917 on Build GRCh38
rs568032861 Chr.2: 24972082-24972082 on Build GRCh38
rs1047677673 Chr.2: 24972066-24972066 on Build GRCh38
rs957520768 Chr.2: 24972014-24972014 on Build GRCh38
rs915613253 Chr.2: 24972021-24972021 on Build GRCh38
rs929086879 Chr.2: 24972218-24972218 on Build GRCh38
rs774023611 Chr.2: 24971941-24971941 on Build GRCh38
rs962381602 Chr.2: 24972031-24972031 on Build GRCh38
rs888701354 Chr.2: 24972074-24972074 on Build GRCh38
rs746132681 Chr.2: 24971778-24971778 on Build GRCh38
rs762498361 Chr.2: 24971981-24971981 on Build GRCh38
rs764657963 Chr.2: 24971866-24971866 on Build GRCh38
rs772027886 Chr.2: 24971789-24971789 on Build GRCh38
rs922464376 Chr.2: 24971806-24971806 on Build GRCh38
rs1036963121 Chr.2: 24972150-24972150 on Build GRCh38
rs754816370 Chr.2: 24971816-24971816 on Build GRCh38
rs967181451 Chr.2: 24972000-24972000 on Build GRCh38
rs978387727 Chr.2: 24972064-24972064 on Build GRCh38
rs765756817 Chr.2: 24971813-24971813 on Build GRCh38
rs775311094 Chr.2: 24971785-24971786 on Build GRCh38
rs749652378 Chr.2: 24971926-24971926 on Build GRCh38
rs959423354 Chr.2: 24972052-24972052 on Build GRCh38
rs898392121 Chr.2: 24972153-24972153 on Build GRCh38
rs949844396 Chr.2: 24971844-24971844 on Build GRCh38
rs770260817 Chr.2: 24971867-24971867 on Build GRCh38
rs534868695 Chr.2: 24972038-24972038 on Build GRCh38
rs759500264 Chr.2: 24972047-24972047 on Build GRCh38
rs932395979 Chr.2: 24972087-24972087 on Build GRCh38

View Full Product Details