Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 28392833-28393341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ31356
Entrez Gene ID
403150
Gene Name
uncharacterized protein FLJ31356
Gene Aliases
-
UniGene
Hs.562970
Gene Chromosome Location
Chr.2: 28384409-28394672 on Build GRCh38
Transcripts
NR_103831.1 NM_005253.3 XM_005264231.3 XM_006711976.2

Gene Symbol
FOSL2
Entrez Gene ID
2355
Gene Name
FOS like 2, AP-1 transcription factor subunit
Gene Aliases
FRA2
UniGene
Hs.220971
Gene Chromosome Location
Chr.2: 28392759-28417312 on Build GRCh38
Transcripts
NR_103831.1 NM_005253.3 XM_005264231.3 XM_006711976.2

SNP Detail

SNP ID Chromosome Location
rs1025607344 Chr.2: 28392925-28392925 on Build GRCh38
rs1057297601 Chr.2: 28393280-28393280 on Build GRCh38
rs562125065 Chr.2: 28393239-28393239 on Build GRCh38
rs1005642441 Chr.2: 28393318-28393318 on Build GRCh38
rs985007210 Chr.2: 28393085-28393085 on Build GRCh38
rs970933855 Chr.2: 28392905-28392905 on Build GRCh38
rs1012341229 Chr.2: 28393155-28393155 on Build GRCh38
rs540377415 Chr.2: 28393137-28393137 on Build GRCh38
rs756229009 Chr.2: 28392881-28392881 on Build GRCh38
rs72195155 Chr.2: 28392945-28392946 on Build GRCh38
rs903403762 Chr.2: 28392954-28392954 on Build GRCh38
rs902796676 Chr.2: 28393109-28393109 on Build GRCh38
rs924082762 Chr.2: 28393287-28393287 on Build GRCh38
rs935572330 Chr.2: 28393117-28393117 on Build GRCh38
rs933523682 Chr.2: 28393019-28393019 on Build GRCh38
rs922381938 Chr.2: 28393245-28393245 on Build GRCh38
rs902138504 Chr.2: 28392902-28392917 on Build GRCh38
rs1034080808 Chr.2: 28392935-28392935 on Build GRCh38
rs771626880 Chr.2: 28392961-28392961 on Build GRCh38
rs1053805110 Chr.2: 28393149-28393149 on Build GRCh38
rs971968943 Chr.2: 28393123-28393123 on Build GRCh38
rs1050598212 Chr.2: 28392946-28392946 on Build GRCh38
rs952841856 Chr.2: 28393082-28393082 on Build GRCh38
rs751553910 Chr.2: 28393150-28393150 on Build GRCh38
rs1013303675 Chr.2: 28393292-28393292 on Build GRCh38
rs1023471385 Chr.2: 28392892-28392892 on Build GRCh38
rs555320989 Chr.2: 28393140-28393140 on Build GRCh38
rs981935697 Chr.2: 28392921-28392921 on Build GRCh38
rs987626854 Chr.2: 28393277-28393277 on Build GRCh38
rs569299061 Chr.2: 28392926-28392926 on Build GRCh38
rs200363012 Chr.2: 28392945-28392945 on Build GRCh38
rs909048148 Chr.2: 28393103-28393103 on Build GRCh38
rs906614713 Chr.2: 28393168-28393168 on Build GRCh38
rs1022985500 Chr.2: 28393296-28393296 on Build GRCh38
rs1025471377 Chr.2: 28393178-28393178 on Build GRCh38
rs975096679 Chr.2: 28393240-28393240 on Build GRCh38
rs544135500 Chr.2: 28393227-28393227 on Build GRCh38
rs1051875977 Chr.2: 28393091-28393091 on Build GRCh38
rs573589381 Chr.2: 28393173-28393173 on Build GRCh38
rs940607235 Chr.2: 28392912-28392912 on Build GRCh38
rs189740937 Chr.2: 28393012-28393012 on Build GRCh38
rs995147107 Chr.2: 28393023-28393023 on Build GRCh38
rs749590372 Chr.2: 28392937-28392940 on Build GRCh38
rs748240757 Chr.2: 28392859-28392859 on Build GRCh38
rs868097685 Chr.2: 28393022-28393022 on Build GRCh38
rs909101550 Chr.2: 28392911-28392911 on Build GRCh38
rs964013865 Chr.2: 28393234-28393234 on Build GRCh38
rs896099142 Chr.2: 28393283-28393283 on Build GRCh38
rs952867468 Chr.2: 28393302-28393302 on Build GRCh38
rs960077568 Chr.2: 28392872-28392872 on Build GRCh38
rs1045592818 Chr.2: 28393157-28393157 on Build GRCh38
rs558196496 Chr.2: 28393000-28393000 on Build GRCh38
rs1028322602 Chr.2: 28393092-28393092 on Build GRCh38
rs1023457748 Chr.2: 28393073-28393073 on Build GRCh38
rs796941522 Chr.2: 28392946-28392947 on Build GRCh38
rs991974039 Chr.2: 28392883-28392883 on Build GRCh38
rs1012722875 Chr.2: 28392880-28392880 on Build GRCh38
rs909512668 Chr.2: 28392953-28392953 on Build GRCh38
rs1005451941 Chr.2: 28393211-28393211 on Build GRCh38
rs894149122 Chr.2: 28393017-28393017 on Build GRCh38
rs935588649 Chr.2: 28393303-28393303 on Build GRCh38
rs951291296 Chr.2: 28393197-28393197 on Build GRCh38
rs1003402557 Chr.2: 28393171-28393171 on Build GRCh38
rs951260110 Chr.2: 28392933-28392933 on Build GRCh38
rs1035991183 Chr.2: 28392923-28392923 on Build GRCh38
rs539470542 Chr.2: 28392947-28392947 on Build GRCh38
rs539487550 Chr.2: 28392857-28392857 on Build GRCh38
rs999069126 Chr.2: 28392986-28392986 on Build GRCh38
rs933037569 Chr.2: 28392940-28392940 on Build GRCh38
rs888896255 Chr.2: 28392949-28392949 on Build GRCh38
rs556774052 Chr.2: 28392941-28392942 on Build GRCh38
rs983819930 Chr.2: 28392944-28392944 on Build GRCh38
rs748627110 Chr.2: 28392946-28392951 on Build GRCh38
rs923225839 Chr.2: 28393138-28393138 on Build GRCh38
rs955106211 Chr.2: 28393254-28393254 on Build GRCh38

View Full Product Details