Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 33586474-33586981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM98A
Entrez Gene ID
25940
Gene Name
family with sequence similarity 98 member A
Gene Aliases
-
UniGene
Hs.468140
Gene Chromosome Location
Chr.2: 33583661-33599362 on Build GRCh38
Transcripts
NM_001304538.1 NM_015475.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4927904 c.662T>C p.L221P Chr.2: 33586620-33586620 on Build GRCh38
6660352 c.656G>A p.R219K Chr.2: 33586626-33586626 on Build GRCh38
5531022 c.652C>T p.R218W Chr.2: 33586630-33586630 on Build GRCh38
7144847 c.646G>A p.E216K Chr.2: 33586636-33586636 on Build GRCh38
4978489 c.700G>C p.G234R Chr.2: 33586582-33586582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747032069 Chr.2: 33586704-33586705 on Build GRCh38
rs192488161 Chr.2: 33586929-33586929 on Build GRCh38
rs758277653 Chr.2: 33586692-33586693 on Build GRCh38
rs779644340 Chr.2: 33586702-33586710 on Build GRCh38
rs201777834 Chr.2: 33586657-33586657 on Build GRCh38
rs267599362 Chr.2: 33586586-33586586 on Build GRCh38
rs751285617 Chr.2: 33586689-33586689 on Build GRCh38
rs750533093 Chr.2: 33586658-33586658 on Build GRCh38
rs138180530 Chr.2: 33586728-33586728 on Build GRCh38
rs752336301 Chr.2: 33586702-33586702 on Build GRCh38
rs539409961 Chr.2: 33586818-33586818 on Build GRCh38
rs866150970 Chr.2: 33586751-33586751 on Build GRCh38
rs938596881 Chr.2: 33586765-33586765 on Build GRCh38
rs901368445 Chr.2: 33586819-33586819 on Build GRCh38
rs138724969 Chr.2: 33586644-33586644 on Build GRCh38
rs746340280 Chr.2: 33586629-33586629 on Build GRCh38
rs1037025341 Chr.2: 33586566-33586566 on Build GRCh38
rs949900105 Chr.2: 33586947-33586947 on Build GRCh38
rs889837961 Chr.2: 33586815-33586815 on Build GRCh38
rs552123551 Chr.2: 33586567-33586567 on Build GRCh38
rs916960433 Chr.2: 33586940-33586940 on Build GRCh38
rs758837280 Chr.2: 33586791-33586791 on Build GRCh38
rs777370097 Chr.2: 33586720-33586720 on Build GRCh38
rs548009306 Chr.2: 33586645-33586645 on Build GRCh38
rs914287023 Chr.2: 33586722-33586722 on Build GRCh38
rs867470093 Chr.2: 33586692-33586692 on Build GRCh38
rs373779415 Chr.2: 33586699-33586699 on Build GRCh38
rs570182380 Chr.2: 33586498-33586498 on Build GRCh38
rs373278223 Chr.2: 33586538-33586538 on Build GRCh38
rs766314676 Chr.2: 33586673-33586673 on Build GRCh38
rs187875880 Chr.2: 33586779-33586779 on Build GRCh38
rs772728744 Chr.2: 33586630-33586630 on Build GRCh38
rs898930005 Chr.2: 33586618-33586618 on Build GRCh38
rs759961791 Chr.2: 33586514-33586514 on Build GRCh38
rs150613673 Chr.2: 33586525-33586528 on Build GRCh38
rs369303305 Chr.2: 33586530-33586530 on Build GRCh38
rs3213961 Chr.2: 33586869-33586869 on Build GRCh38
rs1037342290 Chr.2: 33586537-33586537 on Build GRCh38
rs965655500 Chr.2: 33586647-33586647 on Build GRCh38
rs910822468 Chr.2: 33586825-33586825 on Build GRCh38
rs745826153 Chr.2: 33586875-33586875 on Build GRCh38
rs751373554 Chr.2: 33586589-33586589 on Build GRCh38
rs147004373 Chr.2: 33586639-33586639 on Build GRCh38
rs769239876 Chr.2: 33586619-33586619 on Build GRCh38
rs1021049366 Chr.2: 33586787-33586787 on Build GRCh38
rs987606303 Chr.2: 33586817-33586817 on Build GRCh38
rs747683179 Chr.2: 33586614-33586614 on Build GRCh38
rs373615013 Chr.2: 33586555-33586555 on Build GRCh38
rs761864621 Chr.2: 33586559-33586559 on Build GRCh38
rs151046715 Chr.2: 33586621-33586621 on Build GRCh38
rs1044627061 Chr.2: 33586500-33586500 on Build GRCh38
rs777912636 Chr.2: 33586753-33586753 on Build GRCh38
rs770959031 Chr.2: 33586845-33586845 on Build GRCh38
rs765206074 Chr.2: 33586564-33586564 on Build GRCh38
rs989786991 Chr.2: 33586708-33586708 on Build GRCh38
rs748440465 Chr.2: 33586725-33586725 on Build GRCh38
rs370765954 Chr.2: 33586595-33586595 on Build GRCh38
rs536374008 Chr.2: 33586740-33586740 on Build GRCh38
rs1037378211 Chr.2: 33586769-33586769 on Build GRCh38
rs910768482 Chr.2: 33586513-33586513 on Build GRCh38
rs931275690 Chr.2: 33586800-33586800 on Build GRCh38
rs897091190 Chr.2: 33586798-33586798 on Build GRCh38
rs35346181 Chr.2: 33586638-33586638 on Build GRCh38
rs779280460 Chr.2: 33586653-33586653 on Build GRCh38
rs762824013 Chr.2: 33586663-33586663 on Build GRCh38
rs764238712 Chr.2: 33586546-33586546 on Build GRCh38
rs754790179 Chr.2: 33586690-33586690 on Build GRCh38
rs989735166 Chr.2: 33586948-33586948 on Build GRCh38
rs575654439 Chr.2: 33586856-33586856 on Build GRCh38
rs776773927 Chr.2: 33586640-33586640 on Build GRCh38
rs367623755 Chr.2: 33586706-33586706 on Build GRCh38
rs780775640 Chr.2: 33586730-33586730 on Build GRCh38
rs747248542 Chr.2: 33586805-33586805 on Build GRCh38
rs1031641301 Chr.2: 33586879-33586879 on Build GRCh38
rs750883868 Chr.2: 33586777-33586777 on Build GRCh38
rs371715760 Chr.2: 33586721-33586721 on Build GRCh38
rs928960164 Chr.2: 33586900-33586900 on Build GRCh38
rs775939274 Chr.2: 33586632-33586632 on Build GRCh38
rs758657484 Chr.2: 33586752-33586752 on Build GRCh38
rs377343388 Chr.2: 33586539-33586539 on Build GRCh38
rs377389251 Chr.2: 33586569-33586569 on Build GRCh38

View Full Product Details