Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 33588231-33588774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM98A
Entrez Gene ID
25940
Gene Name
family with sequence similarity 98 member A
Gene Aliases
-
UniGene
Hs.468140
Gene Chromosome Location
Chr.2: 33583661-33599362 on Build GRCh38
Transcripts
NM_001304538.1 NM_015475.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6370159 c.419G>T p.S140I Chr.2: 33588438-33588438 on Build GRCh38
2943191 c.399delA p.A134fs*13 Chr.2: 33588458-33588458 on Build GRCh38
4093992 c.414C>T p.G138G Chr.2: 33588443-33588443 on Build GRCh38
2943189 c.516A>G p.E172E Chr.2: 33588341-33588341 on Build GRCh38
370003 c.351A>T p.S117S Chr.2: 33588506-33588506 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1011206195 Chr.2: 33588719-33588719 on Build GRCh38
rs775558037 Chr.2: 33588290-33588290 on Build GRCh38
rs762587283 Chr.2: 33588367-33588367 on Build GRCh38
rs929582682 Chr.2: 33588350-33588350 on Build GRCh38
rs549521392 Chr.2: 33588292-33588292 on Build GRCh38
rs997993242 Chr.2: 33588572-33588572 on Build GRCh38
rs568120114 Chr.2: 33588270-33588270 on Build GRCh38
rs748873754 Chr.2: 33588520-33588520 on Build GRCh38
rs368754143 Chr.2: 33588480-33588480 on Build GRCh38
rs200115569 Chr.2: 33588442-33588442 on Build GRCh38
rs368717015 Chr.2: 33588319-33588319 on Build GRCh38
rs181897533 Chr.2: 33588412-33588412 on Build GRCh38
rs754198111 Chr.2: 33588555-33588555 on Build GRCh38
rs1002225387 Chr.2: 33588668-33588668 on Build GRCh38
rs185325771 Chr.2: 33588678-33588678 on Build GRCh38
rs768610960 Chr.2: 33588316-33588316 on Build GRCh38
rs746902133 Chr.2: 33588432-33588432 on Build GRCh38
rs111866703 Chr.2: 33588420-33588420 on Build GRCh38
rs756648457 Chr.2: 33588378-33588378 on Build GRCh38
rs764956125 Chr.2: 33588328-33588328 on Build GRCh38
rs892382293 Chr.2: 33588605-33588605 on Build GRCh38
rs189737024 Chr.2: 33588723-33588723 on Build GRCh38
rs529095574 Chr.2: 33588443-33588443 on Build GRCh38
rs557236906 Chr.2: 33588578-33588578 on Build GRCh38
rs779823631 Chr.2: 33588428-33588428 on Build GRCh38
rs761507794 Chr.2: 33588320-33588320 on Build GRCh38
rs1045852900 Chr.2: 33588457-33588457 on Build GRCh38
rs772245188 Chr.2: 33588419-33588419 on Build GRCh38
rs777424389 Chr.2: 33588380-33588381 on Build GRCh38
rs754218793 Chr.2: 33588386-33588386 on Build GRCh38
rs909577512 Chr.2: 33588394-33588394 on Build GRCh38
rs907364923 Chr.2: 33588609-33588609 on Build GRCh38
rs762345082 Chr.2: 33588481-33588481 on Build GRCh38
rs776029943 Chr.2: 33588566-33588571 on Build GRCh38
rs760141147 Chr.2: 33588528-33588529 on Build GRCh38
rs771353246 Chr.2: 33588523-33588524 on Build GRCh38
rs767894823 Chr.2: 33588373-33588373 on Build GRCh38
rs772430283 Chr.2: 33588558-33588559 on Build GRCh38
rs769323680 Chr.2: 33588455-33588455 on Build GRCh38
rs1051372800 Chr.2: 33588575-33588575 on Build GRCh38
rs1056045225 Chr.2: 33588671-33588671 on Build GRCh38
rs765875109 Chr.2: 33588483-33588483 on Build GRCh38
rs764392115 Chr.2: 33588349-33588349 on Build GRCh38
rs761115812 Chr.2: 33588534-33588534 on Build GRCh38
rs376329579 Chr.2: 33588298-33588298 on Build GRCh38
rs941070618 Chr.2: 33588403-33588403 on Build GRCh38
rs751095302 Chr.2: 33588488-33588488 on Build GRCh38
rs778729854 Chr.2: 33588582-33588582 on Build GRCh38
rs62150472 Chr.2: 33588381-33588381 on Build GRCh38
rs1039429451 Chr.2: 33588458-33588458 on Build GRCh38
rs899592124 Chr.2: 33588530-33588530 on Build GRCh38
rs1009475453 Chr.2: 33588645-33588645 on Build GRCh38
rs761701614 Chr.2: 33588444-33588444 on Build GRCh38
rs561820764 Chr.2: 33588359-33588359 on Build GRCh38
rs368454177 Chr.2: 33588537-33588537 on Build GRCh38
rs892834778 Chr.2: 33588714-33588714 on Build GRCh38
rs967093305 Chr.2: 33588725-33588725 on Build GRCh38
rs921959801 Chr.2: 33588259-33588259 on Build GRCh38
rs774566897 Chr.2: 33588526-33588527 on Build GRCh38
rs140113540 Chr.2: 33588387-33588387 on Build GRCh38
rs1049137996 Chr.2: 33588345-33588345 on Build GRCh38
rs779263605 Chr.2: 33588391-33588391 on Build GRCh38
rs753246522 Chr.2: 33588375-33588375 on Build GRCh38
rs778066212 Chr.2: 33588383-33588383 on Build GRCh38

View Full Product Details