Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 36558260-36558798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEZ2
Entrez Gene ID
9637
Gene Name
fasciculation and elongation protein zeta 2
Gene Aliases
HUM3CL
UniGene
Hs.258563
Gene Chromosome Location
Chr.2: 36552239-36598190 on Build GRCh38
Transcripts
NM_001042548.1 NM_005102.2 XM_006712153.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3773972 c.961C>T p.L321F Chr.2: 36558456-36558456 on Build GRCh38
7283778 c.938_939insA p.N313fs*7 Chr.2: 36558478-36558479 on Build GRCh38
3668787 c.973A>T p.T325S Chr.2: 36558444-36558444 on Build GRCh38
4611798 c.938delA p.N313fs*12 Chr.2: 36558479-36558479 on Build GRCh38
6261125 c.974C>A p.T325K Chr.2: 36558443-36558443 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs909661855 Chr.2: 36558550-36558550 on Build GRCh38
rs900252864 Chr.2: 36558314-36558314 on Build GRCh38
rs984345381 Chr.2: 36558477-36558477 on Build GRCh38
rs892259719 Chr.2: 36558398-36558398 on Build GRCh38
rs904647559 Chr.2: 36558717-36558717 on Build GRCh38
rs7423453 Chr.2: 36558487-36558487 on Build GRCh38
rs1056300799 Chr.2: 36558390-36558390 on Build GRCh38
rs76175556 Chr.2: 36558524-36558524 on Build GRCh38
rs559807059 Chr.2: 36558625-36558625 on Build GRCh38
rs1043571350 Chr.2: 36558664-36558664 on Build GRCh38
rs373644845 Chr.2: 36558373-36558373 on Build GRCh38
rs7423450 Chr.2: 36558459-36558459 on Build GRCh38
rs763389236 Chr.2: 36558687-36558688 on Build GRCh38
rs377685278 Chr.2: 36558660-36558660 on Build GRCh38
rs910103576 Chr.2: 36558646-36558646 on Build GRCh38
rs528260265 Chr.2: 36558524-36558525 on Build GRCh38
rs191174760 Chr.2: 36558489-36558489 on Build GRCh38
rs202179209 Chr.2: 36558469-36558469 on Build GRCh38
rs183170431 Chr.2: 36558325-36558325 on Build GRCh38
rs778955330 Chr.2: 36558541-36558544 on Build GRCh38
rs759258050 Chr.2: 36558478-36558479 on Build GRCh38
rs17019091 Chr.2: 36558316-36558316 on Build GRCh38
rs926250260 Chr.2: 36558615-36558615 on Build GRCh38
rs201880256 Chr.2: 36558473-36558473 on Build GRCh38
rs1034219816 Chr.2: 36558595-36558595 on Build GRCh38
rs939261921 Chr.2: 36558387-36558387 on Build GRCh38
rs775813780 Chr.2: 36558514-36558514 on Build GRCh38
rs768043408 Chr.2: 36558479-36558481 on Build GRCh38
rs1021471014 Chr.2: 36558564-36558564 on Build GRCh38
rs1056755121 Chr.2: 36558632-36558632 on Build GRCh38
rs1000242401 Chr.2: 36558742-36558742 on Build GRCh38
rs942968005 Chr.2: 36558651-36558651 on Build GRCh38
rs891563081 Chr.2: 36558720-36558720 on Build GRCh38
rs557218604 Chr.2: 36558366-36558366 on Build GRCh38
rs1010333401 Chr.2: 36558412-36558412 on Build GRCh38
rs536758709 Chr.2: 36558471-36558471 on Build GRCh38
rs1044696522 Chr.2: 36558332-36558332 on Build GRCh38
rs766063293 Chr.2: 36558443-36558443 on Build GRCh38
rs562760319 Chr.2: 36558744-36558744 on Build GRCh38
rs913753406 Chr.2: 36558638-36558638 on Build GRCh38
rs764285592 Chr.2: 36558710-36558710 on Build GRCh38
rs926573666 Chr.2: 36558382-36558382 on Build GRCh38
rs1009870929 Chr.2: 36558557-36558557 on Build GRCh38
rs982954656 Chr.2: 36558532-36558532 on Build GRCh38
rs980442875 Chr.2: 36558609-36558609 on Build GRCh38
rs756392461 Chr.2: 36558525-36558525 on Build GRCh38
rs758303930 Chr.2: 36558430-36558430 on Build GRCh38
rs757808767 Chr.2: 36558501-36558501 on Build GRCh38
rs142784384 Chr.2: 36558688-36558689 on Build GRCh38
rs998812039 Chr.2: 36558504-36558504 on Build GRCh38
rs868176299 Chr.2: 36558308-36558308 on Build GRCh38
rs546272888 Chr.2: 36558296-36558296 on Build GRCh38
rs544475780 Chr.2: 36558642-36558642 on Build GRCh38
rs760043724 Chr.2: 36558478-36558478 on Build GRCh38
rs960383833 Chr.2: 36558598-36558598 on Build GRCh38
rs904449912 Chr.2: 36558566-36558567 on Build GRCh38
rs754499390 Chr.2: 36558470-36558470 on Build GRCh38
rs72789258 Chr.2: 36558775-36558775 on Build GRCh38
rs368745549 Chr.2: 36558464-36558464 on Build GRCh38
rs975986007 Chr.2: 36558684-36558684 on Build GRCh38
rs753074314 Chr.2: 36558522-36558522 on Build GRCh38
rs956146846 Chr.2: 36558388-36558388 on Build GRCh38
rs968494089 Chr.2: 36558450-36558450 on Build GRCh38
rs577147078 Chr.2: 36558593-36558593 on Build GRCh38
rs183669426 Chr.2: 36558573-36558573 on Build GRCh38
rs188279916 Chr.2: 36558414-36558414 on Build GRCh38
rs758031425 Chr.2: 36558444-36558444 on Build GRCh38
rs1021400622 Chr.2: 36558417-36558417 on Build GRCh38
rs761527135 Chr.2: 36558411-36558412 on Build GRCh38
rs754238129 Chr.2: 36558525-36558526 on Build GRCh38
rs7423488 Chr.2: 36558665-36558665 on Build GRCh38
rs1039735764 Chr.2: 36558749-36558749 on Build GRCh38
rs951337042 Chr.2: 36558632-36558633 on Build GRCh38
rs541119817 Chr.2: 36558457-36558457 on Build GRCh38
rs750199049 Chr.2: 36558426-36558426 on Build GRCh38

View Full Product Details