Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 42360968-42361461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX7A2L
Entrez Gene ID
9167
Gene Name
cytochrome c oxidase subunit 7A2 like
Gene Aliases
COX7AR, COX7RP, EB1, SIG81
UniGene
Hs.339639
Gene Chromosome Location
Chr.2: 42335561-42368957 on Build GRCh38
Transcripts
NM_001319036.1 NM_001319037.1 NM_001319038.1 NM_001319040.1 NM_004718.3 NR_134947.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4518240 c.57_58GG>AA p.A20T Chr.2: 42361104-42361105 on Build GRCh38
6883013 c.49G>A p.A17T Chr.2: 42361113-42361113 on Build GRCh38
5397027 c.31A>T p.K11* Chr.2: 42361131-42361131 on Build GRCh38
721854 c.50C>A p.A17D Chr.2: 42361112-42361112 on Build GRCh38
6010092 c.29A>T p.Q10L Chr.2: 42361133-42361133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781228176 Chr.2: 42361160-42361160 on Build GRCh38
rs914573252 Chr.2: 42361251-42361251 on Build GRCh38
rs1019302345 Chr.2: 42361343-42361343 on Build GRCh38
rs139343885 Chr.2: 42361150-42361150 on Build GRCh38
rs868631325 Chr.2: 42360991-42360991 on Build GRCh38
rs1003908456 Chr.2: 42361311-42361311 on Build GRCh38
rs1011616502 Chr.2: 42361029-42361029 on Build GRCh38
rs768081401 Chr.2: 42361086-42361086 on Build GRCh38
rs768755431 Chr.2: 42361171-42361171 on Build GRCh38
rs898358745 Chr.2: 42361423-42361423 on Build GRCh38
rs776932045 Chr.2: 42361005-42361005 on Build GRCh38
rs1002684997 Chr.2: 42361174-42361174 on Build GRCh38
rs772535282 Chr.2: 42361136-42361136 on Build GRCh38
rs377157337 Chr.2: 42361022-42361022 on Build GRCh38
rs201202565 Chr.2: 42361184-42361184 on Build GRCh38
rs745999909 Chr.2: 42361050-42361050 on Build GRCh38
rs763458877 Chr.2: 42361018-42361018 on Build GRCh38
rs367547881 Chr.2: 42361164-42361164 on Build GRCh38
rs953709897 Chr.2: 42361434-42361434 on Build GRCh38
rs779233293 Chr.2: 42361172-42361172 on Build GRCh38
rs761170814 Chr.2: 42361183-42361183 on Build GRCh38
rs775894079 Chr.2: 42361057-42361057 on Build GRCh38
rs765066970 Chr.2: 42361091-42361091 on Build GRCh38
rs781325675 Chr.2: 42361116-42361116 on Build GRCh38
rs886756726 Chr.2: 42361309-42361309 on Build GRCh38
rs958598305 Chr.2: 42361026-42361026 on Build GRCh38
rs111915858 Chr.2: 42361092-42361092 on Build GRCh38
rs747084929 Chr.2: 42361001-42361001 on Build GRCh38
rs747043515 Chr.2: 42360993-42360993 on Build GRCh38
rs951080701 Chr.2: 42361231-42361231 on Build GRCh38
rs769006567 Chr.2: 42361128-42361128 on Build GRCh38
rs751560935 Chr.2: 42361229-42361229 on Build GRCh38
rs765619044 Chr.2: 42361007-42361007 on Build GRCh38
rs774564757 Chr.2: 42361130-42361130 on Build GRCh38
rs1018085023 Chr.2: 42361419-42361419 on Build GRCh38
rs746886088 Chr.2: 42361106-42361106 on Build GRCh38
rs564587690 Chr.2: 42361228-42361228 on Build GRCh38
rs111565073 Chr.2: 42361175-42361175 on Build GRCh38
rs200842940 Chr.2: 42361176-42361176 on Build GRCh38
rs745922132 Chr.2: 42361118-42361118 on Build GRCh38
rs966186302 Chr.2: 42361223-42361223 on Build GRCh38
rs932070243 Chr.2: 42361100-42361100 on Build GRCh38
rs759674646 Chr.2: 42361232-42361232 on Build GRCh38
rs778597421 Chr.2: 42361035-42361035 on Build GRCh38
rs952289930 Chr.2: 42361046-42361046 on Build GRCh38
rs143748823 Chr.2: 42361162-42361162 on Build GRCh38
rs550304803 Chr.2: 42361370-42361370 on Build GRCh38
rs990251030 Chr.2: 42361438-42361438 on Build GRCh38
rs981034710 Chr.2: 42361436-42361436 on Build GRCh38
rs765293030 Chr.2: 42361277-42361277 on Build GRCh38
rs938615639 Chr.2: 42361280-42361280 on Build GRCh38
rs966259892 Chr.2: 42361413-42361413 on Build GRCh38
rs928941758 Chr.2: 42361254-42361254 on Build GRCh38
rs776881420 Chr.2: 42361040-42361040 on Build GRCh38
rs1041709217 Chr.2: 42361192-42361192 on Build GRCh38
rs894307802 Chr.2: 42361290-42361290 on Build GRCh38
rs762843003 Chr.2: 42361159-42361159 on Build GRCh38
rs1027190254 Chr.2: 42361308-42361308 on Build GRCh38
rs749476147 Chr.2: 42361124-42361124 on Build GRCh38
rs758994060 Chr.2: 42361147-42361147 on Build GRCh38
rs140344601 Chr.2: 42361143-42361143 on Build GRCh38
rs973677002 Chr.2: 42361133-42361133 on Build GRCh38
rs763634759 Chr.2: 42361188-42361188 on Build GRCh38
rs777489565 Chr.2: 42361439-42361439 on Build GRCh38
rs773665383 Chr.2: 42361178-42361178 on Build GRCh38
rs770952664 Chr.2: 42361004-42361004 on Build GRCh38
rs116570696 Chr.2: 42361291-42361291 on Build GRCh38
rs755792396 Chr.2: 42361433-42361433 on Build GRCh38
rs377160921 Chr.2: 42361047-42361047 on Build GRCh38
rs546072265 Chr.2: 42361093-42361093 on Build GRCh38
rs764842230 Chr.2: 42361095-42361095 on Build GRCh38
rs374152961 Chr.2: 42361198-42361198 on Build GRCh38
rs995206056 Chr.2: 42361428-42361428 on Build GRCh38
rs1020038285 Chr.2: 42361002-42361002 on Build GRCh38
rs1052204096 Chr.2: 42361372-42361372 on Build GRCh38
rs758961371 Chr.2: 42361193-42361193 on Build GRCh38
rs764630652 Chr.2: 42361019-42361019 on Build GRCh38
rs894199803 Chr.2: 42361179-42361179 on Build GRCh38
rs550968076 Chr.2: 42361194-42361194 on Build GRCh38
rs762232539 Chr.2: 42361085-42361085 on Build GRCh38
rs780104612 Chr.2: 42361206-42361206 on Build GRCh38
rs17850163 Chr.2: 42361141-42361141 on Build GRCh38
rs772036511 Chr.2: 42360995-42360995 on Build GRCh38
rs753828368 Chr.2: 42361209-42361209 on Build GRCh38
rs879005698 Chr.2: 42361324-42361324 on Build GRCh38
rs780507723 Chr.2: 42361203-42361203 on Build GRCh38
rs774861178 Chr.2: 42361185-42361185 on Build GRCh38
rs762567024 Chr.2: 42361187-42361187 on Build GRCh38
rs767425420 Chr.2: 42361201-42361201 on Build GRCh38
rs748436653 Chr.2: 42361173-42361173 on Build GRCh38
rs752340777 Chr.2: 42361152-42361152 on Build GRCh38
rs762502741 Chr.2: 42361153-42361153 on Build GRCh38
rs751249198 Chr.2: 42361190-42361190 on Build GRCh38
rs567834401 Chr.2: 42361406-42361406 on Build GRCh38
rs371146473 Chr.2: 42361084-42361084 on Build GRCh38
rs773075232 Chr.2: 42360996-42360996 on Build GRCh38
rs757125805 Chr.2: 42361107-42361107 on Build GRCh38
rs3764906 Chr.2: 42361055-42361055 on Build GRCh38
rs765820609 Chr.2: 42361028-42361028 on Build GRCh38
rs770973680 Chr.2: 42361048-42361048 on Build GRCh38
rs375801131 Chr.2: 42361205-42361205 on Build GRCh38
rs373291536 Chr.2: 42361066-42361066 on Build GRCh38
rs754534079 Chr.2: 42361094-42361094 on Build GRCh38
rs990649662 Chr.2: 42361242-42361242 on Build GRCh38
rs1006688333 Chr.2: 42361208-42361208 on Build GRCh38
rs1011674723 Chr.2: 42361191-42361191 on Build GRCh38
rs951048162 Chr.2: 42361435-42361435 on Build GRCh38
rs752382071 Chr.2: 42361097-42361097 on Build GRCh38
rs750328081 Chr.2: 42361204-42361204 on Build GRCh38
rs559061083 Chr.2: 42361006-42361006 on Build GRCh38
rs530520671 Chr.2: 42361317-42361317 on Build GRCh38
rs763336757 Chr.2: 42361060-42361060 on Build GRCh38
rs773755117 Chr.2: 42361142-42361142 on Build GRCh38
rs754494173 Chr.2: 42361236-42361236 on Build GRCh38
rs755131565 Chr.2: 42361212-42361212 on Build GRCh38
rs573788850 Chr.2: 42361123-42361123 on Build GRCh38
rs758171642 Chr.2: 42361045-42361045 on Build GRCh38
rs891013883 Chr.2: 42361386-42361386 on Build GRCh38
rs752385712 Chr.2: 42361420-42361420 on Build GRCh38
rs149297722 Chr.2: 42361120-42361120 on Build GRCh38
rs937591832 Chr.2: 42361260-42361260 on Build GRCh38
rs754623670 Chr.2: 42361034-42361034 on Build GRCh38
rs563462982 Chr.2: 42361102-42361102 on Build GRCh38
rs776843991 Chr.2: 42361049-42361049 on Build GRCh38
rs138027067 Chr.2: 42361135-42361135 on Build GRCh38
rs946090696 Chr.2: 42361265-42361265 on Build GRCh38
rs141288244 Chr.2: 42361158-42361158 on Build GRCh38
rs202062223 Chr.2: 42361170-42361170 on Build GRCh38
rs962782679 Chr.2: 42361111-42361111 on Build GRCh38
rs535710805 Chr.2: 42361169-42361169 on Build GRCh38
rs766989036 Chr.2: 42361361-42361361 on Build GRCh38
rs775982023 Chr.2: 42361009-42361009 on Build GRCh38
rs1055512433 Chr.2: 42361285-42361285 on Build GRCh38
rs1041599112 Chr.2: 42361276-42361276 on Build GRCh38

View Full Product Details