Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 43794318-43794820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC2LI1
Entrez Gene ID
51626
Gene Name
dynein cytoplasmic 2 light intermediate chain 1
Gene Aliases
CGI-60, D2LIC, LIC3
UniGene
Hs.371597
Gene Chromosome Location
Chr.2: 43774039-43828492 on Build GRCh38
Transcripts
NM_001193464.1 NM_015522.3 NM_016008.3 XM_005264364.4 XM_005264365.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1021046 c.495G>T p.P165P Chr.2: 43794631-43794631 on Build GRCh38
6226370 c.425T>A p.I142K Chr.2: 43794561-43794561 on Build GRCh38
1021045 c.494C>T p.P165L Chr.2: 43794630-43794630 on Build GRCh38
3426474 c.327T>G p.F109L Chr.2: 43794463-43794463 on Build GRCh38
6440301 c.539C>T p.S180F Chr.2: 43794675-43794675 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768949011 Chr.2: 43794499-43794499 on Build GRCh38
rs115529828 Chr.2: 43794627-43794627 on Build GRCh38
rs755134375 Chr.2: 43794651-43794651 on Build GRCh38
rs756047729 Chr.2: 43794646-43794646 on Build GRCh38
rs766443093 Chr.2: 43794647-43794647 on Build GRCh38
rs146348931 Chr.2: 43794459-43794459 on Build GRCh38
rs769975073 Chr.2: 43794508-43794508 on Build GRCh38
rs770155116 Chr.2: 43794554-43794554 on Build GRCh38
rs779132687 Chr.2: 43794579-43794579 on Build GRCh38
rs755258671 Chr.2: 43794573-43794573 on Build GRCh38
rs528204878 Chr.2: 43794783-43794783 on Build GRCh38
rs964128087 Chr.2: 43794483-43794483 on Build GRCh38
rs901926147 Chr.2: 43794518-43794518 on Build GRCh38
rs749444608 Chr.2: 43794567-43794567 on Build GRCh38
rs772694425 Chr.2: 43794524-43794524 on Build GRCh38
rs758741774 Chr.2: 43794685-43794685 on Build GRCh38
rs762694520 Chr.2: 43794624-43794624 on Build GRCh38
rs748351081 Chr.2: 43794668-43794668 on Build GRCh38
rs775807051 Chr.2: 43794409-43794409 on Build GRCh38
rs368166400 Chr.2: 43794710-43794710 on Build GRCh38
rs1045603641 Chr.2: 43794578-43794578 on Build GRCh38
rs996902213 Chr.2: 43794765-43794765 on Build GRCh38
rs371018698 Chr.2: 43794601-43794601 on Build GRCh38
rs578159100 Chr.2: 43794631-43794631 on Build GRCh38
rs779237011 Chr.2: 43794661-43794661 on Build GRCh38
rs758230490 Chr.2: 43794460-43794460 on Build GRCh38
rs781115586 Chr.2: 43794475-43794475 on Build GRCh38
rs774670800 Chr.2: 43794423-43794423 on Build GRCh38
rs551245502 Chr.2: 43794785-43794785 on Build GRCh38
rs780118430 Chr.2: 43794564-43794564 on Build GRCh38
rs551574531 Chr.2: 43794350-43794350 on Build GRCh38
rs1046443256 Chr.2: 43794388-43794388 on Build GRCh38
rs776935418 Chr.2: 43794476-43794476 on Build GRCh38
rs201948500 Chr.2: 43794485-43794485 on Build GRCh38
rs1030694932 Chr.2: 43794546-43794546 on Build GRCh38
rs953304636 Chr.2: 43794558-43794558 on Build GRCh38
rs995776158 Chr.2: 43794375-43794375 on Build GRCh38
rs751399000 Chr.2: 43794467-43794467 on Build GRCh38
rs773057615 Chr.2: 43794746-43794746 on Build GRCh38
rs773981319 Chr.2: 43794630-43794630 on Build GRCh38
rs200402219 Chr.2: 43794703-43794703 on Build GRCh38
rs34651474 Chr.2: 43794537-43794537 on Build GRCh38
rs1047151200 Chr.2: 43794676-43794676 on Build GRCh38
rs759159149 Chr.2: 43794529-43794529 on Build GRCh38
rs528201408 Chr.2: 43794525-43794525 on Build GRCh38
rs535890611 Chr.2: 43794462-43794462 on Build GRCh38
rs767274747 Chr.2: 43794734-43794734 on Build GRCh38
rs756137309 Chr.2: 43794560-43794560 on Build GRCh38
rs764641476 Chr.2: 43794648-43794648 on Build GRCh38
rs752351797 Chr.2: 43794532-43794532 on Build GRCh38
rs747544028 Chr.2: 43794605-43794605 on Build GRCh38
rs760592460 Chr.2: 43794757-43794757 on Build GRCh38
rs559002219 Chr.2: 43794764-43794764 on Build GRCh38
rs765740446 Chr.2: 43794432-43794432 on Build GRCh38
rs753456602 Chr.2: 43794449-43794449 on Build GRCh38
rs766353351 Chr.2: 43794759-43794759 on Build GRCh38
rs368515052 Chr.2: 43794587-43794587 on Build GRCh38
rs769038785 Chr.2: 43794421-43794421 on Build GRCh38
rs1013166433 Chr.2: 43794371-43794371 on Build GRCh38
rs759097386 Chr.2: 43794451-43794451 on Build GRCh38
rs199776942 Chr.2: 43794351-43794351 on Build GRCh38
rs141762000 Chr.2: 43794534-43794534 on Build GRCh38
rs750300990 Chr.2: 43794639-43794639 on Build GRCh38
rs771636494 Chr.2: 43794700-43794700 on Build GRCh38
rs780205827 Chr.2: 43794497-43794497 on Build GRCh38
rs144608735 Chr.2: 43794586-43794586 on Build GRCh38
rs115668893 Chr.2: 43794431-43794431 on Build GRCh38
rs775149572 Chr.2: 43794623-43794623 on Build GRCh38
rs9309107 Chr.2: 43794687-43794687 on Build GRCh38
rs773433083 Chr.2: 43794715-43794717 on Build GRCh38
rs762222236 Chr.2: 43794521-43794521 on Build GRCh38
rs373759547 Chr.2: 43794559-43794559 on Build GRCh38
rs1034504878 Chr.2: 43794470-43794470 on Build GRCh38
rs748843775 Chr.2: 43794708-43794708 on Build GRCh38
rs898123684 Chr.2: 43794354-43794354 on Build GRCh38
rs757227264 Chr.2: 43794473-43794473 on Build GRCh38
rs3749062 Chr.2: 43794417-43794417 on Build GRCh38
rs907470588 Chr.2: 43794718-43794718 on Build GRCh38
rs571417337 Chr.2: 43794399-43794399 on Build GRCh38
rs752908990 Chr.2: 43794787-43794787 on Build GRCh38
rs772788154 Chr.2: 43794547-43794547 on Build GRCh38
rs928424298 Chr.2: 43794585-43794585 on Build GRCh38
rs749532514 Chr.2: 43794498-43794498 on Build GRCh38
rs762199372 Chr.2: 43794503-43794503 on Build GRCh38
rs57240987 Chr.2: 43794690-43794690 on Build GRCh38
rs775114768 Chr.2: 43794707-43794707 on Build GRCh38
rs763733041 Chr.2: 43794536-43794536 on Build GRCh38
rs767566586 Chr.2: 43794442-43794442 on Build GRCh38
rs769373522 Chr.2: 43794617-43794617 on Build GRCh38
rs76483206 Chr.2: 43794530-43794530 on Build GRCh38
rs748595118 Chr.2: 43794583-43794583 on Build GRCh38

View Full Product Details