Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85363006-85363502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMOD3
Entrez Gene ID
84173
Gene Name
ELMO domain containing 3
Gene Aliases
DFNB88, LST3, RBED1, RBM29
UniGene
Hs.269990
Gene Chromosome Location
Chr.2: 85354394-85391752 on Build GRCh38
Transcripts
NM_001135021.1 NM_001135022.1 NM_001135023.1 NM_032213.4 XM_005264596.3 XM_005264597.3 XM_005264598.3 XM_017005078.1 XM_017005079.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1251130 c.192G>A p.E64E Chr.2: 85363159-85363159 on Build GRCh38
1615171 c.139T>A p.L47M Chr.2: 85363106-85363106 on Build GRCh38
3426726 c.168G>C p.L56L Chr.2: 85363135-85363135 on Build GRCh38
6049661 c.196C>T p.R66C Chr.2: 85363163-85363163 on Build GRCh38
7167815 c.144G>A p.K48K Chr.2: 85363111-85363111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776264234 Chr.2: 85363284-85363285 on Build GRCh38
rs772120151 Chr.2: 85363115-85363115 on Build GRCh38
rs960898143 Chr.2: 85363243-85363243 on Build GRCh38
rs199716491 Chr.2: 85363084-85363084 on Build GRCh38
rs983146978 Chr.2: 85363226-85363226 on Build GRCh38
rs747662366 Chr.2: 85363213-85363230 on Build GRCh38
rs1008474484 Chr.2: 85363135-85363135 on Build GRCh38
rs374954571 Chr.2: 85363164-85363164 on Build GRCh38
rs867976576 Chr.2: 85363380-85363380 on Build GRCh38
rs756654016 Chr.2: 85363131-85363131 on Build GRCh38
rs529077376 Chr.2: 85363177-85363177 on Build GRCh38
rs764268462 Chr.2: 85363097-85363097 on Build GRCh38
rs754245156 Chr.2: 85363281-85363281 on Build GRCh38
rs930145038 Chr.2: 85363120-85363120 on Build GRCh38
rs764321523 Chr.2: 85363201-85363201 on Build GRCh38
rs766348719 Chr.2: 85363172-85363172 on Build GRCh38
rs759718576 Chr.2: 85363102-85363102 on Build GRCh38
rs985450857 Chr.2: 85363064-85363064 on Build GRCh38
rs927697724 Chr.2: 85363472-85363483 on Build GRCh38
rs745897843 Chr.2: 85363101-85363101 on Build GRCh38
rs751647172 Chr.2: 85363206-85363206 on Build GRCh38
rs762312309 Chr.2: 85363053-85363053 on Build GRCh38
rs569552270 Chr.2: 85363463-85363463 on Build GRCh38
rs758488759 Chr.2: 85363095-85363095 on Build GRCh38
rs375589370 Chr.2: 85363271-85363271 on Build GRCh38
rs781711502 Chr.2: 85363129-85363129 on Build GRCh38
rs7564372 Chr.2: 85363163-85363163 on Build GRCh38
rs968745968 Chr.2: 85363042-85363042 on Build GRCh38
rs539757873 Chr.2: 85363279-85363279 on Build GRCh38
rs747840762 Chr.2: 85363160-85363160 on Build GRCh38
rs372284756 Chr.2: 85363082-85363082 on Build GRCh38
rs368317282 Chr.2: 85363077-85363077 on Build GRCh38
rs770157563 Chr.2: 85363050-85363050 on Build GRCh38
rs371908680 Chr.2: 85363140-85363140 on Build GRCh38
rs563364483 Chr.2: 85363052-85363052 on Build GRCh38
rs1011900657 Chr.2: 85363169-85363169 on Build GRCh38
rs759536096 Chr.2: 85363182-85363182 on Build GRCh38
rs773826786 Chr.2: 85363144-85363144 on Build GRCh38
rs369014095 Chr.2: 85363216-85363216 on Build GRCh38
rs776219800 Chr.2: 85363121-85363122 on Build GRCh38
rs749797307 Chr.2: 85363139-85363139 on Build GRCh38
rs746615494 Chr.2: 85363108-85363110 on Build GRCh38
rs906169812 Chr.2: 85363256-85363256 on Build GRCh38
rs746166344 Chr.2: 85363130-85363130 on Build GRCh38
rs369240642 Chr.2: 85363116-85363116 on Build GRCh38
rs1052530031 Chr.2: 85363405-85363405 on Build GRCh38
rs780357560 Chr.2: 85363138-85363138 on Build GRCh38
rs989833561 Chr.2: 85363032-85363032 on Build GRCh38
rs1018383590 Chr.2: 85363137-85363137 on Build GRCh38
rs1010738770 Chr.2: 85363340-85363340 on Build GRCh38
rs373667288 Chr.2: 85363253-85363253 on Build GRCh38
rs116769170 Chr.2: 85363384-85363384 on Build GRCh38
rs752839595 Chr.2: 85363090-85363090 on Build GRCh38
rs1041129101 Chr.2: 85363099-85363099 on Build GRCh38
rs956771408 Chr.2: 85363286-85363286 on Build GRCh38
rs771673993 Chr.2: 85363161-85363161 on Build GRCh38
rs947848781 Chr.2: 85363284-85363284 on Build GRCh38
rs774055360 Chr.2: 85363068-85363068 on Build GRCh38
rs544967777 Chr.2: 85363036-85363036 on Build GRCh38
rs768302094 Chr.2: 85363065-85363065 on Build GRCh38
rs185158687 Chr.2: 85363076-85363076 on Build GRCh38
rs762289558 Chr.2: 85363396-85363396 on Build GRCh38
rs557940265 Chr.2: 85363270-85363270 on Build GRCh38
rs768426016 Chr.2: 85363118-85363128 on Build GRCh38

View Full Product Details