Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 100417389-100417934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST10
Entrez Gene ID
9486
Gene Name
carbohydrate sulfotransferase 10
Gene Aliases
HNK-1ST, HNK1ST
UniGene
Hs.516370
Gene Chromosome Location
Chr.2: 100391860-100417668 on Build GRCh38
Transcripts
NM_004854.4 XM_011512210.1 XM_017005380.1 XM_017005381.1

SNP Detail

SNP ID Chromosome Location
rs867514787 Chr.2: 100417731-100417731 on Build GRCh38
rs1024182483 Chr.2: 100417454-100417454 on Build GRCh38
rs535480727 Chr.2: 100417772-100417772 on Build GRCh38
rs1031529373 Chr.2: 100417606-100417606 on Build GRCh38
rs913029086 Chr.2: 100417529-100417529 on Build GRCh38
rs957266633 Chr.2: 100417607-100417607 on Build GRCh38
rs949365169 Chr.2: 100417838-100417838 on Build GRCh38
rs975841808 Chr.2: 100417757-100417757 on Build GRCh38
rs534581874 Chr.2: 100417890-100417890 on Build GRCh38
rs895160099 Chr.2: 100417589-100417589 on Build GRCh38
rs974840368 Chr.2: 100417451-100417451 on Build GRCh38
rs1055558120 Chr.2: 100417815-100417815 on Build GRCh38
rs998609423 Chr.2: 100417709-100417709 on Build GRCh38
rs910327047 Chr.2: 100417609-100417609 on Build GRCh38
rs371965198 Chr.2: 100417847-100417847 on Build GRCh38
rs977094155 Chr.2: 100417484-100417484 on Build GRCh38
rs1017253830 Chr.2: 100417722-100417722 on Build GRCh38
rs571481515 Chr.2: 100417811-100417811 on Build GRCh38
rs929527013 Chr.2: 100417549-100417550 on Build GRCh38
rs561548876 Chr.2: 100417875-100417876 on Build GRCh38
rs935653048 Chr.2: 100417530-100417530 on Build GRCh38
rs952131404 Chr.2: 100417417-100417417 on Build GRCh38
rs983584209 Chr.2: 100417787-100417787 on Build GRCh38
rs921636861 Chr.2: 100417452-100417452 on Build GRCh38
rs773227664 Chr.2: 100417831-100417831 on Build GRCh38
rs909486465 Chr.2: 100417799-100417799 on Build GRCh38
rs994939372 Chr.2: 100417414-100417414 on Build GRCh38
rs950868399 Chr.2: 100417424-100417424 on Build GRCh38
rs933205095 Chr.2: 100417678-100417689 on Build GRCh38
rs983662874 Chr.2: 100417438-100417438 on Build GRCh38
rs1019651664 Chr.2: 100417441-100417441 on Build GRCh38
rs549947145 Chr.2: 100417720-100417720 on Build GRCh38
rs943132183 Chr.2: 100417617-100417617 on Build GRCh38
rs547984486 Chr.2: 100417595-100417595 on Build GRCh38
rs539520578 Chr.2: 100417719-100417719 on Build GRCh38
rs1031560489 Chr.2: 100417714-100417714 on Build GRCh38
rs942031512 Chr.2: 100417646-100417646 on Build GRCh38
rs942292303 Chr.2: 100417804-100417804 on Build GRCh38
rs1055511275 Chr.2: 100417580-100417580 on Build GRCh38
rs907065498 Chr.2: 100417692-100417692 on Build GRCh38
rs1055123011 Chr.2: 100417667-100417667 on Build GRCh38
rs902136739 Chr.2: 100417543-100417543 on Build GRCh38
rs971278907 Chr.2: 100417472-100417472 on Build GRCh38
rs965730481 Chr.2: 100417445-100417445 on Build GRCh38
rs568442387 Chr.2: 100417573-100417573 on Build GRCh38
rs977322841 Chr.2: 100417604-100417604 on Build GRCh38
rs569755663 Chr.2: 100417736-100417736 on Build GRCh38
rs1024987652 Chr.2: 100417690-100417690 on Build GRCh38
rs531640980 Chr.2: 100417536-100417536 on Build GRCh38
rs910265425 Chr.2: 100417533-100417533 on Build GRCh38
rs13385800 Chr.2: 100417547-100417547 on Build GRCh38
rs989867304 Chr.2: 100417520-100417520 on Build GRCh38
rs887996555 Chr.2: 100417638-100417638 on Build GRCh38
rs866379853 Chr.2: 100417850-100417850 on Build GRCh38
rs372487048 Chr.2: 100417760-100417760 on Build GRCh38
rs966674137 Chr.2: 100417870-100417870 on Build GRCh38
rs368464512 Chr.2: 100417421-100417421 on Build GRCh38
rs1048026216 Chr.2: 100417635-100417635 on Build GRCh38
rs895259194 Chr.2: 100417670-100417670 on Build GRCh38
rs368848889 Chr.2: 100417710-100417710 on Build GRCh38
rs548161848 Chr.2: 100417540-100417540 on Build GRCh38
rs895195822 Chr.2: 100417562-100417562 on Build GRCh38
rs923420498 Chr.2: 100417633-100417633 on Build GRCh38
rs527949789 Chr.2: 100417584-100417584 on Build GRCh38
rs1034181694 Chr.2: 100417561-100417561 on Build GRCh38
rs902574695 Chr.2: 100417856-100417856 on Build GRCh38
rs964847696 Chr.2: 100417751-100417751 on Build GRCh38
rs958804523 Chr.2: 100417433-100417433 on Build GRCh38
rs935726052 Chr.2: 100417477-100417477 on Build GRCh38
rs1047788782 Chr.2: 100417551-100417551 on Build GRCh38
rs572328685 Chr.2: 100417863-100417863 on Build GRCh38
rs570915019 Chr.2: 100417618-100417618 on Build GRCh38
rs976272351 Chr.2: 100417631-100417631 on Build GRCh38
rs907142893 Chr.2: 100417556-100417556 on Build GRCh38
rs556052262 Chr.2: 100417532-100417532 on Build GRCh38

View Full Product Details