Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 134867231-134867731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACMSD
Entrez Gene ID
130013
Gene Name
aminocarboxymuconate semialdehyde decarboxylase
Gene Aliases
-
UniGene
Hs.655728
Gene Chromosome Location
Chr.2: 134838319-134902906 on Build GRCh38
Transcripts
NM_001307983.1 NM_138326.2 XM_005263586.4 XM_005263588.4 XM_005263589.4 XM_005263590.4 XM_011510592.2 XM_017003325.1 XM_017003326.1 NR_036549.1

Gene Symbol
CCNT2-AS1
Entrez Gene ID
100129961
Gene Name
CCNT2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.729028
Gene Chromosome Location
Chr.2: 134866633-134918606 on Build GRCh38
Transcripts
NM_001307983.1 NM_138326.2 XM_005263586.4 XM_005263588.4 XM_005263589.4 XM_005263590.4 XM_011510592.2 XM_017003325.1 XM_017003326.1 NR_036549.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6592196 c.561C>A p.Y187* Chr.2: 134867653-134867653 on Build GRCh38
3961156 c.490G>C p.A164P Chr.2: 134867582-134867582 on Build GRCh38
6328944 c.580+9C>G p.? Chr.2: 134867681-134867681 on Build GRCh38
3115926 c.534G>T p.M178I Chr.2: 134867626-134867626 on Build GRCh38
1006686 c.569C>A p.P190H Chr.2: 134867661-134867661 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148178804 Chr.2: 134867392-134867392 on Build GRCh38
rs985061520 Chr.2: 134867489-134867489 on Build GRCh38
rs562819995 Chr.2: 134867312-134867312 on Build GRCh38
rs942181924 Chr.2: 134867413-134867413 on Build GRCh38
rs1052224941 Chr.2: 134867456-134867456 on Build GRCh38
rs781162614 Chr.2: 134867611-134867611 on Build GRCh38
rs950920544 Chr.2: 134867478-134867478 on Build GRCh38
rs981363289 Chr.2: 134867691-134867691 on Build GRCh38
rs370807501 Chr.2: 134867639-134867639 on Build GRCh38
rs745554810 Chr.2: 134867653-134867653 on Build GRCh38
rs902511231 Chr.2: 134867474-134867474 on Build GRCh38
rs1002690109 Chr.2: 134867403-134867405 on Build GRCh38
rs141124785 Chr.2: 134867418-134867418 on Build GRCh38
rs896542497 Chr.2: 134867703-134867703 on Build GRCh38
rs757421443 Chr.2: 134867585-134867585 on Build GRCh38
rs561802632 Chr.2: 134867283-134867283 on Build GRCh38
rs1000053825 Chr.2: 134867484-134867484 on Build GRCh38
rs182351345 Chr.2: 134867552-134867552 on Build GRCh38
rs1047470309 Chr.2: 134867314-134867314 on Build GRCh38
rs533915923 Chr.2: 134867479-134867479 on Build GRCh38
rs200762839 Chr.2: 134867534-134867534 on Build GRCh38
rs561970448 Chr.2: 134867593-134867593 on Build GRCh38
rs1009507146 Chr.2: 134867328-134867328 on Build GRCh38
rs573008458 Chr.2: 134867310-134867310 on Build GRCh38
rs755993446 Chr.2: 134867621-134867621 on Build GRCh38
rs375120563 Chr.2: 134867537-134867537 on Build GRCh38
rs1032398179 Chr.2: 134867522-134867522 on Build GRCh38
rs557257551 Chr.2: 134867659-134867659 on Build GRCh38
rs769789046 Chr.2: 134867547-134867548 on Build GRCh38
rs769035826 Chr.2: 134867670-134867670 on Build GRCh38
rs575919995 Chr.2: 134867690-134867690 on Build GRCh38
rs576201626 Chr.2: 134867549-134867552 on Build GRCh38
rs997767418 Chr.2: 134867430-134867430 on Build GRCh38
rs550335704 Chr.2: 134867306-134867306 on Build GRCh38
rs528387824 Chr.2: 134867362-134867362 on Build GRCh38
rs777835200 Chr.2: 134867633-134867633 on Build GRCh38
rs566341974 Chr.2: 134867422-134867422 on Build GRCh38
rs761156777 Chr.2: 134867662-134867662 on Build GRCh38
rs969976018 Chr.2: 134867630-134867630 on Build GRCh38
rs909365141 Chr.2: 134867506-134867506 on Build GRCh38
rs143468422 Chr.2: 134867584-134867584 on Build GRCh38
rs779815058 Chr.2: 134867546-134867547 on Build GRCh38
rs777196076 Chr.2: 134867681-134867681 on Build GRCh38
rs764707620 Chr.2: 134867581-134867581 on Build GRCh38
rs532952025 Chr.2: 134867563-134867564 on Build GRCh38
rs200534044 Chr.2: 134867640-134867640 on Build GRCh38
rs144757487 Chr.2: 134867609-134867609 on Build GRCh38
rs748679357 Chr.2: 134867680-134867681 on Build GRCh38
rs765305986 Chr.2: 134867586-134867586 on Build GRCh38
rs199939588 Chr.2: 134867598-134867598 on Build GRCh38
rs1033801225 Chr.2: 134867412-134867412 on Build GRCh38
rs1043912841 Chr.2: 134867335-134867335 on Build GRCh38
rs549109257 Chr.2: 134867509-134867509 on Build GRCh38
rs748679977 Chr.2: 134867635-134867635 on Build GRCh38
rs768198259 Chr.2: 134867548-134867549 on Build GRCh38
rs764017526 Chr.2: 134867551-134867552 on Build GRCh38
rs531430230 Chr.2: 134867552-134867553 on Build GRCh38
rs138317668 Chr.2: 134867549-134867550 on Build GRCh38
rs112570620 Chr.2: 134867305-134867305 on Build GRCh38
rs546463997 Chr.2: 134867432-134867432 on Build GRCh38
rs765379959 Chr.2: 134867701-134867701 on Build GRCh38
rs747995640 Chr.2: 134867614-134867614 on Build GRCh38

View Full Product Details