Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 165676234-165676741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRNP3
Entrez Gene ID
80034
Gene Name
cysteine and serine rich nuclear protein 3
Gene Aliases
FAM130A2, PPP1R73, TAIP-2, TAIP2
UniGene
Hs.470479
Gene Chromosome Location
Chr.2: 165469647-165689407 on Build GRCh38
Transcripts
NM_001172173.1 NM_024969.3 XM_005246865.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6866719 c.428T>C p.V143A Chr.2: 165676331-165676331 on Build GRCh38
3895166 c.601G>A p.E201K Chr.2: 165676504-165676504 on Build GRCh38
4627934 c.535A>G p.T179A Chr.2: 165676438-165676438 on Build GRCh38
6636955 c.661G>T p.D221Y Chr.2: 165676564-165676564 on Build GRCh38
1400716 c.672G>A p.T224T Chr.2: 165676575-165676575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970410514 Chr.2: 165676494-165676494 on Build GRCh38
rs74334465 Chr.2: 165676447-165676447 on Build GRCh38
rs376160936 Chr.2: 165676613-165676613 on Build GRCh38
rs377058956 Chr.2: 165676384-165676384 on Build GRCh38
rs762300895 Chr.2: 165676647-165676647 on Build GRCh38
rs773157959 Chr.2: 165676569-165676569 on Build GRCh38
rs942290744 Chr.2: 165676638-165676638 on Build GRCh38
rs1050855769 Chr.2: 165676586-165676586 on Build GRCh38
rs754321365 Chr.2: 165676528-165676528 on Build GRCh38
rs374471764 Chr.2: 165676287-165676287 on Build GRCh38
rs369735548 Chr.2: 165676553-165676553 on Build GRCh38
rs559756288 Chr.2: 165676355-165676355 on Build GRCh38
rs766099004 Chr.2: 165676659-165676659 on Build GRCh38
rs3731763 Chr.2: 165676527-165676527 on Build GRCh38
rs957982832 Chr.2: 165676444-165676444 on Build GRCh38
rs767539233 Chr.2: 165676440-165676440 on Build GRCh38
rs953751859 Chr.2: 165676284-165676284 on Build GRCh38
rs758001851 Chr.2: 165676511-165676511 on Build GRCh38
rs754378547 Chr.2: 165676320-165676320 on Build GRCh38
rs761399960 Chr.2: 165676503-165676503 on Build GRCh38
rs747967435 Chr.2: 165676405-165676405 on Build GRCh38
rs774477705 Chr.2: 165676271-165676271 on Build GRCh38
rs150163877 Chr.2: 165676688-165676688 on Build GRCh38
rs757771238 Chr.2: 165676323-165676323 on Build GRCh38
rs745421819 Chr.2: 165676359-165676359 on Build GRCh38
rs756526915 Chr.2: 165676302-165676302 on Build GRCh38
rs150240455 Chr.2: 165676462-165676462 on Build GRCh38
rs531777268 Chr.2: 165676573-165676573 on Build GRCh38
rs1016444638 Chr.2: 165676671-165676671 on Build GRCh38
rs79015458 Chr.2: 165676448-165676448 on Build GRCh38
rs148986083 Chr.2: 165676488-165676488 on Build GRCh38
rs746281067 Chr.2: 165676291-165676291 on Build GRCh38
rs764312290 Chr.2: 165676309-165676309 on Build GRCh38
rs757952662 Chr.2: 165676362-165676362 on Build GRCh38
rs779651317 Chr.2: 165676521-165676521 on Build GRCh38
rs769777169 Chr.2: 165676412-165676412 on Build GRCh38
rs563995173 Chr.2: 165676497-165676497 on Build GRCh38
rs763485784 Chr.2: 165676660-165676660 on Build GRCh38
rs147681972 Chr.2: 165676363-165676363 on Build GRCh38
rs200051425 Chr.2: 165676267-165676267 on Build GRCh38
rs371859963 Chr.2: 165676574-165676574 on Build GRCh38
rs746592873 Chr.2: 165676380-165676380 on Build GRCh38
rs762477811 Chr.2: 165676653-165676653 on Build GRCh38
rs528977899 Chr.2: 165676489-165676489 on Build GRCh38
rs766483883 Chr.2: 165676268-165676268 on Build GRCh38
rs371828567 Chr.2: 165676279-165676279 on Build GRCh38
rs983641375 Chr.2: 165676629-165676629 on Build GRCh38
rs760745996 Chr.2: 165676459-165676459 on Build GRCh38
rs972221135 Chr.2: 165676352-165676352 on Build GRCh38
rs867148664 Chr.2: 165676510-165676510 on Build GRCh38
rs1006644664 Chr.2: 165676639-165676639 on Build GRCh38
rs760796141 Chr.2: 165676622-165676622 on Build GRCh38
rs114829797 Chr.2: 165676633-165676633 on Build GRCh38
rs1016931781 Chr.2: 165676321-165676321 on Build GRCh38
rs747939892 Chr.2: 165676557-165676557 on Build GRCh38
rs768509627 Chr.2: 165676393-165676393 on Build GRCh38
rs764377242 Chr.2: 165676463-165676463 on Build GRCh38
rs184029617 Chr.2: 165676580-165676580 on Build GRCh38
rs775797316 Chr.2: 165676458-165676458 on Build GRCh38
rs776223100 Chr.2: 165676397-165676397 on Build GRCh38
rs759649991 Chr.2: 165676422-165676422 on Build GRCh38
rs999378375 Chr.2: 165676370-165676370 on Build GRCh38
rs371719922 Chr.2: 165676624-165676624 on Build GRCh38
rs918352749 Chr.2: 165676587-165676587 on Build GRCh38
rs563081862 Chr.2: 165676564-165676564 on Build GRCh38
rs768810699 Chr.2: 165676628-165676628 on Build GRCh38
rs79811472 Chr.2: 165676446-165676446 on Build GRCh38
rs754213592 Chr.2: 165676485-165676485 on Build GRCh38
rs773108700 Chr.2: 165676413-165676413 on Build GRCh38
rs754653152 Chr.2: 165676533-165676533 on Build GRCh38
rs140860699 Chr.2: 165676294-165676294 on Build GRCh38
rs772147944 Chr.2: 165676575-165676575 on Build GRCh38
rs78556225 Chr.2: 165676545-165676545 on Build GRCh38
rs765769025 Chr.2: 165676268-165676272 on Build GRCh38
rs980836334 Chr.2: 165676466-165676466 on Build GRCh38
rs962153626 Chr.2: 165676346-165676346 on Build GRCh38

View Full Product Details