Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 177436850-177437356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPS
Entrez Gene ID
8540
Gene Name
alkylglycerone phosphate synthase
Gene Aliases
ADAP-S, ADAS, ADHAPS, ADPS, ALDHPSY, RCDP3
UniGene
Hs.516543
Gene Chromosome Location
Chr.2: 177392743-177543836 on Build GRCh38
Transcripts
NM_003659.3 XM_011512041.2 XM_011512042.1 XM_011512043.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1244651 c.545G>A p.R182Q Chr.2: 177436867-177436867 on Build GRCh38
1009933 c.603G>A p.M201I Chr.2: 177437020-177437020 on Build GRCh38
3730855 c.613A>T p.I205F Chr.2: 177437030-177437030 on Build GRCh38
1182232 c.539A>T p.D180V Chr.2: 177436861-177436861 on Build GRCh38
3990771 c.611G>T p.R204L Chr.2: 177437028-177437028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888783611 Chr.2: 177437264-177437268 on Build GRCh38
rs369060661 Chr.2: 177436899-177436899 on Build GRCh38
rs201582276 Chr.2: 177437072-177437072 on Build GRCh38
rs745711263 Chr.2: 177436897-177436897 on Build GRCh38
rs776917165 Chr.2: 177436931-177436932 on Build GRCh38
rs1055037601 Chr.2: 177437303-177437303 on Build GRCh38
rs182602770 Chr.2: 177437067-177437067 on Build GRCh38
rs184809895 Chr.2: 177437256-177437256 on Build GRCh38
rs749441827 Chr.2: 177436969-177436969 on Build GRCh38
rs756070827 Chr.2: 177437105-177437109 on Build GRCh38
rs3834134 Chr.2: 177436969-177436969 on Build GRCh38
rs371316758 Chr.2: 177437089-177437089 on Build GRCh38
rs761362861 Chr.2: 177437069-177437069 on Build GRCh38
rs1010525464 Chr.2: 177436941-177436941 on Build GRCh38
rs897198612 Chr.2: 177436959-177436959 on Build GRCh38
rs947972014 Chr.2: 177437091-177437091 on Build GRCh38
rs972260230 Chr.2: 177437054-177437054 on Build GRCh38
rs1033626775 Chr.2: 177437074-177437074 on Build GRCh38
rs192600490 Chr.2: 177437160-177437160 on Build GRCh38
rs764909922 Chr.2: 177436930-177436930 on Build GRCh38
rs765518497 Chr.2: 177436953-177436953 on Build GRCh38
rs1007777098 Chr.2: 177437281-177437281 on Build GRCh38
rs188886766 Chr.2: 177437327-177437327 on Build GRCh38
rs763611185 Chr.2: 177436925-177436925 on Build GRCh38
rs527342449 Chr.2: 177437232-177437232 on Build GRCh38
rs372372429 Chr.2: 177436927-177436927 on Build GRCh38
rs562754046 Chr.2: 177436987-177436987 on Build GRCh38
rs998188081 Chr.2: 177436990-177436990 on Build GRCh38
rs760039846 Chr.2: 177436922-177436922 on Build GRCh38
rs981096203 Chr.2: 177437107-177437107 on Build GRCh38
rs368443476 Chr.2: 177436965-177436965 on Build GRCh38
rs772951018 Chr.2: 177437073-177437073 on Build GRCh38
rs200503696 Chr.2: 177437005-177437005 on Build GRCh38
rs927782939 Chr.2: 177437109-177437109 on Build GRCh38
rs1019607569 Chr.2: 177437285-177437285 on Build GRCh38
rs767870333 Chr.2: 177436933-177436933 on Build GRCh38
rs564811193 Chr.2: 177437211-177437211 on Build GRCh38
rs937949729 Chr.2: 177437299-177437299 on Build GRCh38
rs763109096 Chr.2: 177436958-177436959 on Build GRCh38
rs762250072 Chr.2: 177436936-177436942 on Build GRCh38
rs761454555 Chr.2: 177436928-177436928 on Build GRCh38
rs1021938690 Chr.2: 177436942-177436942 on Build GRCh38
rs909740048 Chr.2: 177437308-177437308 on Build GRCh38
rs753250521 Chr.2: 177436937-177436937 on Build GRCh38
rs771907891 Chr.2: 177437004-177437004 on Build GRCh38
rs746939822 Chr.2: 177437019-177437019 on Build GRCh38
rs560742425 Chr.2: 177436932-177436932 on Build GRCh38
rs868732459 Chr.2: 177437052-177437052 on Build GRCh38
rs939072710 Chr.2: 177436934-177436934 on Build GRCh38
rs757518400 Chr.2: 177436971-177436971 on Build GRCh38
rs745598152 Chr.2: 177436984-177436984 on Build GRCh38
rs993649986 Chr.2: 177437316-177437317 on Build GRCh38
rs3770002 Chr.2: 177437329-177437329 on Build GRCh38
rs140295002 Chr.2: 177437023-177437023 on Build GRCh38
rs774279058 Chr.2: 177436871-177436871 on Build GRCh38
rs768650581 Chr.2: 177437065-177437065 on Build GRCh38
rs981755614 Chr.2: 177437242-177437242 on Build GRCh38
rs374242073 Chr.2: 177436950-177436950 on Build GRCh38
rs771878828 Chr.2: 177436902-177436902 on Build GRCh38
rs892250295 Chr.2: 177436938-177436938 on Build GRCh38
rs1029633311 Chr.2: 177437041-177437041 on Build GRCh38
rs751625638 Chr.2: 177437081-177437081 on Build GRCh38
rs775517200 Chr.2: 177436920-177436920 on Build GRCh38
rs773452907 Chr.2: 177436959-177436959 on Build GRCh38
rs192263202 Chr.2: 177437328-177437328 on Build GRCh38
rs750572330 Chr.2: 177437011-177437011 on Build GRCh38
rs188926893 Chr.2: 177437126-177437126 on Build GRCh38

View Full Product Details