Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201192987-201193526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP10
Entrez Gene ID
843
Gene Name
caspase 10
Gene Aliases
ALPS2, FLICE2, MCH4
UniGene
Hs.5353
Gene Chromosome Location
Chr.2: 201182881-201229406 on Build GRCh38
Transcripts
NM_001206524.1 NM_001206542.1 NM_001230.4 NM_001306083.1 NM_032974.4 NM_032976.3 NM_032977.3 XM_005246907.2 XM_006712796.3 XM_011511990.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1242091 c.536C>T p.P179L Chr.2: 201193078-201193078 on Build GRCh38
4610063 c.555A>G p.I185M Chr.2: 201193097-201193097 on Build GRCh38
6620557 c.517C>A p.L173I Chr.2: 201193059-201193059 on Build GRCh38
6620558 c.517C>A p.L173I Chr.2: 201193059-201193059 on Build GRCh38
1242090 c.536C>T p.P179L Chr.2: 201193078-201193078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530025770 Chr.2: 201193300-201193300 on Build GRCh38
rs779426881 Chr.2: 201193149-201193149 on Build GRCh38
rs146233833 Chr.2: 201193076-201193076 on Build GRCh38
rs895671630 Chr.2: 201193086-201193086 on Build GRCh38
rs753049035 Chr.2: 201193096-201193096 on Build GRCh38
rs369231920 Chr.2: 201193095-201193095 on Build GRCh38
rs775033671 Chr.2: 201193293-201193293 on Build GRCh38
rs767659964 Chr.2: 201193072-201193072 on Build GRCh38
rs544551849 Chr.2: 201193211-201193211 on Build GRCh38
rs780947440 Chr.2: 201193122-201193122 on Build GRCh38
rs781000749 Chr.2: 201193020-201193020 on Build GRCh38
rs774005416 Chr.2: 201193049-201193049 on Build GRCh38
rs755432849 Chr.2: 201193091-201193091 on Build GRCh38
rs373276469 Chr.2: 201193126-201193126 on Build GRCh38
rs1038389995 Chr.2: 201193359-201193359 on Build GRCh38
rs773452930 Chr.2: 201193074-201193074 on Build GRCh38
rs761753815 Chr.2: 201193428-201193428 on Build GRCh38
rs894511746 Chr.2: 201193106-201193106 on Build GRCh38
rs922744800 Chr.2: 201193261-201193261 on Build GRCh38
rs200877813 Chr.2: 201193092-201193092 on Build GRCh38
rs768707944 Chr.2: 201193048-201193048 on Build GRCh38
rs751615960 Chr.2: 201193107-201193108 on Build GRCh38
rs1037621640 Chr.2: 201193013-201193013 on Build GRCh38
rs560207313 Chr.2: 201193364-201193364 on Build GRCh38
rs977830731 Chr.2: 201193240-201193240 on Build GRCh38
rs41349651 Chr.2: 201193243-201193243 on Build GRCh38
rs758869726 Chr.2: 201193097-201193097 on Build GRCh38
rs1050525585 Chr.2: 201193289-201193289 on Build GRCh38
rs371962581 Chr.2: 201193044-201193044 on Build GRCh38
rs745334080 Chr.2: 201193027-201193027 on Build GRCh38
rs185972225 Chr.2: 201193393-201193393 on Build GRCh38
rs1002152208 Chr.2: 201193414-201193414 on Build GRCh38
rs34562617 Chr.2: 201193497-201193498 on Build GRCh38
rs771684153 Chr.2: 201193258-201193258 on Build GRCh38
rs745495121 Chr.2: 201193139-201193139 on Build GRCh38
rs895629526 Chr.2: 201193398-201193398 on Build GRCh38
rs917234774 Chr.2: 201193198-201193199 on Build GRCh38
rs761610318 Chr.2: 201193071-201193071 on Build GRCh38
rs769455042 Chr.2: 201193029-201193029 on Build GRCh38
rs992951593 Chr.2: 201193155-201193155 on Build GRCh38
rs889310437 Chr.2: 201193326-201193326 on Build GRCh38
rs41387047 Chr.2: 201193382-201193382 on Build GRCh38
rs753989167 Chr.2: 201193090-201193090 on Build GRCh38
rs768940240 Chr.2: 201193170-201193170 on Build GRCh38
rs376619426 Chr.2: 201193148-201193148 on Build GRCh38
rs1027754461 Chr.2: 201193394-201193394 on Build GRCh38
rs546398529 Chr.2: 201193387-201193387 on Build GRCh38
rs766461320 Chr.2: 201193018-201193018 on Build GRCh38
rs550586227 Chr.2: 201193424-201193424 on Build GRCh38
rs371164247 Chr.2: 201193136-201193136 on Build GRCh38
rs748887487 Chr.2: 201193158-201193158 on Build GRCh38
rs946101309 Chr.2: 201193236-201193236 on Build GRCh38
rs538567770 Chr.2: 201193405-201193405 on Build GRCh38
rs960567584 Chr.2: 201193492-201193492 on Build GRCh38
rs964276431 Chr.2: 201193249-201193249 on Build GRCh38
rs548700616 Chr.2: 201193331-201193331 on Build GRCh38
rs941305021 Chr.2: 201193335-201193335 on Build GRCh38

View Full Product Details