Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202376207-202376715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMPR2
Entrez Gene ID
659
Gene Name
bone morphogenetic protein receptor type 2
Gene Aliases
BMPR-II, BMPR3, BMR2, BRK-3, POVD1, PPH1, T-ALK
UniGene
Hs.471119
Gene Chromosome Location
Chr.2: 202376310-202567751 on Build GRCh38
Transcripts
NM_001204.6 XM_011511687.1

SNP Detail

SNP ID Chromosome Location
rs532378406 Chr.2: 202376255-202376255 on Build GRCh38
rs878935632 Chr.2: 202376511-202376511 on Build GRCh38
rs1031907170 Chr.2: 202376462-202376462 on Build GRCh38
rs933858016 Chr.2: 202376245-202376245 on Build GRCh38
rs971534082 Chr.2: 202376415-202376415 on Build GRCh38
rs566733210 Chr.2: 202376607-202376607 on Build GRCh38
rs900999810 Chr.2: 202376231-202376231 on Build GRCh38
rs999451508 Chr.2: 202376457-202376457 on Build GRCh38
rs914912537 Chr.2: 202376506-202376506 on Build GRCh38
rs1012908604 Chr.2: 202376637-202376637 on Build GRCh38
rs1038301740 Chr.2: 202376570-202376570 on Build GRCh38
rs1038354587 Chr.2: 202376335-202376335 on Build GRCh38
rs1024375902 Chr.2: 202376390-202376390 on Build GRCh38
rs963755287 Chr.2: 202376509-202376510 on Build GRCh38
rs771255805 Chr.2: 202376542-202376543 on Build GRCh38
rs202027237 Chr.2: 202376299-202376299 on Build GRCh38
rs886055459 Chr.2: 202376545-202376547 on Build GRCh38
rs886055462 Chr.2: 202376553-202376553 on Build GRCh38
rs914689965 Chr.2: 202376691-202376691 on Build GRCh38
rs375624016 Chr.2: 202376547-202376548 on Build GRCh38
rs989136434 Chr.2: 202376498-202376498 on Build GRCh38
rs886055464 Chr.2: 202376638-202376638 on Build GRCh38
rs867772874 Chr.2: 202376520-202376520 on Build GRCh38
rs941279306 Chr.2: 202376565-202376566 on Build GRCh38
rs377400725 Chr.2: 202376476-202376476 on Build GRCh38
rs1053589959 Chr.2: 202376683-202376683 on Build GRCh38
rs548329280 Chr.2: 202376588-202376588 on Build GRCh38
rs550593985 Chr.2: 202376399-202376399 on Build GRCh38
rs957869805 Chr.2: 202376475-202376475 on Build GRCh38
rs1045772236 Chr.2: 202376663-202376663 on Build GRCh38
rs886055463 Chr.2: 202376604-202376604 on Build GRCh38
rs894526466 Chr.2: 202376352-202376352 on Build GRCh38
rs562633536 Chr.2: 202376542-202376542 on Build GRCh38
rs969724206 Chr.2: 202376342-202376342 on Build GRCh38
rs886055460 Chr.2: 202376556-202376557 on Build GRCh38
rs935220982 Chr.2: 202376572-202376572 on Build GRCh38
rs3044175 Chr.2: 202376545-202376545 on Build GRCh38
rs922369632 Chr.2: 202376527-202376527 on Build GRCh38
rs1005524693 Chr.2: 202376298-202376298 on Build GRCh38
rs887053309 Chr.2: 202376292-202376292 on Build GRCh38
rs879010787 Chr.2: 202376512-202376517 on Build GRCh38
rs570604902 Chr.2: 202376511-202376512 on Build GRCh38
rs1012964111 Chr.2: 202376378-202376378 on Build GRCh38
rs1046957145 Chr.2: 202376285-202376285 on Build GRCh38
rs894470955 Chr.2: 202376618-202376618 on Build GRCh38
rs908434869 Chr.2: 202376559-202376559 on Build GRCh38
rs933827592 Chr.2: 202376531-202376548 on Build GRCh38
rs1039517305 Chr.2: 202376229-202376229 on Build GRCh38
rs568136148 Chr.2: 202376545-202376546 on Build GRCh38
rs530181389 Chr.2: 202376548-202376548 on Build GRCh38
rs886055458 Chr.2: 202376361-202376361 on Build GRCh38
rs886055461 Chr.2: 202376548-202376551 on Build GRCh38
rs565254847 Chr.2: 202376234-202376234 on Build GRCh38

View Full Product Details