Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 210577343-210577842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPS1
Entrez Gene ID
1373
Gene Name
carbamoyl-phosphate synthase 1
Gene Aliases
CPSASE1, PHN
UniGene
Hs.149252
Gene Chromosome Location
Chr.2: 210477682-210679107 on Build GRCh38
Transcripts
NM_001122633.2 NM_001875.4 XM_011510640.1 XM_011510641.2 XM_011510642.2 XM_011510643.2 XM_011510644.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7362522 c.449G>A p.G150E Chr.2: 210577488-210577488 on Build GRCh38
5973577 c.468G>T p.G156G Chr.2: 210577489-210577489 on Build GRCh38
356480 c.454T>C p.W152R Chr.2: 210577493-210577493 on Build GRCh38
402161 c.427T>A p.W143R Chr.2: 210577466-210577466 on Build GRCh38
4090773 c.418C>T p.L140L Chr.2: 210577439-210577439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899304335 Chr.2: 210577570-210577570 on Build GRCh38
rs980324418 Chr.2: 210577773-210577773 on Build GRCh38
rs773239723 Chr.2: 210577471-210577471 on Build GRCh38
rs114819130 Chr.2: 210577488-210577488 on Build GRCh38
rs779623064 Chr.2: 210577427-210577427 on Build GRCh38
rs966206814 Chr.2: 210577701-210577701 on Build GRCh38
rs928463942 Chr.2: 210577800-210577800 on Build GRCh38
rs542855604 Chr.2: 210577628-210577628 on Build GRCh38
rs774291313 Chr.2: 210577508-210577508 on Build GRCh38
rs1038158239 Chr.2: 210577803-210577803 on Build GRCh38
rs371639705 Chr.2: 210577420-210577420 on Build GRCh38
rs539204261 Chr.2: 210577765-210577765 on Build GRCh38
rs373095049 Chr.2: 210577391-210577391 on Build GRCh38
rs938645149 Chr.2: 210577411-210577411 on Build GRCh38
rs757272145 Chr.2: 210577405-210577405 on Build GRCh38
rs1026945138 Chr.2: 210577593-210577593 on Build GRCh38
rs778646781 Chr.2: 210577448-210577448 on Build GRCh38
rs559238210 Chr.2: 210577635-210577635 on Build GRCh38
rs910289060 Chr.2: 210577384-210577384 on Build GRCh38
rs771977949 Chr.2: 210577560-210577560 on Build GRCh38
rs767584857 Chr.2: 210577531-210577531 on Build GRCh38
rs773663393 Chr.2: 210577481-210577481 on Build GRCh38
rs934592598 Chr.2: 210577468-210577468 on Build GRCh38
rs114245218 Chr.2: 210577751-210577751 on Build GRCh38
rs985920134 Chr.2: 210577381-210577381 on Build GRCh38
rs772621090 Chr.2: 210577465-210577465 on Build GRCh38
rs541134224 Chr.2: 210577723-210577723 on Build GRCh38
rs771266355 Chr.2: 210577489-210577489 on Build GRCh38
rs747085519 Chr.2: 210577475-210577475 on Build GRCh38
rs141050942 Chr.2: 210577510-210577510 on Build GRCh38
rs1037456021 Chr.2: 210577568-210577568 on Build GRCh38
rs759800691 Chr.2: 210577599-210577599 on Build GRCh38
rs955666086 Chr.2: 210577735-210577735 on Build GRCh38
rs914408029 Chr.2: 210577524-210577524 on Build GRCh38
rs995762573 Chr.2: 210577595-210577595 on Build GRCh38
rs755620752 Chr.2: 210577543-210577543 on Build GRCh38
rs889995055 Chr.2: 210577603-210577603 on Build GRCh38
rs772576138 Chr.2: 210577459-210577459 on Build GRCh38
rs755742834 Chr.2: 210577426-210577426 on Build GRCh38
rs748599785 Chr.2: 210577456-210577456 on Build GRCh38
rs779380891 Chr.2: 210577447-210577447 on Build GRCh38
rs557749812 Chr.2: 210577395-210577395 on Build GRCh38
rs759865421 Chr.2: 210577547-210577547 on Build GRCh38
rs1017377777 Chr.2: 210577696-210577696 on Build GRCh38
rs528932134 Chr.2: 210577594-210577594 on Build GRCh38
rs577598098 Chr.2: 210577429-210577429 on Build GRCh38
rs1037853254 Chr.2: 210577516-210577516 on Build GRCh38
rs1027240173 Chr.2: 210577596-210577596 on Build GRCh38
rs907265631 Chr.2: 210577550-210577550 on Build GRCh38
rs564589194 Chr.2: 210577737-210577737 on Build GRCh38
rs938717648 Chr.2: 210577566-210577566 on Build GRCh38
rs749925871 Chr.2: 210577533-210577533 on Build GRCh38
rs768648800 Chr.2: 210577441-210577441 on Build GRCh38
rs367807258 Chr.2: 210577415-210577415 on Build GRCh38
rs899253030 Chr.2: 210577814-210577815 on Build GRCh38
rs914479988 Chr.2: 210577758-210577758 on Build GRCh38
rs776843683 Chr.2: 210577530-210577530 on Build GRCh38
rs879087250 Chr.2: 210577556-210577556 on Build GRCh38
rs764002191 Chr.2: 210577485-210577486 on Build GRCh38
rs370155547 Chr.2: 210577392-210577392 on Build GRCh38
rs752873202 Chr.2: 210577606-210577606 on Build GRCh38
rs989613383 Chr.2: 210577652-210577652 on Build GRCh38
rs948584992 Chr.2: 210577764-210577764 on Build GRCh38
rs938776990 Chr.2: 210577812-210577812 on Build GRCh38
rs762692218 Chr.2: 210577382-210577382 on Build GRCh38
rs927883083 Chr.2: 210577739-210577739 on Build GRCh38
rs527810760 Chr.2: 210577717-210577717 on Build GRCh38
rs976136194 Chr.2: 210577417-210577417 on Build GRCh38
rs78365831 Chr.2: 210577582-210577582 on Build GRCh38
rs374439479 Chr.2: 210577454-210577454 on Build GRCh38
rs1022056464 Chr.2: 210577698-210577698 on Build GRCh38

View Full Product Details