Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219212929-219213434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB6
Entrez Gene ID
10058
Gene Name
ATP binding cassette subfamily B member 6 (Langereis blood group)
Gene Aliases
ABC, ABC14, DUH3, LAN, MCOPCB7, MTABC3, PRP, PSHK2, umat
UniGene
Hs.107911
Gene Chromosome Location
Chr.2: 219209766-219218990 on Build GRCh38
Transcripts
NM_005689.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1016611 c.1824C>T p.D608D Chr.2: 219213047-219213047 on Build GRCh38
719916 c.1782C>T p.N594N Chr.2: 219213264-219213264 on Build GRCh38
3426098 c.1783G>A p.V595M Chr.2: 219213263-219213263 on Build GRCh38
301424 c.1748_1749TT>CC p.L583>? Chr.2: 219213297-219213298 on Build GRCh38
3044586 c.1751G>A p.R584H Chr.2: 219213295-219213295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774856775 Chr.2: 219213320-219213320 on Build GRCh38
rs397514758 Chr.2: 219213310-219213310 on Build GRCh38
rs756405506 Chr.2: 219213354-219213354 on Build GRCh38
rs1026355294 Chr.2: 219213262-219213262 on Build GRCh38
rs746605817 Chr.2: 219213224-219213225 on Build GRCh38
rs753330867 Chr.2: 219213058-219213058 on Build GRCh38
rs374541848 Chr.2: 219213046-219213046 on Build GRCh38
rs201104334 Chr.2: 219213371-219213371 on Build GRCh38
rs770668401 Chr.2: 219213312-219213312 on Build GRCh38
rs760435750 Chr.2: 219213213-219213213 on Build GRCh38
rs764674184 Chr.2: 219213245-219213245 on Build GRCh38
rs777669550 Chr.2: 219213301-219213301 on Build GRCh38
rs750467585 Chr.2: 219213147-219213147 on Build GRCh38
rs749302568 Chr.2: 219213022-219213022 on Build GRCh38
rs75164102 Chr.2: 219213202-219213202 on Build GRCh38
rs550424352 Chr.2: 219213303-219213303 on Build GRCh38
rs759073924 Chr.2: 219213243-219213243 on Build GRCh38
rs764585714 Chr.2: 219213256-219213256 on Build GRCh38
rs187275052 Chr.2: 219213207-219213207 on Build GRCh38
rs770875456 Chr.2: 219212958-219212958 on Build GRCh38
rs199746430 Chr.2: 219213399-219213399 on Build GRCh38
rs374648956 Chr.2: 219213234-219213234 on Build GRCh38
rs200153096 Chr.2: 219213250-219213250 on Build GRCh38
rs771909792 Chr.2: 219213208-219213208 on Build GRCh38
rs868199730 Chr.2: 219213240-219213240 on Build GRCh38
rs754686262 Chr.2: 219213225-219213225 on Build GRCh38
rs769332783 Chr.2: 219213344-219213344 on Build GRCh38
rs201931275 Chr.2: 219213336-219213336 on Build GRCh38
rs368167926 Chr.2: 219213264-219213264 on Build GRCh38
rs750626192 Chr.2: 219213288-219213288 on Build GRCh38
rs771893220 Chr.2: 219213299-219213299 on Build GRCh38
rs923617817 Chr.2: 219213398-219213398 on Build GRCh38
rs767172815 Chr.2: 219213041-219213041 on Build GRCh38
rs754351332 Chr.2: 219212996-219212996 on Build GRCh38
rs761698895 Chr.2: 219213038-219213038 on Build GRCh38
rs769298194 Chr.2: 219213318-219213318 on Build GRCh38
rs937815794 Chr.2: 219213094-219213094 on Build GRCh38
rs779733140 Chr.2: 219213223-219213223 on Build GRCh38
rs768694750 Chr.2: 219213025-219213025 on Build GRCh38
rs768364665 Chr.2: 219213281-219213281 on Build GRCh38
rs758812992 Chr.2: 219213071-219213071 on Build GRCh38
rs745913706 Chr.2: 219213104-219213104 on Build GRCh38
rs747150898 Chr.2: 219213012-219213012 on Build GRCh38
rs370348853 Chr.2: 219213194-219213194 on Build GRCh38
rs781472859 Chr.2: 219213092-219213092 on Build GRCh38
rs756279217 Chr.2: 219213294-219213294 on Build GRCh38
rs780847738 Chr.2: 219213240-219213240 on Build GRCh38
rs966358048 Chr.2: 219213184-219213184 on Build GRCh38
rs902185312 Chr.2: 219213186-219213186 on Build GRCh38
rs752837301 Chr.2: 219212980-219212980 on Build GRCh38
rs756642643 Chr.2: 219212981-219212981 on Build GRCh38
rs544994317 Chr.2: 219213131-219213131 on Build GRCh38
rs770723235 Chr.2: 219213406-219213406 on Build GRCh38
rs530259488 Chr.2: 219213017-219213017 on Build GRCh38
rs748036945 Chr.2: 219213266-219213267 on Build GRCh38
rs975345031 Chr.2: 219213219-219213219 on Build GRCh38
rs9288542 Chr.2: 219213341-219213341 on Build GRCh38
rs777941316 Chr.2: 219213083-219213083 on Build GRCh38
rs765698382 Chr.2: 219213063-219213063 on Build GRCh38
rs370710985 Chr.2: 219213330-219213330 on Build GRCh38
rs772881476 Chr.2: 219213212-219213212 on Build GRCh38
rs79512794 Chr.2: 219213036-219213036 on Build GRCh38
rs757676835 Chr.2: 219213163-219213163 on Build GRCh38
rs950736040 Chr.2: 219213270-219213270 on Build GRCh38
rs761423202 Chr.2: 219212976-219212976 on Build GRCh38
rs748186310 Chr.2: 219213296-219213296 on Build GRCh38
rs371348525 Chr.2: 219213317-219213317 on Build GRCh38
rs778457867 Chr.2: 219213064-219213064 on Build GRCh38
rs145526996 Chr.2: 219213284-219213284 on Build GRCh38
rs757414837 Chr.2: 219213090-219213090 on Build GRCh38
rs1020831399 Chr.2: 219213403-219213403 on Build GRCh38
rs185646565 Chr.2: 219213334-219213334 on Build GRCh38
rs755475002 Chr.2: 219212997-219212997 on Build GRCh38
rs755525579 Chr.2: 219213061-219213061 on Build GRCh38
rs1018075482 Chr.2: 219213002-219213002 on Build GRCh38
rs145498806 Chr.2: 219213295-219213295 on Build GRCh38
rs762924312 Chr.2: 219212967-219212967 on Build GRCh38
rs867654978 Chr.2: 219213013-219213013 on Build GRCh38
rs765821064 Chr.2: 219212982-219212982 on Build GRCh38
rs1020612260 Chr.2: 219213182-219213182 on Build GRCh38
rs763571986 Chr.2: 219213326-219213326 on Build GRCh38
rs190137939 Chr.2: 219213263-219213263 on Build GRCh38
rs775589869 Chr.2: 219212963-219212963 on Build GRCh38
rs778083436 Chr.2: 219213010-219213010 on Build GRCh38
rs769955438 Chr.2: 219213191-219213191 on Build GRCh38
rs757942218 Chr.2: 219213049-219213064 on Build GRCh38
rs765991143 Chr.2: 219213225-219213225 on Build GRCh38
rs376961879 Chr.2: 219213059-219213059 on Build GRCh38
rs747258692 Chr.2: 219213084-219213084 on Build GRCh38
rs779305304 Chr.2: 219212998-219212998 on Build GRCh38
rs483352876 Chr.2: 219213280-219213280 on Build GRCh38
rs558725837 Chr.2: 219213246-219213246 on Build GRCh38
rs893583192 Chr.2: 219213121-219213121 on Build GRCh38
rs370947094 Chr.2: 219213000-219213000 on Build GRCh38
rs746895150 Chr.2: 219213401-219213401 on Build GRCh38
rs763984294 Chr.2: 219212968-219212968 on Build GRCh38
rs556020297 Chr.2: 219212972-219212972 on Build GRCh38
rs986239039 Chr.2: 219212977-219212977 on Build GRCh38
rs982057321 Chr.2: 219213276-219213276 on Build GRCh38
rs771055662 Chr.2: 219213015-219213015 on Build GRCh38
rs200205391 Chr.2: 219213396-219213396 on Build GRCh38
rs750681683 Chr.2: 219213350-219213350 on Build GRCh38
rs758584770 Chr.2: 219213005-219213005 on Build GRCh38

View Full Product Details