Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232482762-232483260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECEL1
Entrez Gene ID
9427
Gene Name
endothelin converting enzyme like 1
Gene Aliases
DA5D, DINE, ECEX, XCE
UniGene
Hs.26880
Gene Chromosome Location
Chr.2: 232479827-232487859 on Build GRCh38
Transcripts
NM_001290787.1 NM_004826.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
442482 c.1579G>C p.E527Q Chr.2: 232483107-232483107 on Build GRCh38
4092395 c.1558G>A p.D520N Chr.2: 232483128-232483128 on Build GRCh38
1017820 c.1562C>A p.A521D Chr.2: 232483124-232483124 on Build GRCh38
4433878 c.1631G>A p.R544H Chr.2: 232482905-232482905 on Build GRCh38
5883407 c.1552A>T p.K518* Chr.2: 232483134-232483134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17293690 Chr.2: 232482890-232482890 on Build GRCh38
rs771755274 Chr.2: 232483085-232483085 on Build GRCh38
rs768350456 Chr.2: 232482965-232482965 on Build GRCh38
rs1039557294 Chr.2: 232483233-232483233 on Build GRCh38
rs971628494 Chr.2: 232482862-232482862 on Build GRCh38
rs199916648 Chr.2: 232483147-232483147 on Build GRCh38
rs781610712 Chr.2: 232482920-232482920 on Build GRCh38
rs374850133 Chr.2: 232483175-232483175 on Build GRCh38
rs771842952 Chr.2: 232483150-232483150 on Build GRCh38
rs755908082 Chr.2: 232482905-232482905 on Build GRCh38
rs768420513 Chr.2: 232482876-232482878 on Build GRCh38
rs570941848 Chr.2: 232483011-232483011 on Build GRCh38
rs146779776 Chr.2: 232482904-232482904 on Build GRCh38
rs749652211 Chr.2: 232483076-232483076 on Build GRCh38
rs773792839 Chr.2: 232482967-232482967 on Build GRCh38
rs778563534 Chr.2: 232483083-232483083 on Build GRCh38
rs6750085 Chr.2: 232483209-232483209 on Build GRCh38
rs775813200 Chr.2: 232482935-232482935 on Build GRCh38
rs753094153 Chr.2: 232483222-232483222 on Build GRCh38
rs758614813 Chr.2: 232482913-232482913 on Build GRCh38
rs767625283 Chr.2: 232483128-232483128 on Build GRCh38
rs758359848 Chr.2: 232482815-232482815 on Build GRCh38
rs776590182 Chr.2: 232483174-232483174 on Build GRCh38
rs368803327 Chr.2: 232482994-232482994 on Build GRCh38
rs146083116 Chr.2: 232482889-232482889 on Build GRCh38
rs768207898 Chr.2: 232483081-232483081 on Build GRCh38
rs767550105 Chr.2: 232482902-232482902 on Build GRCh38
rs375349961 Chr.2: 232483217-232483217 on Build GRCh38
rs777886260 Chr.2: 232482817-232482817 on Build GRCh38
rs770211718 Chr.2: 232482934-232482934 on Build GRCh38
rs773233774 Chr.2: 232482975-232482975 on Build GRCh38
rs773885419 Chr.2: 232482883-232482883 on Build GRCh38
rs902560306 Chr.2: 232483229-232483229 on Build GRCh38
rs773111477 Chr.2: 232482843-232482843 on Build GRCh38
rs202085552 Chr.2: 232483210-232483210 on Build GRCh38
rs755331817 Chr.2: 232483145-232483145 on Build GRCh38
rs369526951 Chr.2: 232483140-232483140 on Build GRCh38
rs560560183 Chr.2: 232483233-232483234 on Build GRCh38
rs13002919 Chr.2: 232483056-232483056 on Build GRCh38
rs587777130 Chr.2: 232482887-232482887 on Build GRCh38
rs757715782 Chr.2: 232482917-232482917 on Build GRCh38
rs756676913 Chr.2: 232483132-232483132 on Build GRCh38
rs746811400 Chr.2: 232482828-232482838 on Build GRCh38
rs372045195 Chr.2: 232483220-232483220 on Build GRCh38
rs747510730 Chr.2: 232482826-232482826 on Build GRCh38
rs377348971 Chr.2: 232483202-232483202 on Build GRCh38
rs753016147 Chr.2: 232482906-232482906 on Build GRCh38
rs576879477 Chr.2: 232483214-232483214 on Build GRCh38
rs550130776 Chr.2: 232483119-232483119 on Build GRCh38
rs200937309 Chr.2: 232483218-232483218 on Build GRCh38
rs368587591 Chr.2: 232482851-232482851 on Build GRCh38
rs376563571 Chr.2: 232482801-232482801 on Build GRCh38
rs376977206 Chr.2: 232483084-232483084 on Build GRCh38
rs944613733 Chr.2: 232482867-232482867 on Build GRCh38
rs771460490 Chr.2: 232482838-232482838 on Build GRCh38
rs1190429 Chr.2: 232482880-232482880 on Build GRCh38
rs760665185 Chr.2: 232482981-232482981 on Build GRCh38
rs746089635 Chr.2: 232482932-232482932 on Build GRCh38
rs757484998 Chr.2: 232483045-232483045 on Build GRCh38
rs760757121 Chr.2: 232483092-232483092 on Build GRCh38
rs759307695 Chr.2: 232483100-232483100 on Build GRCh38
rs576364780 Chr.2: 232483226-232483226 on Build GRCh38
rs754160637 Chr.2: 232483144-232483144 on Build GRCh38
rs113658806 Chr.2: 232483068-232483068 on Build GRCh38
rs747871273 Chr.2: 232483148-232483148 on Build GRCh38
rs182355168 Chr.2: 232483156-232483156 on Build GRCh38
rs542185300 Chr.2: 232483239-232483239 on Build GRCh38
rs547898666 Chr.2: 232482982-232482982 on Build GRCh38
rs770934372 Chr.2: 232482798-232482798 on Build GRCh38
rs202046121 Chr.2: 232482885-232482885 on Build GRCh38
rs750498508 Chr.2: 232483129-232483129 on Build GRCh38
rs775086793 Chr.2: 232482873-232482873 on Build GRCh38
rs374002082 Chr.2: 232483071-232483071 on Build GRCh38
rs797045548 Chr.2: 232483188-232483188 on Build GRCh38
rs772816522 Chr.2: 232483091-232483091 on Build GRCh38
rs1000027670 Chr.2: 232483232-232483232 on Build GRCh38
rs770098089 Chr.2: 232482927-232482927 on Build GRCh38
rs142971707 Chr.2: 232483170-232483170 on Build GRCh38
rs368556568 Chr.2: 232483239-232483240 on Build GRCh38
rs374586939 Chr.2: 232483028-232483028 on Build GRCh38
rs965875713 Chr.2: 232482863-232482863 on Build GRCh38
rs776303095 Chr.2: 232483099-232483099 on Build GRCh38
rs747095205 Chr.2: 232482916-232482916 on Build GRCh38
rs562073116 Chr.2: 232482854-232482854 on Build GRCh38
rs762550443 Chr.2: 232482877-232482877 on Build GRCh38
rs746461050 Chr.2: 232483160-232483160 on Build GRCh38
rs763138696 Chr.2: 232482990-232482990 on Build GRCh38
rs976845737 Chr.2: 232483094-232483094 on Build GRCh38
rs757285509 Chr.2: 232483060-232483060 on Build GRCh38
rs569970209 Chr.2: 232483127-232483127 on Build GRCh38
rs375170118 Chr.2: 232483109-232483109 on Build GRCh38
rs761723743 Chr.2: 232482901-232482901 on Build GRCh38
rs762090965 Chr.2: 232483189-232483189 on Build GRCh38
rs372342600 Chr.2: 232482964-232482964 on Build GRCh38
rs781682011 Chr.2: 232483067-232483067 on Build GRCh38
rs587777131 Chr.2: 232482850-232482850 on Build GRCh38
rs960913879 Chr.2: 232483046-232483046 on Build GRCh38
rs71398722 Chr.2: 232483232-232483233 on Build GRCh38
rs957732664 Chr.2: 232482816-232482816 on Build GRCh38
rs746265437 Chr.2: 232482921-232482921 on Build GRCh38
rs769439158 Chr.2: 232482872-232482872 on Build GRCh38
rs971576250 Chr.2: 232483178-232483179 on Build GRCh38
rs780468559 Chr.2: 232483072-232483072 on Build GRCh38
rs61731717 Chr.2: 232482871-232482871 on Build GRCh38
rs748267683 Chr.2: 232482802-232482802 on Build GRCh38
rs753663032 Chr.2: 232482988-232482988 on Build GRCh38
rs770974424 Chr.2: 232483097-232483097 on Build GRCh38
rs764253018 Chr.2: 232483122-232483122 on Build GRCh38
rs761330693 Chr.2: 232482968-232482968 on Build GRCh38
rs34056187 Chr.2: 232482852-232482853 on Build GRCh38
rs374731576 Chr.2: 232482971-232482971 on Build GRCh38
rs1010356091 Chr.2: 232483117-232483117 on Build GRCh38
rs1047887566 Chr.2: 232482831-232482831 on Build GRCh38
rs751758652 Chr.2: 232483124-232483124 on Build GRCh38
rs890986775 Chr.2: 232483010-232483010 on Build GRCh38
rs568689441 Chr.2: 232483183-232483183 on Build GRCh38

View Full Product Details