Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240482103-240482503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKMY1
Entrez Gene ID
51281
Gene Name
ankyrin repeat and MYND domain containing 1
Gene Aliases
ZMYND13
UniGene
Hs.656615
Gene Chromosome Location
Chr.2: 240468632-240561110 on Build GRCh38
Transcripts
NM_001282771.1 NM_001282780.1 NM_001282781.1 NM_001308375.1 NM_016552.3 NM_017844.3 XM_011511301.1 XM_011511317.1 XM_011511318.1 XM_011511322.1 XM_017004271.1 XM_017004272.1 XM_017004273.1 XM_017004274.1 XM_017004275.1 XM_017004276.1 XM_017004277.1 XM_017004278.1 XM_017004279.1 XM_017004280.1 XM_017004281.1 XM_017004282.1 XM_017004283.1 XM_017004284.1 XM_017004285.1 XM_017004286.1 XM_017004288.1 XM_017004290.1 XM_017004291.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3909981 c.1881C>T p.P627P Chr.2: 240482248-240482248 on Build GRCh38
7355549 c.1910G>A p.S637N Chr.2: 240482219-240482219 on Build GRCh38
6415138 c.1901A>T p.K634M Chr.2: 240482228-240482228 on Build GRCh38
3909980 c.2553C>T p.P851P Chr.2: 240482248-240482248 on Build GRCh38
7202135 c.2600A>T p.D867V Chr.2: 240482201-240482201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905941059 Chr.2: 240482392-240482392 on Build GRCh38
rs10804408 Chr.2: 240482429-240482429 on Build GRCh38
rs541201575 Chr.2: 240482422-240482422 on Build GRCh38
rs747196781 Chr.2: 240482227-240482229 on Build GRCh38
rs76812266 Chr.2: 240482473-240482473 on Build GRCh38
rs781480258 Chr.2: 240482246-240482247 on Build GRCh38
rs777298013 Chr.2: 240482261-240482261 on Build GRCh38
rs950950161 Chr.2: 240482413-240482413 on Build GRCh38
rs1036269051 Chr.2: 240482397-240482397 on Build GRCh38
rs548508562 Chr.2: 240482316-240482316 on Build GRCh38
rs1026209133 Chr.2: 240482435-240482435 on Build GRCh38
rs773939917 Chr.2: 240482251-240482251 on Build GRCh38
rs980618439 Chr.2: 240482133-240482134 on Build GRCh38
rs943145664 Chr.2: 240482287-240482287 on Build GRCh38
rs76082075 Chr.2: 240482191-240482191 on Build GRCh38
rs755362771 Chr.2: 240482233-240482233 on Build GRCh38
rs970572811 Chr.2: 240482127-240482127 on Build GRCh38
rs1011942821 Chr.2: 240482294-240482294 on Build GRCh38
rs113913655 Chr.2: 240482368-240482368 on Build GRCh38
rs141633070 Chr.2: 240482182-240482182 on Build GRCh38
rs758584713 Chr.2: 240482176-240482176 on Build GRCh38
rs745615959 Chr.2: 240482209-240482209 on Build GRCh38
rs1035299596 Chr.2: 240482133-240482133 on Build GRCh38
rs530769521 Chr.2: 240482349-240482349 on Build GRCh38
rs375234055 Chr.2: 240482260-240482260 on Build GRCh38
rs116348675 Chr.2: 240482239-240482239 on Build GRCh38
rs780291082 Chr.2: 240482133-240482133 on Build GRCh38
rs748966956 Chr.2: 240482237-240482237 on Build GRCh38
rs921878405 Chr.2: 240482212-240482212 on Build GRCh38
rs1023417065 Chr.2: 240482380-240482380 on Build GRCh38
rs753065902 Chr.2: 240482280-240482280 on Build GRCh38
rs921909745 Chr.2: 240482443-240482443 on Build GRCh38
rs775201277 Chr.2: 240482328-240482328 on Build GRCh38
rs765326129 Chr.2: 240482140-240482140 on Build GRCh38
rs569829146 Chr.2: 240482320-240482320 on Build GRCh38
rs763182014 Chr.2: 240482288-240482288 on Build GRCh38
rs909325262 Chr.2: 240482423-240482423 on Build GRCh38
rs974736139 Chr.2: 240482436-240482436 on Build GRCh38
rs769611422 Chr.2: 240482218-240482218 on Build GRCh38
rs540302878 Chr.2: 240482122-240482122 on Build GRCh38
rs368388634 Chr.2: 240482295-240482295 on Build GRCh38
rs751602664 Chr.2: 240482304-240482304 on Build GRCh38
rs147717110 Chr.2: 240482448-240482448 on Build GRCh38
rs367775435 Chr.2: 240482275-240482275 on Build GRCh38
rs570485915 Chr.2: 240482374-240482374 on Build GRCh38
rs923853708 Chr.2: 240482463-240482463 on Build GRCh38
rs781141352 Chr.2: 240482206-240482206 on Build GRCh38
rs764308828 Chr.2: 240482297-240482297 on Build GRCh38
rs1051202980 Chr.2: 240482252-240482252 on Build GRCh38
rs757123975 Chr.2: 240482199-240482199 on Build GRCh38
rs771745495 Chr.2: 240482281-240482281 on Build GRCh38
rs987310692 Chr.2: 240482457-240482457 on Build GRCh38
rs370499899 Chr.2: 240482138-240482138 on Build GRCh38
rs760007389 Chr.2: 240482270-240482270 on Build GRCh38
rs752939513 Chr.2: 240482146-240482146 on Build GRCh38
rs779754517 Chr.2: 240482230-240482230 on Build GRCh38

View Full Product Details