Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2795228-2795722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPXM1
Entrez Gene ID
56265
Gene Name
carboxypeptidase X (M14 family), member 1
Gene Aliases
CPX1, CPXM
UniGene
Hs.659346
Gene Chromosome Location
Chr.20: 2794069-2800646 on Build GRCh38
Transcripts
NM_001184699.1 NM_019609.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1632222 c.1837G>A p.A613T Chr.20: 2795300-2795300 on Build GRCh38
6302833 c.1704G>A p.W568* Chr.20: 2795615-2795615 on Build GRCh38
3972492 c.1797C>T p.H599H Chr.20: 2795340-2795340 on Build GRCh38
6298385 c.1749C>A p.T583T Chr.20: 2795388-2795388 on Build GRCh38
390815 c.1633G>T p.D545Y Chr.20: 2795686-2795686 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765326895 Chr.20: 2795570-2795570 on Build GRCh38
rs748946318 Chr.20: 2795466-2795466 on Build GRCh38
rs781722698 Chr.20: 2795454-2795454 on Build GRCh38
rs767317980 Chr.20: 2795565-2795565 on Build GRCh38
rs1027598497 Chr.20: 2795624-2795624 on Build GRCh38
rs892951831 Chr.20: 2795519-2795519 on Build GRCh38
rs770416813 Chr.20: 2795254-2795254 on Build GRCh38
rs147765286 Chr.20: 2795603-2795603 on Build GRCh38
rs377710258 Chr.20: 2795274-2795274 on Build GRCh38
rs762006674 Chr.20: 2795427-2795427 on Build GRCh38
rs1004487590 Chr.20: 2795260-2795260 on Build GRCh38
rs746438167 Chr.20: 2795565-2795565 on Build GRCh38
rs761705901 Chr.20: 2795556-2795556 on Build GRCh38
rs771466865 Chr.20: 2795340-2795341 on Build GRCh38
rs778953404 Chr.20: 2795421-2795421 on Build GRCh38
rs936559798 Chr.20: 2795698-2795698 on Build GRCh38
rs141526809 Chr.20: 2795677-2795677 on Build GRCh38
rs1056051386 Chr.20: 2795517-2795517 on Build GRCh38
rs374663889 Chr.20: 2795609-2795609 on Build GRCh38
rs774906068 Chr.20: 2795462-2795462 on Build GRCh38
rs191472527 Chr.20: 2795626-2795626 on Build GRCh38
rs1033135068 Chr.20: 2795564-2795565 on Build GRCh38
rs774231916 Chr.20: 2795555-2795555 on Build GRCh38
rs376638657 Chr.20: 2795459-2795459 on Build GRCh38
rs760577440 Chr.20: 2795568-2795568 on Build GRCh38
rs199564388 Chr.20: 2795271-2795271 on Build GRCh38
rs1053862808 Chr.20: 2795689-2795689 on Build GRCh38
rs182655680 Chr.20: 2795272-2795272 on Build GRCh38
rs115394567 Chr.20: 2795564-2795564 on Build GRCh38
rs767505445 Chr.20: 2795386-2795386 on Build GRCh38
rs1035982905 Chr.20: 2795496-2795496 on Build GRCh38
rs755097782 Chr.20: 2795283-2795283 on Build GRCh38
rs758656907 Chr.20: 2795429-2795429 on Build GRCh38
rs756605193 Chr.20: 2795627-2795627 on Build GRCh38
rs758603446 Chr.20: 2795589-2795589 on Build GRCh38
rs768482754 Chr.20: 2795551-2795551 on Build GRCh38
rs778104642 Chr.20: 2795433-2795433 on Build GRCh38
rs761830537 Chr.20: 2795363-2795363 on Build GRCh38
rs200029209 Chr.20: 2795648-2795648 on Build GRCh38
rs750813847 Chr.20: 2795684-2795684 on Build GRCh38
rs938078645 Chr.20: 2795516-2795516 on Build GRCh38
rs752893032 Chr.20: 2795422-2795422 on Build GRCh38
rs779200453 Chr.20: 2795292-2795292 on Build GRCh38
rs920083503 Chr.20: 2795385-2795385 on Build GRCh38
rs771491989 Chr.20: 2795307-2795307 on Build GRCh38
rs760122188 Chr.20: 2795252-2795252 on Build GRCh38
rs757667358 Chr.20: 2795606-2795606 on Build GRCh38
rs201970765 Chr.20: 2795270-2795270 on Build GRCh38
rs538584869 Chr.20: 2795647-2795647 on Build GRCh38
rs763022497 Chr.20: 2795672-2795672 on Build GRCh38
rs746357181 Chr.20: 2795612-2795612 on Build GRCh38
rs147866857 Chr.20: 2795642-2795642 on Build GRCh38
rs145736623 Chr.20: 2795602-2795602 on Build GRCh38
rs764257542 Chr.20: 2795673-2795673 on Build GRCh38
rs151104845 Chr.20: 2795261-2795261 on Build GRCh38
rs533953788 Chr.20: 2795505-2795505 on Build GRCh38
rs74866360 Chr.20: 2795273-2795273 on Build GRCh38
rs905042088 Chr.20: 2795534-2795534 on Build GRCh38
rs566174236 Chr.20: 2795481-2795481 on Build GRCh38
rs775487980 Chr.20: 2795662-2795662 on Build GRCh38
rs149777086 Chr.20: 2795641-2795641 on Build GRCh38
rs372739739 Chr.20: 2795256-2795256 on Build GRCh38
rs778615093 Chr.20: 2795692-2795692 on Build GRCh38
rs549143936 Chr.20: 2795530-2795530 on Build GRCh38
rs748821645 Chr.20: 2795628-2795628 on Build GRCh38
rs751469005 Chr.20: 2795349-2795349 on Build GRCh38
rs747428139 Chr.20: 2795451-2795451 on Build GRCh38
rs752389496 Chr.20: 2795263-2795263 on Build GRCh38
rs1001086455 Chr.20: 2795540-2795540 on Build GRCh38
rs748471153 Chr.20: 2795299-2795299 on Build GRCh38
rs769295245 Chr.20: 2795330-2795330 on Build GRCh38
rs908942159 Chr.20: 2795480-2795480 on Build GRCh38
rs772447877 Chr.20: 2795300-2795300 on Build GRCh38
rs1019250052 Chr.20: 2795586-2795586 on Build GRCh38
rs201609174 Chr.20: 2795633-2795633 on Build GRCh38
rs963797346 Chr.20: 2795324-2795324 on Build GRCh38
rs756588229 Chr.20: 2795687-2795687 on Build GRCh38
rs762842667 Chr.20: 2795265-2795265 on Build GRCh38
rs759082888 Chr.20: 2795257-2795257 on Build GRCh38
rs751762428 Chr.20: 2795674-2795674 on Build GRCh38
rs773697213 Chr.20: 2795572-2795572 on Build GRCh38
rs775234521 Chr.20: 2795340-2795340 on Build GRCh38
rs6115674 Chr.20: 2795590-2795590 on Build GRCh38
rs186626222 Chr.20: 2795301-2795301 on Build GRCh38
rs753359247 Chr.20: 2795691-2795691 on Build GRCh38
rs769992463 Chr.20: 2795678-2795680 on Build GRCh38
rs781760114 Chr.20: 2795555-2795556 on Build GRCh38
rs1011528614 Chr.20: 2795529-2795529 on Build GRCh38
rs753793658 Chr.20: 2795401-2795401 on Build GRCh38
rs150695261 Chr.20: 2795346-2795346 on Build GRCh38
rs752838139 Chr.20: 2795575-2795575 on Build GRCh38
rs767923118 Chr.20: 2795681-2795681 on Build GRCh38
rs201409887 Chr.20: 2795688-2795688 on Build GRCh38
rs747803655 Chr.20: 2795699-2795699 on Build GRCh38
rs371175094 Chr.20: 2795319-2795319 on Build GRCh38
rs781462252 Chr.20: 2795610-2795610 on Build GRCh38
rs759094843 Chr.20: 2795323-2795323 on Build GRCh38
rs755116809 Chr.20: 2795414-2795414 on Build GRCh38
rs142650829 Chr.20: 2795458-2795458 on Build GRCh38
rs768357907 Chr.20: 2795630-2795630 on Build GRCh38
rs548047354 Chr.20: 2795472-2795472 on Build GRCh38

View Full Product Details